Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40334883-40335377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf52
Entrez Gene ID
388115
Gene Name
chromosome 15 open reading frame 52
Gene Aliases
-
UniGene
Hs.32433
Gene Chromosome Location
Chr.15: 40331452-40340967 on Build GRCh38
Transcripts
NM_207380.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4884890 c.1514G>T p.S505I Chr.15: 40335249-40335249 on Build GRCh38
6554883 c.802C>A p.P268T Chr.15: 40335331-40335331 on Build GRCh38
1372639 c.1523C>T p.T508M Chr.15: 40335240-40335240 on Build GRCh38
4846479 c.1389G>C p.W463C Chr.15: 40335374-40335374 on Build GRCh38
6376663 c.956G>T p.G319V Chr.15: 40335177-40335177 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs533392306 Chr.15: 40335021-40335021 on Build GRCh38
rs770216748 Chr.15: 40335186-40335186 on Build GRCh38
rs183595585 Chr.15: 40334945-40334945 on Build GRCh38
rs768986155 Chr.15: 40335241-40335241 on Build GRCh38
rs376941842 Chr.15: 40335129-40335129 on Build GRCh38
rs746197395 Chr.15: 40335356-40335356 on Build GRCh38
rs374965872 Chr.15: 40334904-40334904 on Build GRCh38
rs545140671 Chr.15: 40334976-40334976 on Build GRCh38
rs771177717 Chr.15: 40335254-40335254 on Build GRCh38
rs771984214 Chr.15: 40334971-40334971 on Build GRCh38
rs955666416 Chr.15: 40335032-40335032 on Build GRCh38
rs142452705 Chr.15: 40335173-40335173 on Build GRCh38
rs369415884 Chr.15: 40335110-40335110 on Build GRCh38
rs945088275 Chr.15: 40335040-40335040 on Build GRCh38
rs894664120 Chr.15: 40335191-40335191 on Build GRCh38
rs760423120 Chr.15: 40335204-40335204 on Build GRCh38
rs916362591 Chr.15: 40335352-40335352 on Build GRCh38
rs781424201 Chr.15: 40335309-40335309 on Build GRCh38
rs1008328669 Chr.15: 40335055-40335055 on Build GRCh38
rs989754933 Chr.15: 40335278-40335278 on Build GRCh38
rs747299842 Chr.15: 40335180-40335180 on Build GRCh38
rs372473196 Chr.15: 40335153-40335153 on Build GRCh38
rs372826018 Chr.15: 40335300-40335300 on Build GRCh38
rs930168121 Chr.15: 40334941-40334941 on Build GRCh38
rs952696741 Chr.15: 40335096-40335096 on Build GRCh38
rs776974702 Chr.15: 40335127-40335127 on Build GRCh38
rs983379932 Chr.15: 40335097-40335097 on Build GRCh38
rs1035897217 Chr.15: 40334929-40334929 on Build GRCh38
rs372988249 Chr.15: 40335308-40335308 on Build GRCh38
rs775795228 Chr.15: 40335116-40335116 on Build GRCh38
rs572485964 Chr.15: 40335240-40335240 on Build GRCh38
rs774734344 Chr.15: 40335343-40335343 on Build GRCh38
rs762359041 Chr.15: 40335128-40335128 on Build GRCh38
rs768598403 Chr.15: 40335181-40335181 on Build GRCh38
rs141713705 Chr.15: 40334989-40334989 on Build GRCh38
rs146699813 Chr.15: 40335247-40335247 on Build GRCh38
rs200868200 Chr.15: 40335246-40335246 on Build GRCh38
rs376572354 Chr.15: 40335219-40335219 on Build GRCh38
rs764269177 Chr.15: 40335125-40335125 on Build GRCh38
rs538441829 Chr.15: 40335239-40335239 on Build GRCh38
rs753911947 Chr.15: 40335301-40335302 on Build GRCh38
rs201431730 Chr.15: 40335188-40335188 on Build GRCh38
rs974334596 Chr.15: 40334920-40334920 on Build GRCh38
rs1004196352 Chr.15: 40335208-40335208 on Build GRCh38
rs148064363 Chr.15: 40334961-40334961 on Build GRCh38
rs193099892 Chr.15: 40335089-40335089 on Build GRCh38
rs547738234 Chr.15: 40335088-40335088 on Build GRCh38
rs151308893 Chr.15: 40335183-40335183 on Build GRCh38
rs752416206 Chr.15: 40335145-40335145 on Build GRCh38
rs750851571 Chr.15: 40335130-40335130 on Build GRCh38
rs1056512857 Chr.15: 40334957-40334957 on Build GRCh38
rs570603935 Chr.15: 40335146-40335146 on Build GRCh38
rs555662079 Chr.15: 40335017-40335017 on Build GRCh38
rs761368609 Chr.15: 40335275-40335275 on Build GRCh38
rs139346626 Chr.15: 40335195-40335195 on Build GRCh38
rs760628332 Chr.15: 40335156-40335156 on Build GRCh38
rs112466704 Chr.15: 40335074-40335074 on Build GRCh38
rs759824874 Chr.15: 40335265-40335265 on Build GRCh38
rs773718356 Chr.15: 40335192-40335192 on Build GRCh38
rs1031069192 Chr.15: 40335016-40335016 on Build GRCh38
rs556409470 Chr.15: 40334927-40334927 on Build GRCh38
rs947761258 Chr.15: 40335144-40335144 on Build GRCh38
rs757332648 Chr.15: 40335172-40335172 on Build GRCh38
rs558204056 Chr.15: 40335255-40335255 on Build GRCh38
rs764883604 Chr.15: 40335291-40335291 on Build GRCh38
rs555755039 Chr.15: 40335221-40335221 on Build GRCh38
rs140554165 Chr.15: 40335122-40335122 on Build GRCh38
rs1043614440 Chr.15: 40335149-40335149 on Build GRCh38
rs781273348 Chr.15: 40335242-40335242 on Build GRCh38
rs939081807 Chr.15: 40335335-40335335 on Build GRCh38
rs894707363 Chr.15: 40335087-40335087 on Build GRCh38
rs140264738 Chr.15: 40335301-40335301 on Build GRCh38
rs766796608 Chr.15: 40335198-40335198 on Build GRCh38
rs369701147 Chr.15: 40335184-40335184 on Build GRCh38
rs769832239 Chr.15: 40335319-40335319 on Build GRCh38
rs556433174 Chr.15: 40335349-40335349 on Build GRCh38
rs1015630386 Chr.15: 40335213-40335213 on Build GRCh38
rs545869746 Chr.15: 40335042-40335042 on Build GRCh38
rs144372797 Chr.15: 40335218-40335218 on Build GRCh38
rs553275907 Chr.15: 40334936-40334937 on Build GRCh38
rs369154599 Chr.15: 40335142-40335142 on Build GRCh38
rs748164206 Chr.15: 40335243-40335243 on Build GRCh38
rs750420083 Chr.15: 40335227-40335227 on Build GRCh38
rs539289289 Chr.15: 40335151-40335151 on Build GRCh38
rs147193136 Chr.15: 40335068-40335068 on Build GRCh38
rs775301727 Chr.15: 40335058-40335058 on Build GRCh38
rs777877148 Chr.15: 40335322-40335322 on Build GRCh38
rs199502649 Chr.15: 40335339-40335339 on Build GRCh38
rs769881318 Chr.15: 40335245-40335245 on Build GRCh38
rs772354058 Chr.15: 40335115-40335115 on Build GRCh38
rs145243262 Chr.15: 40335302-40335302 on Build GRCh38
rs564677335 Chr.15: 40335014-40335014 on Build GRCh38
rs368027083 Chr.15: 40334946-40334946 on Build GRCh38
rs749781667 Chr.15: 40335337-40335337 on Build GRCh38
rs1018259517 Chr.15: 40335057-40335057 on Build GRCh38
rs964047017 Chr.15: 40335067-40335067 on Build GRCh38
rs749157219 Chr.15: 40335154-40335154 on Build GRCh38
rs772016566 Chr.15: 40335174-40335174 on Build GRCh38
rs758762768 Chr.15: 40335141-40335141 on Build GRCh38
rs1047467039 Chr.15: 40334942-40334942 on Build GRCh38
rs753569582 Chr.15: 40335270-40335270 on Build GRCh38
rs763745528 Chr.15: 40335267-40335267 on Build GRCh38
rs750366646 Chr.15: 40335292-40335292 on Build GRCh38
rs908340225 Chr.15: 40334956-40334956 on Build GRCh38
rs748397358 Chr.15: 40335314-40335314 on Build GRCh38
rs188495379 Chr.15: 40334965-40334965 on Build GRCh38

View Full Product Details