Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40336432-40336932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf52
Entrez Gene ID
388115
Gene Name
chromosome 15 open reading frame 52
Gene Aliases
-
UniGene
Hs.32433
Gene Chromosome Location
Chr.15: 40331452-40340967 on Build GRCh38
Transcripts
NM_207380.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4938917 c.894A>G p.K298K Chr.15: 40336794-40336794 on Build GRCh38
6614428 c.997A>C p.R333R Chr.15: 40336596-40336596 on Build GRCh38
1372641 c.900G>A p.Q300Q Chr.15: 40336788-40336788 on Build GRCh38
7229255 c.937G>A p.G313R Chr.15: 40336656-40336656 on Build GRCh38
6369435 c.362A>T p.K121M Chr.15: 40336601-40336601 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1035989619 Chr.15: 40336884-40336884 on Build GRCh38
rs749505310 Chr.15: 40336557-40336557 on Build GRCh38
rs746467935 Chr.15: 40336539-40336539 on Build GRCh38
rs764569483 Chr.15: 40336525-40336525 on Build GRCh38
rs377283083 Chr.15: 40336739-40336739 on Build GRCh38
rs562536344 Chr.15: 40336912-40336912 on Build GRCh38
rs760005239 Chr.15: 40336835-40336835 on Build GRCh38
rs1027961482 Chr.15: 40336859-40336859 on Build GRCh38
rs1028223128 Chr.15: 40336585-40336585 on Build GRCh38
rs559510099 Chr.15: 40336867-40336867 on Build GRCh38
rs143912259 Chr.15: 40336794-40336794 on Build GRCh38
rs763426354 Chr.15: 40336589-40336589 on Build GRCh38
rs775838823 Chr.15: 40336581-40336581 on Build GRCh38
rs1045372741 Chr.15: 40336653-40336653 on Build GRCh38
rs752561012 Chr.15: 40336630-40336630 on Build GRCh38
rs755023679 Chr.15: 40336846-40336846 on Build GRCh38
rs759287482 Chr.15: 40336687-40336687 on Build GRCh38
rs368116601 Chr.15: 40336662-40336662 on Build GRCh38
rs777855939 Chr.15: 40336635-40336635 on Build GRCh38
rs200782061 Chr.15: 40336679-40336679 on Build GRCh38
rs370895955 Chr.15: 40336577-40336577 on Build GRCh38
rs764363675 Chr.15: 40336583-40336583 on Build GRCh38
rs200126169 Chr.15: 40336607-40336607 on Build GRCh38
rs370005721 Chr.15: 40336743-40336743 on Build GRCh38
rs751145680 Chr.15: 40336680-40336680 on Build GRCh38
rs146973724 Chr.15: 40336608-40336608 on Build GRCh38
rs747064803 Chr.15: 40336729-40336729 on Build GRCh38
rs754543566 Chr.15: 40336542-40336542 on Build GRCh38
rs141368637 Chr.15: 40336544-40336544 on Build GRCh38
rs376584935 Chr.15: 40336702-40336702 on Build GRCh38
rs750177087 Chr.15: 40336820-40336820 on Build GRCh38
rs780799483 Chr.15: 40336628-40336628 on Build GRCh38
rs761304163 Chr.15: 40336582-40336582 on Build GRCh38
rs1005427902 Chr.15: 40336908-40336908 on Build GRCh38
rs758115534 Chr.15: 40336537-40336537 on Build GRCh38
rs749032102 Chr.15: 40336637-40336637 on Build GRCh38
rs765272093 Chr.15: 40336818-40336818 on Build GRCh38
rs370442829 Chr.15: 40336773-40336773 on Build GRCh38
rs777096458 Chr.15: 40336740-40336740 on Build GRCh38
rs781303978 Chr.15: 40336851-40336851 on Build GRCh38
rs924012464 Chr.15: 40336567-40336567 on Build GRCh38
rs780472606 Chr.15: 40336733-40336733 on Build GRCh38
rs201560388 Chr.15: 40336778-40336778 on Build GRCh38
rs749515396 Chr.15: 40336554-40336554 on Build GRCh38
rs765970665 Chr.15: 40336604-40336604 on Build GRCh38
rs557258914 Chr.15: 40336509-40336509 on Build GRCh38
rs757332488 Chr.15: 40336817-40336817 on Build GRCh38
rs757096139 Chr.15: 40336707-40336707 on Build GRCh38
rs367840861 Chr.15: 40336704-40336704 on Build GRCh38
rs377691451 Chr.15: 40336822-40336822 on Build GRCh38
rs370144055 Chr.15: 40336538-40336538 on Build GRCh38
rs758775313 Chr.15: 40336531-40336534 on Build GRCh38
rs1043748893 Chr.15: 40336560-40336560 on Build GRCh38
rs762181230 Chr.15: 40336594-40336594 on Build GRCh38
rs563889496 Chr.15: 40336657-40336657 on Build GRCh38
rs770871955 Chr.15: 40336641-40336641 on Build GRCh38
rs746074054 Chr.15: 40336710-40336710 on Build GRCh38
rs769096017 Chr.15: 40336667-40336667 on Build GRCh38
rs773197120 Chr.15: 40336550-40336550 on Build GRCh38
rs200667931 Chr.15: 40336543-40336543 on Build GRCh38
rs772271858 Chr.15: 40336724-40336724 on Build GRCh38
rs753814378 Chr.15: 40336814-40336814 on Build GRCh38
rs771209616 Chr.15: 40336568-40336568 on Build GRCh38
rs371905877 Chr.15: 40336701-40336701 on Build GRCh38
rs12911108 Chr.15: 40336470-40336470 on Build GRCh38
rs774726809 Chr.15: 40336575-40336575 on Build GRCh38
rs756140701 Chr.15: 40336632-40336632 on Build GRCh38
rs202100643 Chr.15: 40336728-40336728 on Build GRCh38
rs200478802 Chr.15: 40336694-40336694 on Build GRCh38
rs760221617 Chr.15: 40336789-40336789 on Build GRCh38
rs775026611 Chr.15: 40336787-40336787 on Build GRCh38
rs891968769 Chr.15: 40336533-40336533 on Build GRCh38
rs888116621 Chr.15: 40336907-40336907 on Build GRCh38
rs138396559 Chr.15: 40336656-40336656 on Build GRCh38
rs985673585 Chr.15: 40336717-40336717 on Build GRCh38
rs767675571 Chr.15: 40336578-40336578 on Build GRCh38
rs1014632160 Chr.15: 40336666-40336666 on Build GRCh38
rs750063847 Chr.15: 40336536-40336536 on Build GRCh38
rs532200169 Chr.15: 40336497-40336497 on Build GRCh38
rs769450654 Chr.15: 40336549-40336549 on Build GRCh38
rs777149549 Chr.15: 40336668-40336668 on Build GRCh38
rs1053784035 Chr.15: 40336744-40336744 on Build GRCh38
rs747239067 Chr.15: 40336845-40336845 on Build GRCh38
rs767137937 Chr.15: 40336779-40336779 on Build GRCh38
rs747508438 Chr.15: 40336844-40336844 on Build GRCh38
rs779236778 Chr.15: 40336645-40336645 on Build GRCh38
rs573879759 Chr.15: 40336523-40336523 on Build GRCh38
rs373181231 Chr.15: 40336751-40336751 on Build GRCh38
rs778909877 Chr.15: 40336709-40336709 on Build GRCh38
rs753857383 Chr.15: 40336706-40336706 on Build GRCh38
rs758153228 Chr.15: 40336829-40336829 on Build GRCh38
rs754137129 Chr.15: 40336586-40336586 on Build GRCh38

View Full Product Details