Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43775391-43775890 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELL3
Entrez Gene ID
80237
Gene Name
elongation factor for RNA polymerase II 3
Gene Aliases
-
UniGene
Hs.706346
Gene Chromosome Location
Chr.15: 43772600-43777304 on Build GRCh38
Transcripts
NM_025165.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6905029 c.368C>T p.S123L Chr.15: 43775837-43775837 on Build GRCh38
4187671 c.329T>G p.L110R Chr.15: 43775876-43775876 on Build GRCh38
3361411 c.558G>A p.Q186Q Chr.15: 43775536-43775536 on Build GRCh38
280772 c.439T>C p.S147P Chr.15: 43775766-43775766 on Build GRCh38
962065 c.325C>T p.R109C Chr.15: 43775880-43775880 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369172951 Chr.15: 43775480-43775480 on Build GRCh38
rs778743007 Chr.15: 43775658-43775658 on Build GRCh38
rs767665514 Chr.15: 43775765-43775765 on Build GRCh38
rs982638159 Chr.15: 43775545-43775545 on Build GRCh38
rs770900024 Chr.15: 43775419-43775419 on Build GRCh38
rs751833506 Chr.15: 43775631-43775631 on Build GRCh38
rs758385778 Chr.15: 43775788-43775788 on Build GRCh38
rs1055674381 Chr.15: 43775672-43775672 on Build GRCh38
rs764290325 Chr.15: 43775857-43775857 on Build GRCh38
rs1016837640 Chr.15: 43775838-43775838 on Build GRCh38
rs771558827 Chr.15: 43775737-43775737 on Build GRCh38
rs780729535 Chr.15: 43775817-43775817 on Build GRCh38
rs963543797 Chr.15: 43775443-43775443 on Build GRCh38
rs778880317 Chr.15: 43775585-43775585 on Build GRCh38
rs933696109 Chr.15: 43775647-43775648 on Build GRCh38
rs372579644 Chr.15: 43775491-43775491 on Build GRCh38
rs780192751 Chr.15: 43775779-43775780 on Build GRCh38
rs918311101 Chr.15: 43775476-43775476 on Build GRCh38
rs754758260 Chr.15: 43775815-43775815 on Build GRCh38
rs146412353 Chr.15: 43775473-43775473 on Build GRCh38
rs761028156 Chr.15: 43775855-43775855 on Build GRCh38
rs35454865 Chr.15: 43775787-43775787 on Build GRCh38
rs774959695 Chr.15: 43775844-43775844 on Build GRCh38
rs909782821 Chr.15: 43775563-43775563 on Build GRCh38
rs764170894 Chr.15: 43775771-43775771 on Build GRCh38
rs767759116 Chr.15: 43775643-43775643 on Build GRCh38
rs766277984 Chr.15: 43775799-43775799 on Build GRCh38
rs779697747 Chr.15: 43775607-43775607 on Build GRCh38
rs750799344 Chr.15: 43775496-43775496 on Build GRCh38
rs754632661 Chr.15: 43775691-43775691 on Build GRCh38
rs752761104 Chr.15: 43775644-43775644 on Build GRCh38
rs909458385 Chr.15: 43775435-43775435 on Build GRCh38
rs760626651 Chr.15: 43775719-43775719 on Build GRCh38
rs369931248 Chr.15: 43775652-43775652 on Build GRCh38
rs766790843 Chr.15: 43775626-43775626 on Build GRCh38
rs531883967 Chr.15: 43775683-43775683 on Build GRCh38
rs532357510 Chr.15: 43775850-43775850 on Build GRCh38
rs138259199 Chr.15: 43775821-43775821 on Build GRCh38
rs768290291 Chr.15: 43775610-43775610 on Build GRCh38
rs982564314 Chr.15: 43775837-43775837 on Build GRCh38
rs755120703 Chr.15: 43775543-43775543 on Build GRCh38
rs745655619 Chr.15: 43775422-43775422 on Build GRCh38
rs763374753 Chr.15: 43775498-43775498 on Build GRCh38
rs774906694 Chr.15: 43775742-43775742 on Build GRCh38
rs766331099 Chr.15: 43775868-43775868 on Build GRCh38
rs766739456 Chr.15: 43775537-43775537 on Build GRCh38
rs776738443 Chr.15: 43775858-43775858 on Build GRCh38
rs199935246 Chr.15: 43775804-43775804 on Build GRCh38
rs768099954 Chr.15: 43775488-43775488 on Build GRCh38
rs201197535 Chr.15: 43775551-43775551 on Build GRCh38
rs761359622 Chr.15: 43775617-43775617 on Build GRCh38
rs775088692 Chr.15: 43775433-43775433 on Build GRCh38
rs894566244 Chr.15: 43775803-43775803 on Build GRCh38
rs202152081 Chr.15: 43775823-43775823 on Build GRCh38
rs745697910 Chr.15: 43775586-43775586 on Build GRCh38
rs747639403 Chr.15: 43775713-43775713 on Build GRCh38
rs773868260 Chr.15: 43775721-43775721 on Build GRCh38
rs748210585 Chr.15: 43775789-43775790 on Build GRCh38
rs375557328 Chr.15: 43775565-43775565 on Build GRCh38
rs778770300 Chr.15: 43775509-43775510 on Build GRCh38
rs776059471 Chr.15: 43775614-43775614 on Build GRCh38
rs370565230 Chr.15: 43775818-43775818 on Build GRCh38
rs140782164 Chr.15: 43775687-43775687 on Build GRCh38
rs775733605 Chr.15: 43775768-43775768 on Build GRCh38
rs997636421 Chr.15: 43775745-43775745 on Build GRCh38
rs750306906 Chr.15: 43775679-43775679 on Build GRCh38
rs746790926 Chr.15: 43775688-43775688 on Build GRCh38
rs773728364 Chr.15: 43775618-43775618 on Build GRCh38
rs775583137 Chr.15: 43775854-43775854 on Build GRCh38
rs369771946 Chr.15: 43775625-43775625 on Build GRCh38
rs771751830 Chr.15: 43775427-43775427 on Build GRCh38
rs771808909 Chr.15: 43775604-43775604 on Build GRCh38
rs373378172 Chr.15: 43775697-43775697 on Build GRCh38
rs749745166 Chr.15: 43775733-43775733 on Build GRCh38
rs77744401 Chr.15: 43775748-43775748 on Build GRCh38
rs368627967 Chr.15: 43775859-43775859 on Build GRCh38
rs757191401 Chr.15: 43775580-43775580 on Build GRCh38
rs745751156 Chr.15: 43775588-43775589 on Build GRCh38
rs759949645 Chr.15: 43775746-43775746 on Build GRCh38
rs375110222 Chr.15: 43775420-43775420 on Build GRCh38
rs765758493 Chr.15: 43775492-43775492 on Build GRCh38
rs746241390 Chr.15: 43775848-43775848 on Build GRCh38
rs992677007 Chr.15: 43775434-43775434 on Build GRCh38
rs373227136 Chr.15: 43775639-43775639 on Build GRCh38
rs772427384 Chr.15: 43775849-43775849 on Build GRCh38
rs552668420 Chr.15: 43775656-43775657 on Build GRCh38
rs972217989 Chr.15: 43775459-43775459 on Build GRCh38
rs760687773 Chr.15: 43775769-43775769 on Build GRCh38
rs751782159 Chr.15: 43775538-43775538 on Build GRCh38
rs142769297 Chr.15: 43775832-43775832 on Build GRCh38
rs746600602 Chr.15: 43775608-43775608 on Build GRCh38
rs748786821 Chr.15: 43775826-43775826 on Build GRCh38

View Full Product Details