Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43794688-43795195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERF2
Entrez Gene ID
10169
Gene Name
small EDRK-rich factor 2
Gene Aliases
4F5REL, FAM2C, H4F5REL, HsT17089
UniGene
Hs.424126
Gene Chromosome Location
Chr.15: 43777096-43796089 on Build GRCh38
Transcripts
NM_001018108.3 NM_001199875.1 NM_001199876.1 NM_001199877.1 NM_001199878.1 NR_037672.1 NR_037673.1 NM_001258031.1 NM_001258032.1

Gene Symbol
SERF2-C15ORF63
Entrez Gene ID
100529067
Gene Name
SERF2-C15orf63 readthrough
Gene Aliases
-
UniGene
Hs.730672
Gene Chromosome Location
Chr.15: 43791976-43802589 on Build GRCh38
Transcripts
NM_001018108.3 NM_001199875.1 NM_001199876.1 NM_001199877.1 NM_001199878.1 NR_037672.1 NR_037673.1 NM_001258031.1 NM_001258032.1

Gene Symbol
SERINC4
Entrez Gene ID
619189
Gene Name
serine incorporator 4
Gene Aliases
-
UniGene
Hs.424126
Gene Chromosome Location
Chr.15: 43794174-43800097 on Build GRCh38
Transcripts
NM_001018108.3 NM_001199875.1 NM_001199876.1 NM_001199877.1 NM_001199878.1 NR_037672.1 NR_037673.1 NM_001258031.1 NM_001258032.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6257357 c.642C>A p.F214L Chr.15: 43795183-43795183 on Build GRCh38
235330 c.711G>T p.L237L Chr.15: 43795114-43795114 on Build GRCh38
4054882 c.1514G>A p.R505H Chr.15: 43795043-43795043 on Build GRCh38
263584 c.729A>G p.P243P Chr.15: 43795096-43795096 on Build GRCh38
3727650 c.1382G>A p.G461D Chr.15: 43795175-43795175 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145705656 Chr.15: 43795044-43795044 on Build GRCh38
rs769313998 Chr.15: 43794958-43794958 on Build GRCh38
rs754483746 Chr.15: 43795043-43795043 on Build GRCh38
rs772352476 Chr.15: 43795167-43795168 on Build GRCh38
rs910727618 Chr.15: 43794726-43794726 on Build GRCh38
rs549760199 Chr.15: 43794882-43794882 on Build GRCh38
rs368021438 Chr.15: 43795165-43795165 on Build GRCh38
rs552453789 Chr.15: 43794831-43794831 on Build GRCh38
rs574879721 Chr.15: 43795038-43795038 on Build GRCh38
rs143486742 Chr.15: 43795018-43795018 on Build GRCh38
rs192719626 Chr.15: 43794742-43794742 on Build GRCh38
rs764346378 Chr.15: 43795016-43795016 on Build GRCh38
rs151106509 Chr.15: 43795089-43795089 on Build GRCh38
rs576323903 Chr.15: 43795004-43795004 on Build GRCh38
rs143808872 Chr.15: 43795050-43795050 on Build GRCh38
rs987677598 Chr.15: 43794720-43794720 on Build GRCh38
rs963757034 Chr.15: 43794776-43794776 on Build GRCh38
rs187951529 Chr.15: 43794969-43794969 on Build GRCh38
rs368967498 Chr.15: 43795047-43795047 on Build GRCh38
rs1056857418 Chr.15: 43794892-43794892 on Build GRCh38
rs567696997 Chr.15: 43794900-43794900 on Build GRCh38
rs981114582 Chr.15: 43794828-43794828 on Build GRCh38
rs992720015 Chr.15: 43795118-43795118 on Build GRCh38
rs772205377 Chr.15: 43795070-43795070 on Build GRCh38
rs369732267 Chr.15: 43795040-43795040 on Build GRCh38
rs372775523 Chr.15: 43794954-43794954 on Build GRCh38
rs919669117 Chr.15: 43795081-43795081 on Build GRCh38
rs1040393416 Chr.15: 43794918-43794918 on Build GRCh38
rs192035160 Chr.15: 43795041-43795041 on Build GRCh38
rs778103282 Chr.15: 43795121-43795121 on Build GRCh38
rs374113255 Chr.15: 43795140-43795140 on Build GRCh38
rs146664766 Chr.15: 43795108-43795108 on Build GRCh38
rs908182437 Chr.15: 43794829-43794829 on Build GRCh38
rs751982774 Chr.15: 43794888-43794888 on Build GRCh38
rs773087720 Chr.15: 43795083-43795083 on Build GRCh38
rs754998121 Chr.15: 43794957-43794958 on Build GRCh38
rs757346933 Chr.15: 43795055-43795059 on Build GRCh38
rs567698201 Chr.15: 43794740-43794740 on Build GRCh38
rs569110366 Chr.15: 43794840-43794840 on Build GRCh38
rs764032205 Chr.15: 43795022-43795022 on Build GRCh38
rs755103256 Chr.15: 43794896-43794896 on Build GRCh38
rs113812701 Chr.15: 43795126-43795126 on Build GRCh38
rs376787676 Chr.15: 43795078-43795078 on Build GRCh38
rs370605763 Chr.15: 43795125-43795125 on Build GRCh38
rs903149602 Chr.15: 43794934-43794934 on Build GRCh38
rs572392894 Chr.15: 43794795-43794795 on Build GRCh38
rs938601226 Chr.15: 43794891-43794891 on Build GRCh38
rs567095907 Chr.15: 43794778-43794778 on Build GRCh38
rs746466410 Chr.15: 43794955-43794955 on Build GRCh38
rs771038548 Chr.15: 43794995-43794995 on Build GRCh38
rs919562211 Chr.15: 43794813-43794813 on Build GRCh38
rs531821428 Chr.15: 43795129-43795129 on Build GRCh38
rs971500728 Chr.15: 43794816-43794816 on Build GRCh38
rs375421621 Chr.15: 43795076-43795076 on Build GRCh38
rs973756953 Chr.15: 43794844-43794845 on Build GRCh38
rs908211842 Chr.15: 43795098-43795098 on Build GRCh38
rs756727241 Chr.15: 43795120-43795120 on Build GRCh38
rs910686594 Chr.15: 43795130-43795130 on Build GRCh38
rs554027040 Chr.15: 43794912-43794912 on Build GRCh38
rs774277001 Chr.15: 43795168-43795168 on Build GRCh38
rs753706823 Chr.15: 43795023-43795023 on Build GRCh38
rs766074141 Chr.15: 43795091-43795091 on Build GRCh38
rs777667165 Chr.15: 43795036-43795041 on Build GRCh38
rs778250947 Chr.15: 43795049-43795049 on Build GRCh38
rs920763946 Chr.15: 43794856-43794858 on Build GRCh38
rs931019896 Chr.15: 43794832-43794832 on Build GRCh38
rs771216898 Chr.15: 43795057-43795057 on Build GRCh38
rs112335120 Chr.15: 43794980-43794980 on Build GRCh38
rs149013820 Chr.15: 43795046-43795046 on Build GRCh38
rs752235354 Chr.15: 43795110-43795110 on Build GRCh38
rs781239638 Chr.15: 43794906-43794906 on Build GRCh38
rs747047754 Chr.15: 43795141-43795141 on Build GRCh38
rs772256664 Chr.15: 43795132-43795132 on Build GRCh38
rs773710636 Chr.15: 43794985-43794985 on Build GRCh38
rs143602754 Chr.15: 43795037-43795037 on Build GRCh38
rs1012744412 Chr.15: 43795011-43795011 on Build GRCh38
rs755434251 Chr.15: 43795067-43795068 on Build GRCh38
rs774531261 Chr.15: 43794999-43794999 on Build GRCh38
rs138021432 Chr.15: 43795162-43795162 on Build GRCh38
rs948780561 Chr.15: 43794940-43794940 on Build GRCh38
rs548863288 Chr.15: 43795079-43795079 on Build GRCh38
rs930808404 Chr.15: 43794875-43794875 on Build GRCh38
rs779182068 Chr.15: 43795126-43795127 on Build GRCh38
rs371045123 Chr.15: 43795065-43795065 on Build GRCh38
rs577023776 Chr.15: 43794922-43794922 on Build GRCh38
rs757496538 Chr.15: 43795128-43795128 on Build GRCh38
rs142556047 Chr.15: 43795094-43795094 on Build GRCh38
rs375681174 Chr.15: 43795039-43795039 on Build GRCh38
rs770220121 Chr.15: 43794984-43794984 on Build GRCh38
rs185398419 Chr.15: 43794808-43794808 on Build GRCh38
rs373214024 Chr.15: 43795054-43795054 on Build GRCh38
rs537519374 Chr.15: 43794869-43794869 on Build GRCh38
rs991444346 Chr.15: 43794755-43794757 on Build GRCh38
rs763295298 Chr.15: 43794992-43794992 on Build GRCh38
rs552855311 Chr.15: 43795066-43795066 on Build GRCh38
rs1043170181 Chr.15: 43794964-43794964 on Build GRCh38
rs769788389 Chr.15: 43795155-43795155 on Build GRCh38
rs533590161 Chr.15: 43794902-43794902 on Build GRCh38
rs1050669355 Chr.15: 43794904-43794904 on Build GRCh38
rs746103485 Chr.15: 43795131-43795132 on Build GRCh38
rs369518718 Chr.15: 43795041-43795046 on Build GRCh38
rs761732921 Chr.15: 43795028-43795028 on Build GRCh38

View Full Product Details