Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 44490580-44491114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTDSPL2
Entrez Gene ID
51496
Gene Name
CTD small phosphatase like 2
Gene Aliases
HSPC058, HSPC129
UniGene
Hs.497967
Gene Chromosome Location
Chr.15: 44427329-44527774 on Build GRCh38
Transcripts
NM_016396.2 XM_005254441.2 XM_006720565.2 XM_011521665.2 XM_017022306.1 XM_017022307.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6404167 c.671G>A p.R224H Chr.15: 44490979-44490979 on Build GRCh38
3706592 c.656A>C p.E219A Chr.15: 44490964-44490964 on Build GRCh38
1608408 c.691+3G>T p.? Chr.15: 44491002-44491002 on Build GRCh38
3816170 c.670C>T p.R224C Chr.15: 44490978-44490978 on Build GRCh38
4833593 c.534A>G p.Q178Q Chr.15: 44490842-44490842 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151090996 Chr.15: 44491062-44491062 on Build GRCh38
rs771280790 Chr.15: 44490946-44490946 on Build GRCh38
rs374115658 Chr.15: 44490963-44490963 on Build GRCh38
rs966323206 Chr.15: 44490711-44490711 on Build GRCh38
rs143796729 Chr.15: 44490976-44490976 on Build GRCh38
rs558812327 Chr.15: 44490991-44490991 on Build GRCh38
rs746517109 Chr.15: 44490962-44490962 on Build GRCh38
rs984618838 Chr.15: 44490753-44490753 on Build GRCh38
rs892105710 Chr.15: 44490618-44490618 on Build GRCh38
rs371327729 Chr.15: 44490744-44490744 on Build GRCh38
rs763329991 Chr.15: 44490793-44490793 on Build GRCh38
rs774669228 Chr.15: 44490948-44490948 on Build GRCh38
rs1031076560 Chr.15: 44490671-44490671 on Build GRCh38
rs755439766 Chr.15: 44491010-44491014 on Build GRCh38
rs567332994 Chr.15: 44490710-44490710 on Build GRCh38
rs746465774 Chr.15: 44490761-44490761 on Build GRCh38
rs79423674 Chr.15: 44490657-44490657 on Build GRCh38
rs149011224 Chr.15: 44490679-44490679 on Build GRCh38
rs1056224185 Chr.15: 44490993-44490993 on Build GRCh38
rs1056500921 Chr.15: 44490859-44490859 on Build GRCh38
rs544558634 Chr.15: 44491078-44491078 on Build GRCh38
rs748680681 Chr.15: 44491043-44491044 on Build GRCh38
rs754395573 Chr.15: 44490909-44490909 on Build GRCh38
rs367955013 Chr.15: 44490967-44490967 on Build GRCh38
rs775639601 Chr.15: 44490957-44490957 on Build GRCh38
rs765009326 Chr.15: 44490870-44490870 on Build GRCh38
rs751260874 Chr.15: 44490978-44490978 on Build GRCh38
rs75179450 Chr.15: 44490820-44490820 on Build GRCh38
rs757192550 Chr.15: 44490850-44490850 on Build GRCh38
rs1056157872 Chr.15: 44490614-44490614 on Build GRCh38
rs764416424 Chr.15: 44490627-44490627 on Build GRCh38
rs773676249 Chr.15: 44490788-44490788 on Build GRCh38
rs771475034 Chr.15: 44491020-44491020 on Build GRCh38
rs934826593 Chr.15: 44490785-44490785 on Build GRCh38
rs764970317 Chr.15: 44490613-44490613 on Build GRCh38
rs746147823 Chr.15: 44491030-44491030 on Build GRCh38
rs371137544 Chr.15: 44490949-44490949 on Build GRCh38
rs751839128 Chr.15: 44490763-44490763 on Build GRCh38
rs780730012 Chr.15: 44490910-44490910 on Build GRCh38
rs34612266 Chr.15: 44490806-44490806 on Build GRCh38
rs749694507 Chr.15: 44491002-44491002 on Build GRCh38
rs779598773 Chr.15: 44490759-44490759 on Build GRCh38
rs984780669 Chr.15: 44490748-44490748 on Build GRCh38
rs766673797 Chr.15: 44490753-44490755 on Build GRCh38
rs780633326 Chr.15: 44490985-44490985 on Build GRCh38
rs975038816 Chr.15: 44491087-44491087 on Build GRCh38
rs772325010 Chr.15: 44491032-44491032 on Build GRCh38
rs946382606 Chr.15: 44490624-44490624 on Build GRCh38
rs964096650 Chr.15: 44491086-44491086 on Build GRCh38
rs749603385 Chr.15: 44490938-44490938 on Build GRCh38
rs111427983 Chr.15: 44490740-44490740 on Build GRCh38
rs779393977 Chr.15: 44491022-44491022 on Build GRCh38
rs865835916 Chr.15: 44490972-44490972 on Build GRCh38
rs1048817139 Chr.15: 44490605-44490605 on Build GRCh38
rs758640490 Chr.15: 44490735-44490735 on Build GRCh38
rs761715308 Chr.15: 44490961-44490961 on Build GRCh38
rs778751700 Chr.15: 44490741-44490741 on Build GRCh38
rs750206776 Chr.15: 44490873-44490873 on Build GRCh38
rs772087408 Chr.15: 44490955-44490955 on Build GRCh38
rs750460927 Chr.15: 44490986-44490986 on Build GRCh38
rs758062180 Chr.15: 44490898-44490898 on Build GRCh38
rs748643122 Chr.15: 44490924-44490924 on Build GRCh38
rs779617362 Chr.15: 44490902-44490902 on Build GRCh38
rs904040089 Chr.15: 44490663-44490663 on Build GRCh38
rs778337108 Chr.15: 44490933-44490933 on Build GRCh38
rs759650841 Chr.15: 44490821-44490821 on Build GRCh38
rs745564737 Chr.15: 44490743-44490743 on Build GRCh38
rs781537962 Chr.15: 44491042-44491043 on Build GRCh38
rs879661423 Chr.15: 44490988-44490988 on Build GRCh38
rs892543897 Chr.15: 44491042-44491042 on Build GRCh38
rs776962075 Chr.15: 44490776-44490776 on Build GRCh38
rs753772537 Chr.15: 44490842-44490842 on Build GRCh38
rs537399555 Chr.15: 44490661-44490661 on Build GRCh38
rs886078938 Chr.15: 44490606-44490606 on Build GRCh38
rs199944005 Chr.15: 44490834-44490834 on Build GRCh38
rs753701913 Chr.15: 44490738-44490738 on Build GRCh38
rs754627394 Chr.15: 44490979-44490979 on Build GRCh38
rs896155526 Chr.15: 44490906-44490906 on Build GRCh38
rs373778549 Chr.15: 44490765-44490765 on Build GRCh38
rs376756628 Chr.15: 44490908-44490908 on Build GRCh38
rs766692694 Chr.15: 44490800-44490800 on Build GRCh38
rs551724124 Chr.15: 44490637-44490637 on Build GRCh38
rs1000921718 Chr.15: 44490666-44490666 on Build GRCh38
rs946350102 Chr.15: 44490851-44490851 on Build GRCh38
rs75936503 Chr.15: 44490817-44490817 on Build GRCh38
rs987200950 Chr.15: 44490681-44490681 on Build GRCh38
rs147437657 Chr.15: 44490787-44490787 on Build GRCh38
rs948279684 Chr.15: 44490965-44490965 on Build GRCh38
rs755621291 Chr.15: 44491051-44491051 on Build GRCh38

View Full Product Details