Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 45099329-45099849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUOX2
Entrez Gene ID
50506
Gene Name
dual oxidase 2
Gene Aliases
LNOX2, NOXEF2, P138-TOX, TDH6, THOX2
UniGene
Hs.71377
Gene Chromosome Location
Chr.15: 45092653-45114161 on Build GRCh38
Transcripts
NM_014080.4 XM_005254421.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1749171 c.3282C>G p.F1094L Chr.15: 45099795-45099795 on Build GRCh38
3501625 c.3251G>A p.R1084Q Chr.15: 45099826-45099826 on Build GRCh38
1749170 c.3289A>G p.I1097V Chr.15: 45099788-45099788 on Build GRCh38
4487808 c.3231C>T p.L1077L Chr.15: 45099846-45099846 on Build GRCh38
2153879 c.3268G>A p.V1090I Chr.15: 45099809-45099809 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1009807148 Chr.15: 45099474-45099475 on Build GRCh38
rs200459845 Chr.15: 45099389-45099389 on Build GRCh38
rs533512371 Chr.15: 45099438-45099438 on Build GRCh38
rs750241140 Chr.15: 45099681-45099681 on Build GRCh38
rs149907592 Chr.15: 45099769-45099769 on Build GRCh38
rs374660171 Chr.15: 45099403-45099403 on Build GRCh38
rs954731065 Chr.15: 45099505-45099505 on Build GRCh38
rs765163513 Chr.15: 45099680-45099680 on Build GRCh38
rs377720106 Chr.15: 45099460-45099460 on Build GRCh38
rs375243329 Chr.15: 45099744-45099744 on Build GRCh38
rs139115517 Chr.15: 45099764-45099764 on Build GRCh38
rs200344635 Chr.15: 45099776-45099776 on Build GRCh38
rs767782942 Chr.15: 45099702-45099702 on Build GRCh38
rs779924144 Chr.15: 45099734-45099735 on Build GRCh38
rs147719149 Chr.15: 45099762-45099762 on Build GRCh38
rs531536885 Chr.15: 45099749-45099749 on Build GRCh38
rs373924991 Chr.15: 45099658-45099658 on Build GRCh38
rs745608872 Chr.15: 45099474-45099474 on Build GRCh38
rs368975704 Chr.15: 45099827-45099827 on Build GRCh38
rs771715647 Chr.15: 45099388-45099388 on Build GRCh38
rs770819190 Chr.15: 45099360-45099360 on Build GRCh38
rs558919433 Chr.15: 45099826-45099826 on Build GRCh38
rs779150021 Chr.15: 45099708-45099708 on Build GRCh38
rs772253164 Chr.15: 45099760-45099760 on Build GRCh38
rs752785319 Chr.15: 45099669-45099669 on Build GRCh38
rs748194265 Chr.15: 45099737-45099737 on Build GRCh38
rs533557003 Chr.15: 45099809-45099809 on Build GRCh38
rs928951102 Chr.15: 45099650-45099650 on Build GRCh38
rs267604227 Chr.15: 45099406-45099406 on Build GRCh38
rs770266156 Chr.15: 45099689-45099689 on Build GRCh38
rs958531745 Chr.15: 45099534-45099534 on Build GRCh38
rs368652053 Chr.15: 45099699-45099699 on Build GRCh38
rs755674247 Chr.15: 45099789-45099789 on Build GRCh38
rs760172069 Chr.15: 45099399-45099399 on Build GRCh38
rs144923217 Chr.15: 45099770-45099770 on Build GRCh38
rs113632824 Chr.15: 45099710-45099710 on Build GRCh38
rs776772769 Chr.15: 45099817-45099817 on Build GRCh38
rs200000982 Chr.15: 45099449-45099449 on Build GRCh38
rs763092853 Chr.15: 45099429-45099429 on Build GRCh38
rs751311879 Chr.15: 45099728-45099728 on Build GRCh38
rs758318135 Chr.15: 45099418-45099420 on Build GRCh38
rs762702337 Chr.15: 45099712-45099712 on Build GRCh38
rs372219394 Chr.15: 45099711-45099711 on Build GRCh38
rs576035700 Chr.15: 45099612-45099612 on Build GRCh38
rs189585637 Chr.15: 45099682-45099682 on Build GRCh38
rs752091319 Chr.15: 45099737-45099737 on Build GRCh38
rs372199883 Chr.15: 45099804-45099804 on Build GRCh38
rs1032040322 Chr.15: 45099723-45099723 on Build GRCh38
rs768120439 Chr.15: 45099499-45099499 on Build GRCh38
rs760707357 Chr.15: 45099765-45099765 on Build GRCh38
rs139366597 Chr.15: 45099535-45099535 on Build GRCh38
rs757212555 Chr.15: 45099462-45099462 on Build GRCh38
rs766569538 Chr.15: 45099430-45099430 on Build GRCh38
rs752327099 Chr.15: 45099780-45099780 on Build GRCh38
rs759378295 Chr.15: 45099822-45099822 on Build GRCh38
rs766216114 Chr.15: 45099726-45099726 on Build GRCh38
rs145586290 Chr.15: 45099730-45099730 on Build GRCh38
rs567703268 Chr.15: 45099415-45099415 on Build GRCh38
rs886051191 Chr.15: 45099404-45099404 on Build GRCh38
rs112152834 Chr.15: 45099368-45099368 on Build GRCh38
rs1043807777 Chr.15: 45099701-45099701 on Build GRCh38
rs777375808 Chr.15: 45099793-45099793 on Build GRCh38
rs777115289 Chr.15: 45099401-45099401 on Build GRCh38
rs755622327 Chr.15: 45099732-45099732 on Build GRCh38
rs111276476 Chr.15: 45099559-45099559 on Build GRCh38
rs147540920 Chr.15: 45099686-45099686 on Build GRCh38
rs375943962 Chr.15: 45099378-45099378 on Build GRCh38
rs201877498 Chr.15: 45099653-45099653 on Build GRCh38
rs767324555 Chr.15: 45099824-45099824 on Build GRCh38
rs959718972 Chr.15: 45099739-45099739 on Build GRCh38
rs917130174 Chr.15: 45099651-45099651 on Build GRCh38
rs866257357 Chr.15: 45099413-45099413 on Build GRCh38
rs202085990 Chr.15: 45099810-45099810 on Build GRCh38
rs746176090 Chr.15: 45099761-45099761 on Build GRCh38
rs79480907 Chr.15: 45099816-45099816 on Build GRCh38
rs267604228 Chr.15: 45099753-45099753 on Build GRCh38
rs368746258 Chr.15: 45099400-45099400 on Build GRCh38
rs769776374 Chr.15: 45099752-45099752 on Build GRCh38
rs779741540 Chr.15: 45099486-45099486 on Build GRCh38
rs758000012 Chr.15: 45099484-45099484 on Build GRCh38
rs201282348 Chr.15: 45099687-45099687 on Build GRCh38
rs746301343 Chr.15: 45099808-45099808 on Build GRCh38
rs939551285 Chr.15: 45099683-45099683 on Build GRCh38
rs767200123 Chr.15: 45099778-45099778 on Build GRCh38
rs941026356 Chr.15: 45099661-45099661 on Build GRCh38
rs111315629 Chr.15: 45099369-45099370 on Build GRCh38
rs755108099 Chr.15: 45099456-45099456 on Build GRCh38
rs376833157 Chr.15: 45099819-45099819 on Build GRCh38
rs370794299 Chr.15: 45099657-45099657 on Build GRCh38
rs374614801 Chr.15: 45099694-45099694 on Build GRCh38
rs369781783 Chr.15: 45099496-45099496 on Build GRCh38
rs269869 Chr.15: 45099368-45099368 on Build GRCh38
rs766234674 Chr.15: 45099774-45099774 on Build GRCh38
rs150893514 Chr.15: 45099463-45099463 on Build GRCh38
rs751416598 Chr.15: 45099775-45099775 on Build GRCh38
rs973093293 Chr.15: 45099544-45099544 on Build GRCh38
rs559663716 Chr.15: 45099695-45099695 on Build GRCh38
rs115150281 Chr.15: 45099511-45099511 on Build GRCh38
rs368488511 Chr.15: 45099748-45099748 on Build GRCh38
rs1003699344 Chr.15: 45099391-45099391 on Build GRCh38
rs1010952104 Chr.15: 45099446-45099446 on Build GRCh38
rs201362841 Chr.15: 45099820-45099820 on Build GRCh38
rs376481943 Chr.15: 45099521-45099521 on Build GRCh38
rs753806363 Chr.15: 45099461-45099461 on Build GRCh38
rs770272807 Chr.15: 45099518-45099518 on Build GRCh38
rs753221072 Chr.15: 45099795-45099795 on Build GRCh38
rs747602398 Chr.15: 45099688-45099688 on Build GRCh38
rs376681974 Chr.15: 45099613-45099613 on Build GRCh38
rs200647474 Chr.15: 45099731-45099731 on Build GRCh38
rs899978179 Chr.15: 45099416-45099416 on Build GRCh38
rs886051192 Chr.15: 45099489-45099489 on Build GRCh38
rs974140902 Chr.15: 45099791-45099791 on Build GRCh38
rs777168376 Chr.15: 45099510-45099510 on Build GRCh38
rs929569738 Chr.15: 45099370-45099370 on Build GRCh38
rs762974325 Chr.15: 45099772-45099772 on Build GRCh38
rs375752800 Chr.15: 45099670-45099670 on Build GRCh38
rs920274747 Chr.15: 45099646-45099646 on Build GRCh38
rs868606571 Chr.15: 45099478-45099478 on Build GRCh38
rs757796376 Chr.15: 45099798-45099798 on Build GRCh38

View Full Product Details