Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 45118905-45119406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUOXA1
Entrez Gene ID
90527
Gene Name
dual oxidase maturation factor 1
Gene Aliases
NIP, NUMBIP, mol
UniGene
Hs.356664
Gene Chromosome Location
Chr.15: 45117366-45129938 on Build GRCh38
Transcripts
NM_001276264.1 NM_001276265.1 NM_001276266.1 NM_001276267.1 NM_001276268.1 NM_144565.3 XM_006720743.3 XM_006720745.1 XM_006720746.1 XM_006720747.3 XM_006720749.1 XM_006720751.3 XM_006720752.3 XM_006720754.1 XM_006720755.1 XM_011522181.2 XM_011522182.1 XM_011522183.2 XM_011522184.1 XM_011522185.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4148792 c.1029A>C p.L343F Chr.15: 45119109-45119109 on Build GRCh38
4826774 c.965C>G p.S322* Chr.15: 45119173-45119173 on Build GRCh38
7247073 c.840G>T p.K280N Chr.15: 45119298-45119298 on Build GRCh38
4389987 c.979A>G p.T327A Chr.15: 45119159-45119159 on Build GRCh38
5383398 c.980C>T p.T327I Chr.15: 45119158-45119158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200256639 Chr.15: 45119187-45119187 on Build GRCh38
rs777816961 Chr.15: 45119371-45119371 on Build GRCh38
rs564524035 Chr.15: 45119354-45119354 on Build GRCh38
rs757481210 Chr.15: 45119216-45119216 on Build GRCh38
rs926101072 Chr.15: 45118966-45118966 on Build GRCh38
rs199855983 Chr.15: 45119198-45119198 on Build GRCh38
rs776495746 Chr.15: 45119067-45119067 on Build GRCh38
rs773823827 Chr.15: 45119276-45119276 on Build GRCh38
rs765096521 Chr.15: 45119160-45119160 on Build GRCh38
rs934837823 Chr.15: 45118978-45118978 on Build GRCh38
rs773158418 Chr.15: 45119265-45119265 on Build GRCh38
rs778017134 Chr.15: 45119116-45119116 on Build GRCh38
rs16977686 Chr.15: 45119201-45119201 on Build GRCh38
rs750080774 Chr.15: 45119083-45119083 on Build GRCh38
rs770825058 Chr.15: 45119382-45119382 on Build GRCh38
rs537172649 Chr.15: 45119034-45119034 on Build GRCh38
rs763849904 Chr.15: 45119064-45119064 on Build GRCh38
rs987726607 Chr.15: 45119309-45119309 on Build GRCh38
rs754388768 Chr.15: 45119197-45119210 on Build GRCh38
rs781486796 Chr.15: 45119099-45119099 on Build GRCh38
rs978901903 Chr.15: 45118934-45118934 on Build GRCh38
rs1049265294 Chr.15: 45119223-45119223 on Build GRCh38
rs752170231 Chr.15: 45119094-45119094 on Build GRCh38
rs1012157708 Chr.15: 45119056-45119056 on Build GRCh38
rs201954258 Chr.15: 45119161-45119161 on Build GRCh38
rs1004692281 Chr.15: 45119024-45119024 on Build GRCh38
rs776609963 Chr.15: 45118990-45118990 on Build GRCh38
rs946471839 Chr.15: 45119137-45119137 on Build GRCh38
rs150452217 Chr.15: 45119196-45119196 on Build GRCh38
rs565975715 Chr.15: 45118985-45118985 on Build GRCh38
rs760652352 Chr.15: 45119062-45119062 on Build GRCh38
rs768088968 Chr.15: 45119191-45119191 on Build GRCh38
rs1039115653 Chr.15: 45118998-45118998 on Build GRCh38
rs766903143 Chr.15: 45119279-45119279 on Build GRCh38
rs371948395 Chr.15: 45119299-45119299 on Build GRCh38
rs975350744 Chr.15: 45119061-45119061 on Build GRCh38
rs749566812 Chr.15: 45119207-45119207 on Build GRCh38
rs776385926 Chr.15: 45119151-45119151 on Build GRCh38
rs556593213 Chr.15: 45119050-45119050 on Build GRCh38
rs749481036 Chr.15: 45119118-45119118 on Build GRCh38
rs534349863 Chr.15: 45118993-45118993 on Build GRCh38
rs372874489 Chr.15: 45119114-45119114 on Build GRCh38
rs905413917 Chr.15: 45118954-45118954 on Build GRCh38
rs933918251 Chr.15: 45119307-45119307 on Build GRCh38
rs896415858 Chr.15: 45119055-45119055 on Build GRCh38
rs886255710 Chr.15: 45119021-45119021 on Build GRCh38
rs541103488 Chr.15: 45119301-45119301 on Build GRCh38
rs748031965 Chr.15: 45119238-45119238 on Build GRCh38
rs184157954 Chr.15: 45118999-45118999 on Build GRCh38
rs750581745 Chr.15: 45119331-45119331 on Build GRCh38
rs774124760 Chr.15: 45119123-45119123 on Build GRCh38
rs367665353 Chr.15: 45119111-45119111 on Build GRCh38
rs868806123 Chr.15: 45118994-45118994 on Build GRCh38
rs774398321 Chr.15: 45119122-45119122 on Build GRCh38
rs764968386 Chr.15: 45119074-45119074 on Build GRCh38
rs989037277 Chr.15: 45118980-45118980 on Build GRCh38
rs140559023 Chr.15: 45119189-45119189 on Build GRCh38
rs374524880 Chr.15: 45119179-45119179 on Build GRCh38
rs745874216 Chr.15: 45119141-45119141 on Build GRCh38
rs766339299 Chr.15: 45119095-45119095 on Build GRCh38
rs79897809 Chr.15: 45119329-45119329 on Build GRCh38
rs761445781 Chr.15: 45119072-45119072 on Build GRCh38
rs762832270 Chr.15: 45119033-45119033 on Build GRCh38
rs143792283 Chr.15: 45119268-45119268 on Build GRCh38
rs749120729 Chr.15: 45119372-45119372 on Build GRCh38
rs753121085 Chr.15: 45119192-45119192 on Build GRCh38
rs148849457 Chr.15: 45119297-45119297 on Build GRCh38
rs201535627 Chr.15: 45119222-45119222 on Build GRCh38
rs921659832 Chr.15: 45119001-45119001 on Build GRCh38
rs772024127 Chr.15: 45119225-45119225 on Build GRCh38
rs773070416 Chr.15: 45119177-45119177 on Build GRCh38
rs779985265 Chr.15: 45119332-45119332 on Build GRCh38
rs946417989 Chr.15: 45119369-45119369 on Build GRCh38
rs747056739 Chr.15: 45119334-45119334 on Build GRCh38
rs1022436785 Chr.15: 45118930-45118930 on Build GRCh38
rs545798327 Chr.15: 45119018-45119018 on Build GRCh38
rs761719309 Chr.15: 45119156-45119156 on Build GRCh38
rs754407796 Chr.15: 45119188-45119188 on Build GRCh38
rs890699865 Chr.15: 45118931-45118931 on Build GRCh38
rs781231191 Chr.15: 45119234-45119234 on Build GRCh38
rs765986697 Chr.15: 45119092-45119092 on Build GRCh38
rs757575945 Chr.15: 45119319-45119319 on Build GRCh38
rs930381960 Chr.15: 45119013-45119013 on Build GRCh38
rs3803330 Chr.15: 45118982-45118982 on Build GRCh38
rs769769517 Chr.15: 45119252-45119252 on Build GRCh38
rs535602524 Chr.15: 45118981-45118981 on Build GRCh38
rs754204563 Chr.15: 45119300-45119300 on Build GRCh38
rs753309736 Chr.15: 45119281-45119281 on Build GRCh38
rs759131675 Chr.15: 45119277-45119277 on Build GRCh38
rs766073228 Chr.15: 45119184-45119184 on Build GRCh38
rs770741347 Chr.15: 45119275-45119275 on Build GRCh38
rs1043838472 Chr.15: 45119159-45119159 on Build GRCh38
rs758026133 Chr.15: 45119090-45119090 on Build GRCh38
rs199559225 Chr.15: 45119221-45119221 on Build GRCh38
rs952072391 Chr.15: 45119215-45119215 on Build GRCh38
rs569866625 Chr.15: 45118962-45118962 on Build GRCh38

View Full Product Details