Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 51453448-51453954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMXL2
Entrez Gene ID
23312
Gene Name
Dmx like 2
Gene Aliases
PEPNS, RC3
UniGene
Hs.511386
Gene Chromosome Location
Chr.15: 51447724-51622833 on Build GRCh38
Transcripts
NM_001174116.1 NM_001174117.1 NM_015263.3 XM_005254255.1 XM_005254256.1 XM_017022034.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1629590 c.8568T>C p.S2856S Chr.15: 51453612-51453612 on Build GRCh38
5800354 c.8618C>T p.S2873F Chr.15: 51453562-51453562 on Build GRCh38
7102060 c.8596C>G p.L2866V Chr.15: 51453584-51453584 on Build GRCh38
4971062 c.8539-4G>A p.? Chr.15: 51453645-51453645 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780647665 Chr.15: 51453519-51453519 on Build GRCh38
rs186815238 Chr.15: 51453850-51453850 on Build GRCh38
rs1023120618 Chr.15: 51453803-51453803 on Build GRCh38
rs758932790 Chr.15: 51453658-51453658 on Build GRCh38
rs1004287330 Chr.15: 51453777-51453777 on Build GRCh38
rs762703183 Chr.15: 51453562-51453562 on Build GRCh38
rs1019433281 Chr.15: 51453842-51453842 on Build GRCh38
rs757951844 Chr.15: 51453512-51453512 on Build GRCh38
rs748029047 Chr.15: 51453526-51453526 on Build GRCh38
rs946537529 Chr.15: 51453648-51453648 on Build GRCh38
rs770954045 Chr.15: 51453571-51453571 on Build GRCh38
rs183783275 Chr.15: 51453827-51453827 on Build GRCh38
rs758205027 Chr.15: 51453677-51453678 on Build GRCh38
rs1039329505 Chr.15: 51453617-51453617 on Build GRCh38
rs890787725 Chr.15: 51453764-51453764 on Build GRCh38
rs1002949499 Chr.15: 51453831-51453831 on Build GRCh38
rs577363200 Chr.15: 51453843-51453843 on Build GRCh38
rs189672861 Chr.15: 51453926-51453926 on Build GRCh38
rs563966899 Chr.15: 51453793-51453793 on Build GRCh38
rs762280638 Chr.15: 51453614-51453614 on Build GRCh38
rs750900599 Chr.15: 51453511-51453511 on Build GRCh38
rs774556103 Chr.15: 51453482-51453482 on Build GRCh38
rs367871337 Chr.15: 51453924-51453924 on Build GRCh38
rs887238961 Chr.15: 51453704-51453707 on Build GRCh38
rs760503397 Chr.15: 51453724-51453724 on Build GRCh38
rs561674212 Chr.15: 51453697-51453697 on Build GRCh38
rs878877563 Chr.15: 51453499-51453499 on Build GRCh38
rs979343951 Chr.15: 51453911-51453911 on Build GRCh38
rs970147396 Chr.15: 51453508-51453508 on Build GRCh38
rs919576879 Chr.15: 51453875-51453875 on Build GRCh38
rs1013203248 Chr.15: 51453763-51453763 on Build GRCh38
rs1020842269 Chr.15: 51453880-51453880 on Build GRCh38
rs931000380 Chr.15: 51453888-51453889 on Build GRCh38
rs769469101 Chr.15: 51453529-51453529 on Build GRCh38
rs923769489 Chr.15: 51453478-51453478 on Build GRCh38
rs201906356 Chr.15: 51453646-51453646 on Build GRCh38
rs192441641 Chr.15: 51453749-51453749 on Build GRCh38
rs956502446 Chr.15: 51453823-51453823 on Build GRCh38
rs1037249154 Chr.15: 51453790-51453790 on Build GRCh38
rs376767735 Chr.15: 51453659-51453659 on Build GRCh38
rs1031766146 Chr.15: 51453801-51453801 on Build GRCh38
rs368467532 Chr.15: 51453555-51453555 on Build GRCh38
rs745761670 Chr.15: 51453490-51453490 on Build GRCh38
rs372234458 Chr.15: 51453594-51453594 on Build GRCh38
rs374268189 Chr.15: 51453518-51453518 on Build GRCh38
rs867765971 Chr.15: 51453635-51453635 on Build GRCh38
rs977558174 Chr.15: 51453474-51453474 on Build GRCh38
rs539924813 Chr.15: 51453645-51453645 on Build GRCh38
rs919625375 Chr.15: 51453633-51453633 on Build GRCh38
rs867593783 Chr.15: 51453857-51453857 on Build GRCh38
rs968389801 Chr.15: 51453895-51453895 on Build GRCh38
rs754429972 Chr.15: 51453689-51453689 on Build GRCh38
rs559235172 Chr.15: 51453753-51453753 on Build GRCh38
rs995665921 Chr.15: 51453799-51453799 on Build GRCh38
rs575413040 Chr.15: 51453483-51453483 on Build GRCh38
rs1057374328 Chr.15: 51453748-51453749 on Build GRCh38
rs773320191 Chr.15: 51453541-51453541 on Build GRCh38
rs780960778 Chr.15: 51453690-51453690 on Build GRCh38
rs779560213 Chr.15: 51453517-51453517 on Build GRCh38
rs866255949 Chr.15: 51453735-51453735 on Build GRCh38
rs763982493 Chr.15: 51453774-51453774 on Build GRCh38
rs775720567 Chr.15: 51453584-51453584 on Build GRCh38
rs751093727 Chr.15: 51453683-51453683 on Build GRCh38
rs760618182 Chr.15: 51453585-51453585 on Build GRCh38
rs72729254 Chr.15: 51453719-51453719 on Build GRCh38

View Full Product Details