Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 52547443-52547946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARPP19
Entrez Gene ID
10776
Gene Name
cAMP regulated phosphoprotein 19
Gene Aliases
ARPP-16, ARPP-19, ARPP16, ENSAL
UniGene
Hs.512908
Gene Chromosome Location
Chr.15: 52547045-52569446 on Build GRCh38
Transcripts
NM_001306191.1 NM_001306195.1 NM_001306196.1 NM_006628.5 XM_017021868.1 XM_017021869.1 XM_017021870.1 XM_017021871.1

SNP Detail

SNP ID Chromosome Location
rs996397799 Chr.15: 52547500-52547500 on Build GRCh38
rs980671031 Chr.15: 52547628-52547628 on Build GRCh38
rs186764558 Chr.15: 52547833-52547833 on Build GRCh38
rs553631559 Chr.15: 52547585-52547585 on Build GRCh38
rs747756756 Chr.15: 52547470-52547473 on Build GRCh38
rs535253515 Chr.15: 52547474-52547474 on Build GRCh38
rs936260647 Chr.15: 52547642-52547642 on Build GRCh38
rs973132979 Chr.15: 52547835-52547835 on Build GRCh38
rs1040153154 Chr.15: 52547679-52547680 on Build GRCh38
rs767800254 Chr.15: 52547663-52547663 on Build GRCh38
rs917304893 Chr.15: 52547582-52547582 on Build GRCh38
rs952051756 Chr.15: 52547516-52547516 on Build GRCh38
rs563177409 Chr.15: 52547773-52547773 on Build GRCh38
rs984665048 Chr.15: 52547517-52547517 on Build GRCh38
rs1006153797 Chr.15: 52547778-52547778 on Build GRCh38
rs62023329 Chr.15: 52547616-52547616 on Build GRCh38
rs887574517 Chr.15: 52547728-52547728 on Build GRCh38
rs1016952588 Chr.15: 52547534-52547534 on Build GRCh38
rs996061960 Chr.15: 52547718-52547718 on Build GRCh38
rs961342502 Chr.15: 52547552-52547552 on Build GRCh38
rs1019086760 Chr.15: 52547482-52547482 on Build GRCh38
rs560987054 Chr.15: 52547909-52547909 on Build GRCh38
rs936148560 Chr.15: 52547889-52547889 on Build GRCh38
rs775736958 Chr.15: 52547473-52547474 on Build GRCh38
rs576101111 Chr.15: 52547613-52547613 on Build GRCh38
rs372546244 Chr.15: 52547522-52547522 on Build GRCh38
rs181175136 Chr.15: 52547492-52547492 on Build GRCh38
rs751699575 Chr.15: 52547606-52547606 on Build GRCh38
rs572130969 Chr.15: 52547471-52547471 on Build GRCh38
rs1061827 Chr.15: 52547485-52547485 on Build GRCh38
rs11539781 Chr.15: 52547826-52547826 on Build GRCh38
rs554655537 Chr.15: 52547607-52547607 on Build GRCh38
rs980172733 Chr.15: 52547886-52547886 on Build GRCh38
rs578125056 Chr.15: 52547797-52547797 on Build GRCh38
rs762932646 Chr.15: 52547493-52547493 on Build GRCh38
rs972833478 Chr.15: 52547565-52547565 on Build GRCh38
rs927608073 Chr.15: 52547635-52547637 on Build GRCh38
rs891422668 Chr.15: 52547466-52547466 on Build GRCh38
rs764054361 Chr.15: 52547570-52547570 on Build GRCh38
rs1014828769 Chr.15: 52547784-52547784 on Build GRCh38
rs891481241 Chr.15: 52547669-52547669 on Build GRCh38
rs549062725 Chr.15: 52547892-52547892 on Build GRCh38
rs1024428995 Chr.15: 52547868-52547868 on Build GRCh38
rs1029243759 Chr.15: 52547719-52547719 on Build GRCh38
rs961395140 Chr.15: 52547807-52547807 on Build GRCh38
rs771607097 Chr.15: 52547608-52547608 on Build GRCh38
rs945696817 Chr.15: 52547671-52547671 on Build GRCh38
rs913471482 Chr.15: 52547919-52547919 on Build GRCh38
rs578123963 Chr.15: 52547659-52547659 on Build GRCh38
rs560219193 Chr.15: 52547847-52547847 on Build GRCh38
rs971886724 Chr.15: 52547883-52547883 on Build GRCh38
rs11857682 Chr.15: 52547888-52547888 on Build GRCh38
rs1028943653 Chr.15: 52547512-52547512 on Build GRCh38
rs901581100 Chr.15: 52547716-52547716 on Build GRCh38

View Full Product Details