Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 52549397-52549909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARPP19
Entrez Gene ID
10776
Gene Name
cAMP regulated phosphoprotein 19
Gene Aliases
ARPP-16, ARPP-19, ARPP16, ENSAL
UniGene
Hs.512908
Gene Chromosome Location
Chr.15: 52547045-52569446 on Build GRCh38
Transcripts
NM_001306191.1 NM_001306195.1 NM_001306196.1 NM_006628.5 XM_017021868.1 XM_017021869.1 XM_017021870.1 XM_017021871.1

SNP Detail

SNP ID Chromosome Location
rs959114442 Chr.15: 52549671-52549671 on Build GRCh38
rs192719247 Chr.15: 52549440-52549440 on Build GRCh38
rs774314612 Chr.15: 52549518-52549523 on Build GRCh38
rs60316609 Chr.15: 52549875-52549876 on Build GRCh38
rs530674055 Chr.15: 52549501-52549501 on Build GRCh38
rs183117887 Chr.15: 52549484-52549484 on Build GRCh38
rs933295397 Chr.15: 52549468-52549468 on Build GRCh38
rs141579132 Chr.15: 52549441-52549441 on Build GRCh38
rs978053532 Chr.15: 52549753-52549753 on Build GRCh38
rs570460613 Chr.15: 52549640-52549640 on Build GRCh38
rs978381594 Chr.15: 52549453-52549453 on Build GRCh38
rs993776776 Chr.15: 52549537-52549537 on Build GRCh38
rs756984259 Chr.15: 52549718-52549718 on Build GRCh38
rs1048419800 Chr.15: 52549545-52549545 on Build GRCh38
rs973455667 Chr.15: 52549839-52549839 on Build GRCh38
rs371650109 Chr.15: 52549816-52549816 on Build GRCh38
rs769065082 Chr.15: 52549864-52549865 on Build GRCh38
rs954763482 Chr.15: 52549804-52549804 on Build GRCh38
rs188544453 Chr.15: 52549692-52549692 on Build GRCh38
rs527375020 Chr.15: 52549464-52549464 on Build GRCh38
rs565997848 Chr.15: 52549775-52549775 on Build GRCh38
rs1048002931 Chr.15: 52549874-52549875 on Build GRCh38
rs756785186 Chr.15: 52549535-52549535 on Build GRCh38
rs1022095027 Chr.15: 52549709-52549709 on Build GRCh38
rs987869699 Chr.15: 52549805-52549805 on Build GRCh38
rs559660370 Chr.15: 52549426-52549426 on Build GRCh38
rs767400502 Chr.15: 52549863-52549864 on Build GRCh38
rs910627724 Chr.15: 52549490-52549490 on Build GRCh38
rs780499843 Chr.15: 52549835-52549835 on Build GRCh38
rs920777870 Chr.15: 52549847-52549847 on Build GRCh38
rs906220939 Chr.15: 52549586-52549586 on Build GRCh38
rs11539780 Chr.15: 52549425-52549425 on Build GRCh38
rs551974835 Chr.15: 52549597-52549597 on Build GRCh38
rs1003605438 Chr.15: 52549614-52549614 on Build GRCh38
rs924649334 Chr.15: 52549465-52549465 on Build GRCh38
rs554063688 Chr.15: 52549729-52549729 on Build GRCh38
rs780622968 Chr.15: 52549668-52549668 on Build GRCh38
rs1037875770 Chr.15: 52549508-52549508 on Build GRCh38
rs144286553 Chr.15: 52549862-52549863 on Build GRCh38
rs966603486 Chr.15: 52549448-52549448 on Build GRCh38

View Full Product Details