Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 52550291-52550836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARPP19
Entrez Gene ID
10776
Gene Name
cAMP regulated phosphoprotein 19
Gene Aliases
ARPP-16, ARPP-19, ARPP16, ENSAL
UniGene
Hs.512908
Gene Chromosome Location
Chr.15: 52547045-52569446 on Build GRCh38
Transcripts
NM_001306191.1 NM_001306195.1 NM_001306196.1 NM_006628.5 XM_017021868.1 XM_017021869.1 XM_017021870.1 XM_017021871.1

SNP Detail

SNP ID Chromosome Location
rs564179437 Chr.15: 52550320-52550320 on Build GRCh38
rs752208206 Chr.15: 52550359-52550362 on Build GRCh38
rs74016837 Chr.15: 52550724-52550724 on Build GRCh38
rs866992956 Chr.15: 52550682-52550682 on Build GRCh38
rs141582878 Chr.15: 52550655-52550655 on Build GRCh38
rs886464720 Chr.15: 52550814-52550814 on Build GRCh38
rs776276391 Chr.15: 52550777-52550777 on Build GRCh38
rs6493566 Chr.15: 52550362-52550362 on Build GRCh38
rs999406221 Chr.15: 52550330-52550332 on Build GRCh38
rs35885704 Chr.15: 52550431-52550431 on Build GRCh38
rs796658749 Chr.15: 52550611-52550611 on Build GRCh38
rs181304212 Chr.15: 52550665-52550665 on Build GRCh38
rs964733917 Chr.15: 52550432-52550432 on Build GRCh38
rs11857272 Chr.15: 52550494-52550494 on Build GRCh38
rs923619110 Chr.15: 52550693-52550693 on Build GRCh38
rs567717200 Chr.15: 52550513-52550513 on Build GRCh38
rs777081571 Chr.15: 52550457-52550457 on Build GRCh38
rs902253747 Chr.15: 52550329-52550329 on Build GRCh38
rs1046953842 Chr.15: 52550811-52550811 on Build GRCh38
rs531717929 Chr.15: 52550616-52550617 on Build GRCh38
rs950720122 Chr.15: 52550622-52550622 on Build GRCh38
rs189267071 Chr.15: 52550643-52550643 on Build GRCh38
rs138993084 Chr.15: 52550594-52550594 on Build GRCh38
rs184434867 Chr.15: 52550545-52550545 on Build GRCh38
rs1028079009 Chr.15: 52550617-52550618 on Build GRCh38
rs759952871 Chr.15: 52550478-52550478 on Build GRCh38
rs898300101 Chr.15: 52550769-52550769 on Build GRCh38
rs4776056 Chr.15: 52550707-52550707 on Build GRCh38
rs766979254 Chr.15: 52550570-52550570 on Build GRCh38
rs150778286 Chr.15: 52550626-52550626 on Build GRCh38
rs1043372118 Chr.15: 52550554-52550554 on Build GRCh38
rs890977369 Chr.15: 52550596-52550596 on Build GRCh38
rs192347686 Chr.15: 52550346-52550346 on Build GRCh38
rs150870446 Chr.15: 52550664-52550664 on Build GRCh38
rs536589260 Chr.15: 52550514-52550514 on Build GRCh38
rs946420961 Chr.15: 52550538-52550538 on Build GRCh38
rs117898381 Chr.15: 52550505-52550505 on Build GRCh38
rs748632681 Chr.15: 52550468-52550468 on Build GRCh38
rs748968095 Chr.15: 52550743-52550743 on Build GRCh38
rs570038244 Chr.15: 52550673-52550674 on Build GRCh38
rs72623989 Chr.15: 52550531-52550531 on Build GRCh38
rs186483866 Chr.15: 52550672-52550672 on Build GRCh38
rs979624925 Chr.15: 52550685-52550685 on Build GRCh38
rs564608069 Chr.15: 52550625-52550626 on Build GRCh38
rs563413259 Chr.15: 52550696-52550696 on Build GRCh38
rs528089915 Chr.15: 52550456-52550456 on Build GRCh38
rs968274827 Chr.15: 52550679-52550679 on Build GRCh38
rs4776055 Chr.15: 52550659-52550659 on Build GRCh38
rs1051317872 Chr.15: 52550717-52550717 on Build GRCh38
rs995382155 Chr.15: 52550617-52550617 on Build GRCh38
rs960884405 Chr.15: 52550521-52550521 on Build GRCh38
rs1006830078 Chr.15: 52550416-52550416 on Build GRCh38
rs545644990 Chr.15: 52550745-52550745 on Build GRCh38
rs991055758 Chr.15: 52550522-52550522 on Build GRCh38
rs1006713752 Chr.15: 52550610-52550610 on Build GRCh38
rs995437693 Chr.15: 52550806-52550806 on Build GRCh38
rs766733614 Chr.15: 52550435-52550435 on Build GRCh38
rs189187576 Chr.15: 52550753-52550753 on Build GRCh38

View Full Product Details