Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 52551717-52552257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARPP19
Entrez Gene ID
10776
Gene Name
cAMP regulated phosphoprotein 19
Gene Aliases
ARPP-16, ARPP-19, ARPP16, ENSAL
UniGene
Hs.512908
Gene Chromosome Location
Chr.15: 52547045-52569446 on Build GRCh38
Transcripts
NM_001306191.1 NM_001306195.1 NM_001306196.1 NM_006628.5 XM_017021868.1 XM_017021869.1 XM_017021870.1 XM_017021871.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6871069 c.324C>T p.S108S Chr.15: 52551949-52551949 on Build GRCh38
1741372 c.338G>T p.*113L Chr.15: 52551935-52551935 on Build GRCh38
6291241 c.328C>T p.L110L Chr.15: 52551945-52551945 on Build GRCh38
5726776 c.242C>T p.A81V Chr.15: 52552031-52552031 on Build GRCh38
962859 c.173A>C p.K58T Chr.15: 52552100-52552100 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971900646 Chr.15: 52552193-52552193 on Build GRCh38
rs925250241 Chr.15: 52552083-52552083 on Build GRCh38
rs896227826 Chr.15: 52552129-52552129 on Build GRCh38
rs1019402015 Chr.15: 52552223-52552223 on Build GRCh38
rs776746187 Chr.15: 52552018-52552018 on Build GRCh38
rs763475402 Chr.15: 52551912-52551912 on Build GRCh38
rs373266168 Chr.15: 52552107-52552107 on Build GRCh38
rs747291355 Chr.15: 52551989-52551989 on Build GRCh38
rs773835778 Chr.15: 52552084-52552084 on Build GRCh38
rs190624189 Chr.15: 52552226-52552226 on Build GRCh38
rs992731567 Chr.15: 52551957-52551957 on Build GRCh38
rs749070045 Chr.15: 52551974-52551974 on Build GRCh38
rs373556910 Chr.15: 52551902-52551902 on Build GRCh38
rs1045160022 Chr.15: 52551750-52551750 on Build GRCh38
rs765890390 Chr.15: 52551899-52551899 on Build GRCh38
rs776897303 Chr.15: 52552168-52552168 on Build GRCh38
rs11539783 Chr.15: 52551969-52551969 on Build GRCh38
rs903943028 Chr.15: 52551751-52551751 on Build GRCh38
rs557025438 Chr.15: 52552184-52552184 on Build GRCh38
rs765932557 Chr.15: 52551915-52551915 on Build GRCh38
rs375653113 Chr.15: 52551973-52551973 on Build GRCh38
rs200974565 Chr.15: 52552119-52552119 on Build GRCh38
rs186917395 Chr.15: 52552136-52552136 on Build GRCh38
rs892659853 Chr.15: 52552119-52552119 on Build GRCh38
rs144580227 Chr.15: 52552178-52552178 on Build GRCh38
rs889663089 Chr.15: 52552208-52552208 on Build GRCh38
rs528673270 Chr.15: 52552027-52552027 on Build GRCh38
rs773910974 Chr.15: 52552025-52552025 on Build GRCh38
rs201676094 Chr.15: 52551988-52551988 on Build GRCh38
rs767115876 Chr.15: 52552112-52552112 on Build GRCh38
rs183208589 Chr.15: 52551865-52551865 on Build GRCh38
rs960123828 Chr.15: 52551948-52551948 on Build GRCh38
rs762429384 Chr.15: 52551889-52551889 on Build GRCh38
rs151240833 Chr.15: 52551964-52551964 on Build GRCh38
rs754379191 Chr.15: 52551939-52551939 on Build GRCh38
rs900082430 Chr.15: 52552204-52552204 on Build GRCh38
rs952498722 Chr.15: 52551910-52551910 on Build GRCh38
rs1019862013 Chr.15: 52551856-52551856 on Build GRCh38
rs908385716 Chr.15: 52551932-52551932 on Build GRCh38
rs750179875 Chr.15: 52552118-52552119 on Build GRCh38
rs569226018 Chr.15: 52552201-52552201 on Build GRCh38
rs112530318 Chr.15: 52551921-52551921 on Build GRCh38
rs552822810 Chr.15: 52551875-52551875 on Build GRCh38
rs764946186 Chr.15: 52551932-52551935 on Build GRCh38
rs747919014 Chr.15: 52551924-52551924 on Build GRCh38
rs868718681 Chr.15: 52551828-52551828 on Build GRCh38
rs763964968 Chr.15: 52552141-52552141 on Build GRCh38
rs1026775305 Chr.15: 52551905-52551905 on Build GRCh38
rs752200440 Chr.15: 52552124-52552124 on Build GRCh38
rs866018715 Chr.15: 52551878-52551878 on Build GRCh38
rs571088882 Chr.15: 52551943-52551943 on Build GRCh38
rs755680947 Chr.15: 52552133-52552133 on Build GRCh38
rs892598678 Chr.15: 52551819-52551819 on Build GRCh38
rs756966368 Chr.15: 52552148-52552148 on Build GRCh38
rs199641757 Chr.15: 52552139-52552139 on Build GRCh38
rs1008473308 Chr.15: 52552222-52552222 on Build GRCh38
rs777862029 Chr.15: 52551965-52551965 on Build GRCh38
rs776968155 Chr.15: 52551888-52551888 on Build GRCh38
rs1052623310 Chr.15: 52551815-52551815 on Build GRCh38
rs369014666 Chr.15: 52552019-52552019 on Build GRCh38
rs751236830 Chr.15: 52551927-52551927 on Build GRCh38
rs111757752 Chr.15: 52552194-52552194 on Build GRCh38
rs1008500222 Chr.15: 52551845-52551845 on Build GRCh38
rs770729483 Chr.15: 52551984-52551984 on Build GRCh38
rs201256320 Chr.15: 52552015-52552015 on Build GRCh38
rs117862230 Chr.15: 52551797-52551797 on Build GRCh38
rs369826005 Chr.15: 52551882-52551882 on Build GRCh38
rs753781758 Chr.15: 52552144-52552144 on Build GRCh38
rs1045044978 Chr.15: 52552057-52552057 on Build GRCh38
rs1042050390 Chr.15: 52552164-52552164 on Build GRCh38

View Full Product Details