Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 52581910-52582408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM214A
Entrez Gene ID
56204
Gene Name
family with sequence similarity 214 member A
Gene Aliases
KIAA1370
UniGene
Hs.152385
Gene Chromosome Location
Chr.15: 52581321-52709817 on Build GRCh38
Transcripts
NM_001286495.1 NM_019600.3 NR_104457.1 XM_005254547.2 XM_005254548.2 XM_011521789.2 XM_011521790.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4682915 c.3196T>G p.S1066A Chr.15: 52582189-52582189 on Build GRCh38
1608477 c.3140_3147delGAAAGTCA p.R1047fs*4 Chr.15: 52582238-52582245 on Build GRCh38
4517290 c.3125_3126GG>AT p.R1042H Chr.15: 52582259-52582260 on Build GRCh38
5551441 c.3120C>T p.D1040D Chr.15: 52582265-52582265 on Build GRCh38
5478927 c.3182A>C p.K1061T Chr.15: 52582203-52582203 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977317216 Chr.15: 52581942-52581942 on Build GRCh38
rs879078901 Chr.15: 52582222-52582222 on Build GRCh38
rs763182531 Chr.15: 52582321-52582321 on Build GRCh38
rs901626112 Chr.15: 52582100-52582100 on Build GRCh38
rs762951047 Chr.15: 52582153-52582153 on Build GRCh38
rs372153473 Chr.15: 52582231-52582231 on Build GRCh38
rs773493119 Chr.15: 52582134-52582134 on Build GRCh38
rs897459555 Chr.15: 52582340-52582340 on Build GRCh38
rs201372446 Chr.15: 52582316-52582316 on Build GRCh38
rs765359192 Chr.15: 52582129-52582129 on Build GRCh38
rs369015694 Chr.15: 52582264-52582264 on Build GRCh38
rs924511643 Chr.15: 52581967-52581967 on Build GRCh38
rs377506717 Chr.15: 52582110-52582110 on Build GRCh38
rs766474472 Chr.15: 52582182-52582182 on Build GRCh38
rs113862970 Chr.15: 52582070-52582070 on Build GRCh38
rs375475675 Chr.15: 52582244-52582244 on Build GRCh38
rs887335557 Chr.15: 52582203-52582203 on Build GRCh38
rs780976457 Chr.15: 52582322-52582322 on Build GRCh38
rs1025833577 Chr.15: 52582108-52582108 on Build GRCh38
rs756780019 Chr.15: 52582261-52582261 on Build GRCh38
rs752011284 Chr.15: 52582205-52582205 on Build GRCh38
rs771189651 Chr.15: 52582345-52582345 on Build GRCh38
rs748929090 Chr.15: 52582025-52582025 on Build GRCh38
rs201947269 Chr.15: 52582291-52582291 on Build GRCh38
rs370561381 Chr.15: 52582265-52582265 on Build GRCh38
rs768499507 Chr.15: 52582317-52582317 on Build GRCh38
rs774382212 Chr.15: 52582348-52582348 on Build GRCh38
rs923509948 Chr.15: 52582048-52582048 on Build GRCh38
rs905089083 Chr.15: 52582380-52582380 on Build GRCh38
rs1054499179 Chr.15: 52581981-52581981 on Build GRCh38
rs967785533 Chr.15: 52581991-52581991 on Build GRCh38
rs760588991 Chr.15: 52582125-52582125 on Build GRCh38
rs745413880 Chr.15: 52582274-52582274 on Build GRCh38
rs745868675 Chr.15: 52582358-52582358 on Build GRCh38
rs375329211 Chr.15: 52582318-52582318 on Build GRCh38
rs777665140 Chr.15: 52582128-52582129 on Build GRCh38
rs758068713 Chr.15: 52581973-52581973 on Build GRCh38
rs1046090529 Chr.15: 52582366-52582366 on Build GRCh38
rs998746488 Chr.15: 52582105-52582105 on Build GRCh38
rs779799066 Chr.15: 52581955-52581955 on Build GRCh38
rs756514693 Chr.15: 52582236-52582236 on Build GRCh38
rs755454638 Chr.15: 52582217-52582217 on Build GRCh38
rs780102159 Chr.15: 52582312-52582312 on Build GRCh38
rs777225690 Chr.15: 52582237-52582237 on Build GRCh38
rs569713549 Chr.15: 52581990-52581990 on Build GRCh38
rs949885338 Chr.15: 52581937-52581937 on Build GRCh38
rs368419269 Chr.15: 52582223-52582223 on Build GRCh38
rs1024336397 Chr.15: 52582273-52582273 on Build GRCh38

View Full Product Details