Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 57420853-57421371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CGNL1
Entrez Gene ID
84952
Gene Name
cingulin like 1
Gene Aliases
JACOP, PCING
UniGene
Hs.148989
Gene Chromosome Location
Chr.15: 57376487-57550727 on Build GRCh38
Transcripts
NM_001252335.1 NM_032866.4 XM_005254726.3 XM_005254727.4 XM_011522120.1 XM_011522121.1 XM_017022685.1 XM_017022686.1

SNP Detail

SNP ID Chromosome Location
rs191772200 Chr.15: 57421345-57421345 on Build GRCh38
rs896833869 Chr.15: 57420974-57420974 on Build GRCh38
rs894921743 Chr.15: 57420911-57420911 on Build GRCh38
rs865994282 Chr.15: 57420940-57420940 on Build GRCh38
rs1018453777 Chr.15: 57421047-57421047 on Build GRCh38
rs1051340954 Chr.15: 57421315-57421315 on Build GRCh38
rs72738080 Chr.15: 57421096-57421096 on Build GRCh38
rs147770762 Chr.15: 57421241-57421241 on Build GRCh38
rs776805177 Chr.15: 57421311-57421311 on Build GRCh38
rs1706373 Chr.15: 57421213-57421213 on Build GRCh38
rs906361707 Chr.15: 57420884-57420884 on Build GRCh38
rs778206465 Chr.15: 57421191-57421191 on Build GRCh38
rs939210864 Chr.15: 57420888-57420888 on Build GRCh38
rs945262376 Chr.15: 57420876-57420876 on Build GRCh38
rs977959177 Chr.15: 57421106-57421106 on Build GRCh38
rs542536490 Chr.15: 57421003-57421003 on Build GRCh38
rs990269277 Chr.15: 57421066-57421066 on Build GRCh38
rs11071316 Chr.15: 57421035-57421035 on Build GRCh38
rs564056325 Chr.15: 57421110-57421110 on Build GRCh38
rs896797285 Chr.15: 57421283-57421283 on Build GRCh38
rs967380410 Chr.15: 57421100-57421100 on Build GRCh38
rs148614464 Chr.15: 57421002-57421002 on Build GRCh38
rs957645740 Chr.15: 57421058-57421058 on Build GRCh38
rs377373395 Chr.15: 57421104-57421105 on Build GRCh38
rs376750136 Chr.15: 57421018-57421018 on Build GRCh38
rs770869055 Chr.15: 57421271-57421271 on Build GRCh38
rs144153245 Chr.15: 57421019-57421019 on Build GRCh38
rs927924091 Chr.15: 57421117-57421117 on Build GRCh38
rs200899646 Chr.15: 57421299-57421299 on Build GRCh38
rs1042298777 Chr.15: 57420878-57420878 on Build GRCh38
rs546230713 Chr.15: 57421134-57421134 on Build GRCh38
rs559764764 Chr.15: 57420937-57420937 on Build GRCh38
rs11858037 Chr.15: 57420908-57420908 on Build GRCh38
rs368870189 Chr.15: 57421127-57421127 on Build GRCh38
rs537669005 Chr.15: 57421327-57421327 on Build GRCh38
rs566128671 Chr.15: 57421143-57421143 on Build GRCh38
rs186219423 Chr.15: 57421144-57421144 on Build GRCh38
rs993875000 Chr.15: 57421305-57421305 on Build GRCh38
rs28576065 Chr.15: 57420931-57420931 on Build GRCh38
rs80168281 Chr.15: 57421120-57421120 on Build GRCh38
rs1016969337 Chr.15: 57420951-57420951 on Build GRCh38
rs548878215 Chr.15: 57421145-57421145 on Build GRCh38

View Full Product Details