Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 57437900-57438401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CGNL1
Entrez Gene ID
84952
Gene Name
cingulin like 1
Gene Aliases
JACOP, PCING
UniGene
Hs.148989
Gene Chromosome Location
Chr.15: 57376487-57550727 on Build GRCh38
Transcripts
NM_001252335.1 NM_032866.4 XM_005254726.3 XM_005254727.4 XM_011522120.1 XM_011522121.1 XM_017022685.1 XM_017022686.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3887223 c.211G>A p.E71K Chr.15: 57438210-57438210 on Build GRCh38
310069 c.190C>T p.L64L Chr.15: 57438189-57438189 on Build GRCh38
4490644 c.366C>T p.L122L Chr.15: 57438365-57438365 on Build GRCh38
2219128 c.184C>T p.R62W Chr.15: 57438183-57438183 on Build GRCh38
4187167 c.214A>T p.N72Y Chr.15: 57438213-57438213 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779573168 Chr.15: 57438376-57438376 on Build GRCh38
rs974227907 Chr.15: 57438300-57438300 on Build GRCh38
rs759215334 Chr.15: 57438192-57438192 on Build GRCh38
rs746480640 Chr.15: 57437954-57437954 on Build GRCh38
rs1043457311 Chr.15: 57438209-57438209 on Build GRCh38
rs143019005 Chr.15: 57438203-57438203 on Build GRCh38
rs765193693 Chr.15: 57438290-57438290 on Build GRCh38
rs777659946 Chr.15: 57438051-57438051 on Build GRCh38
rs777819228 Chr.15: 57438157-57438157 on Build GRCh38
rs781271183 Chr.15: 57437959-57437959 on Build GRCh38
rs768679656 Chr.15: 57437957-57437957 on Build GRCh38
rs754539162 Chr.15: 57438125-57438125 on Build GRCh38
rs1023784739 Chr.15: 57438148-57438148 on Build GRCh38
rs772505623 Chr.15: 57438330-57438330 on Build GRCh38
rs764627311 Chr.15: 57438196-57438196 on Build GRCh38
rs762174793 Chr.15: 57438008-57438008 on Build GRCh38
rs61998108 Chr.15: 57438140-57438140 on Build GRCh38
rs564485602 Chr.15: 57438348-57438348 on Build GRCh38
rs779543265 Chr.15: 57438030-57438030 on Build GRCh38
rs748030532 Chr.15: 57438047-57438047 on Build GRCh38
rs766223967 Chr.15: 57438129-57438129 on Build GRCh38
rs773538446 Chr.15: 57438068-57438070 on Build GRCh38
rs763225818 Chr.15: 57438114-57438114 on Build GRCh38
rs753839950 Chr.15: 57438269-57438269 on Build GRCh38
rs775685005 Chr.15: 57437990-57437990 on Build GRCh38
rs755678494 Chr.15: 57438197-57438197 on Build GRCh38
rs565854948 Chr.15: 57438232-57438232 on Build GRCh38
rs752850285 Chr.15: 57438302-57438302 on Build GRCh38
rs773580516 Chr.15: 57438183-57438183 on Build GRCh38
rs367680036 Chr.15: 57438217-57438217 on Build GRCh38
rs2934441 Chr.15: 57438206-57438206 on Build GRCh38
rs771348097 Chr.15: 57438247-57438247 on Build GRCh38
rs192521923 Chr.15: 57438119-57438119 on Build GRCh38
rs760493042 Chr.15: 57438111-57438111 on Build GRCh38
rs767096185 Chr.15: 57438136-57438136 on Build GRCh38
rs776742721 Chr.15: 57438175-57438175 on Build GRCh38
rs777664973 Chr.15: 57438318-57438318 on Build GRCh38
rs866506108 Chr.15: 57438016-57438016 on Build GRCh38
rs764148295 Chr.15: 57438202-57438202 on Build GRCh38
rs770353590 Chr.15: 57438182-57438182 on Build GRCh38
rs769654103 Chr.15: 57437966-57437966 on Build GRCh38
rs747163175 Chr.15: 57438167-57438167 on Build GRCh38
rs777158079 Chr.15: 57438097-57438097 on Build GRCh38
rs146561570 Chr.15: 57437998-57437998 on Build GRCh38
rs150676873 Chr.15: 57438323-57438323 on Build GRCh38
rs1280393 Chr.15: 57438359-57438359 on Build GRCh38
rs546056290 Chr.15: 57438299-57438299 on Build GRCh38
rs752701723 Chr.15: 57438141-57438141 on Build GRCh38
rs761709518 Chr.15: 57438274-57438274 on Build GRCh38
rs1008700156 Chr.15: 57437926-57437926 on Build GRCh38
rs753930634 Chr.15: 57438375-57438375 on Build GRCh38
rs999602107 Chr.15: 57438070-57438070 on Build GRCh38
rs781292917 Chr.15: 57438044-57438044 on Build GRCh38
rs769807707 Chr.15: 57438236-57438236 on Build GRCh38
rs371804275 Chr.15: 57438358-57438358 on Build GRCh38
rs1035255197 Chr.15: 57438085-57438085 on Build GRCh38
rs758177120 Chr.15: 57438293-57438293 on Build GRCh38
rs759947371 Chr.15: 57438252-57438252 on Build GRCh38
rs746289208 Chr.15: 57438329-57438329 on Build GRCh38
rs779469261 Chr.15: 57438324-57438324 on Build GRCh38
rs944597549 Chr.15: 57438365-57438365 on Build GRCh38
rs769742701 Chr.15: 57438050-57438050 on Build GRCh38
rs756168597 Chr.15: 57438145-57438145 on Build GRCh38
rs770069570 Chr.15: 57438044-57438045 on Build GRCh38
rs754696787 Chr.15: 57438184-57438184 on Build GRCh38
rs372675578 Chr.15: 57438029-57438029 on Build GRCh38
rs760360365 Chr.15: 57438342-57438342 on Build GRCh38
rs751149283 Chr.15: 57438041-57438041 on Build GRCh38
rs114236402 Chr.15: 57437970-57437970 on Build GRCh38
rs750393487 Chr.15: 57438058-57438058 on Build GRCh38
rs141997044 Chr.15: 57438268-57438268 on Build GRCh38
rs761385726 Chr.15: 57438355-57438355 on Build GRCh38
rs371054975 Chr.15: 57438347-57438347 on Build GRCh38
rs184748613 Chr.15: 57438242-57438242 on Build GRCh38
rs775264968 Chr.15: 57438267-57438267 on Build GRCh38
rs750100614 Chr.15: 57438018-57438018 on Build GRCh38
rs756250743 Chr.15: 57438311-57438311 on Build GRCh38
rs752938070 Chr.15: 57438362-57438362 on Build GRCh38
rs1038914184 Chr.15: 57437948-57437948 on Build GRCh38
rs756984832 Chr.15: 57438160-57438160 on Build GRCh38
rs776047162 Chr.15: 57438349-57438349 on Build GRCh38
rs755236164 Chr.15: 57438218-57438218 on Build GRCh38
rs750288593 Chr.15: 57438292-57438292 on Build GRCh38
rs748198919 Chr.15: 57438177-57438177 on Build GRCh38
rs768336144 Chr.15: 57438346-57438346 on Build GRCh38
rs778528325 Chr.15: 57438215-57438215 on Build GRCh38
rs774082923 Chr.15: 57438188-57438188 on Build GRCh38
rs760861852 Chr.15: 57438092-57438092 on Build GRCh38
rs946541290 Chr.15: 57438208-57438208 on Build GRCh38
rs182623732 Chr.15: 57437977-57437977 on Build GRCh38
rs766123717 Chr.15: 57438360-57438360 on Build GRCh38
rs200914383 Chr.15: 57438234-57438234 on Build GRCh38
rs139101816 Chr.15: 57438042-57438042 on Build GRCh38
rs751116729 Chr.15: 57438120-57438120 on Build GRCh38
rs773775045 Chr.15: 57438239-57438239 on Build GRCh38
rs929968334 Chr.15: 57438161-57438161 on Build GRCh38
rs552662762 Chr.15: 57438096-57438096 on Build GRCh38
rs771914487 Chr.15: 57438258-57438258 on Build GRCh38
rs768750302 Chr.15: 57438171-57438171 on Build GRCh38
rs778022294 Chr.15: 57438367-57438367 on Build GRCh38
rs765549295 Chr.15: 57438014-57438014 on Build GRCh38
rs377283226 Chr.15: 57438194-57438194 on Build GRCh38
rs749365303 Chr.15: 57438319-57438319 on Build GRCh38
rs749013050 Chr.15: 57438062-57438062 on Build GRCh38
rs760636959 Chr.15: 57437991-57437991 on Build GRCh38
rs139876229 Chr.15: 57438339-57438339 on Build GRCh38
rs762262580 Chr.15: 57438109-57438109 on Build GRCh38
rs866945743 Chr.15: 57437995-57437995 on Build GRCh38
rs758899965 Chr.15: 57438220-57438220 on Build GRCh38
rs566024340 Chr.15: 57438110-57438110 on Build GRCh38
rs749171957 Chr.15: 57438158-57438158 on Build GRCh38
rs753388710 Chr.15: 57438151-57438151 on Build GRCh38
rs779569537 Chr.15: 57437952-57437952 on Build GRCh38
rs764648283 Chr.15: 57437993-57437993 on Build GRCh38
rs774625872 Chr.15: 57438250-57438250 on Build GRCh38
rs775568562 Chr.15: 57438066-57438066 on Build GRCh38
rs199798727 Chr.15: 57437960-57437960 on Build GRCh38
rs772375615 Chr.15: 57438063-57438063 on Build GRCh38
rs763359098 Chr.15: 57438283-57438285 on Build GRCh38

View Full Product Details