Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 57591822-57592328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCOM1
Entrez Gene ID
145781
Gene Name
GRINL1A complex locus 1
Gene Aliases
GRINL1A, Gcom2, MYZAP, MYZAP-POLR2M, gcom
UniGene
Hs.437256
Gene Chromosome Location
Chr.15: 57591904-57717557 on Build GRCh38
Transcripts
NM_001018090.6 NM_001018091.6 NM_001285900.3 NR_104367.2 NR_104368.2 NR_104369.2 NR_104370.2 NR_104371.3 NM_001018100.4 NM_152451.7

Gene Symbol
MYZAP
Entrez Gene ID
100820829
Gene Name
myocardial zonula adherens protein
Gene Aliases
GCOM1, Gup, MYOZAP
UniGene
Hs.437256
Gene Chromosome Location
Chr.15: 57591904-57685364 on Build GRCh38
Transcripts
NM_001018090.6 NM_001018091.6 NM_001285900.3 NR_104367.2 NR_104368.2 NR_104369.2 NR_104370.2 NR_104371.3 NM_001018100.4 NM_152451.7

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3744677 c.4C>T p.L2L Chr.15: 57592038-57592038 on Build GRCh38
5790544 c.14C>A p.T5K Chr.15: 57592048-57592048 on Build GRCh38
5790546 c.14C>A p.T5K Chr.15: 57592048-57592048 on Build GRCh38
3744676 c.4C>T p.L2L Chr.15: 57592038-57592038 on Build GRCh38
5790545 c.14C>A p.T5K Chr.15: 57592048-57592048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965749986 Chr.15: 57592165-57592165 on Build GRCh38
rs564526066 Chr.15: 57592046-57592046 on Build GRCh38
rs748735582 Chr.15: 57592242-57592242 on Build GRCh38
rs1000535451 Chr.15: 57591848-57591848 on Build GRCh38
rs76940573 Chr.15: 57592098-57592098 on Build GRCh38
rs1014237410 Chr.15: 57591878-57591878 on Build GRCh38
rs923183174 Chr.15: 57592195-57592195 on Build GRCh38
rs982809553 Chr.15: 57592137-57592137 on Build GRCh38
rs1025180675 Chr.15: 57591882-57591882 on Build GRCh38
rs936446414 Chr.15: 57592214-57592214 on Build GRCh38
rs573379038 Chr.15: 57591987-57591987 on Build GRCh38
rs969953357 Chr.15: 57591895-57591895 on Build GRCh38
rs1050213537 Chr.15: 57592011-57592011 on Build GRCh38
rs542609269 Chr.15: 57592236-57592236 on Build GRCh38
rs892123858 Chr.15: 57592050-57592050 on Build GRCh38
rs1052176048 Chr.15: 57592218-57592218 on Build GRCh38
rs747424571 Chr.15: 57592081-57592081 on Build GRCh38
rs12902584 Chr.15: 57591908-57591908 on Build GRCh38
rs2453089 Chr.15: 57592021-57592021 on Build GRCh38
rs376140185 Chr.15: 57591940-57591940 on Build GRCh38
rs976399795 Chr.15: 57591989-57591989 on Build GRCh38
rs528382845 Chr.15: 57591966-57591966 on Build GRCh38
rs544581323 Chr.15: 57592031-57592031 on Build GRCh38
rs757466633 Chr.15: 57592070-57592071 on Build GRCh38
rs542491438 Chr.15: 57591999-57591999 on Build GRCh38
rs1002347693 Chr.15: 57592091-57592091 on Build GRCh38
rs985974314 Chr.15: 57592199-57592199 on Build GRCh38
rs952824947 Chr.15: 57592106-57592106 on Build GRCh38
rs768953798 Chr.15: 57592201-57592210 on Build GRCh38
rs865782710 Chr.15: 57592146-57592146 on Build GRCh38
rs1015585138 Chr.15: 57592142-57592142 on Build GRCh38
rs112902884 Chr.15: 57592295-57592295 on Build GRCh38
rs1046044265 Chr.15: 57592238-57592238 on Build GRCh38
rs923162948 Chr.15: 57591990-57591990 on Build GRCh38
rs905457106 Chr.15: 57592264-57592264 on Build GRCh38
rs73422816 Chr.15: 57592297-57592297 on Build GRCh38
rs1027032427 Chr.15: 57591919-57591919 on Build GRCh38
rs980931056 Chr.15: 57591909-57591909 on Build GRCh38
rs769052776 Chr.15: 57592019-57592019 on Build GRCh38
rs982761890 Chr.15: 57591954-57591954 on Build GRCh38
rs796094590 Chr.15: 57592169-57592169 on Build GRCh38
rs925090242 Chr.15: 57592210-57592210 on Build GRCh38
rs866723396 Chr.15: 57592001-57592001 on Build GRCh38
rs1013872576 Chr.15: 57592071-57592071 on Build GRCh38
rs149008805 Chr.15: 57592018-57592018 on Build GRCh38
rs1025658573 Chr.15: 57592079-57592079 on Build GRCh38
rs533865982 Chr.15: 57591965-57591965 on Build GRCh38
rs1690328 Chr.15: 57591983-57591983 on Build GRCh38
rs557751823 Chr.15: 57591870-57591870 on Build GRCh38
rs12902591 Chr.15: 57591921-57591921 on Build GRCh38
rs953296613 Chr.15: 57592197-57592197 on Build GRCh38
rs775491208 Chr.15: 57592088-57592088 on Build GRCh38
rs879587575 Chr.15: 57591929-57591929 on Build GRCh38
rs913617726 Chr.15: 57592230-57592230 on Build GRCh38
rs780242925 Chr.15: 57592026-57592026 on Build GRCh38
rs571228758 Chr.15: 57591960-57591960 on Build GRCh38
rs12902754 Chr.15: 57592010-57592010 on Build GRCh38
rs952775911 Chr.15: 57591934-57591934 on Build GRCh38
rs746032149 Chr.15: 57592005-57592005 on Build GRCh38
rs892752150 Chr.15: 57591872-57591872 on Build GRCh38

View Full Product Details