Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 58771418-58771921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM63B
Entrez Gene ID
54629
Gene Name
family with sequence similarity 63 member B
Gene Aliases
-
UniGene
Hs.591122
Gene Chromosome Location
Chr.15: 58771192-58861900 on Build GRCh38
Transcripts
NM_001040450.2 NM_001040453.2 XM_011521687.1 XM_011521688.1 XM_011521689.1 XM_011521690.1 XM_017022332.1 XM_017022333.1 XM_017022334.1 XM_017022335.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5980764 c.358G>A p.G120S Chr.15: 58771753-58771753 on Build GRCh38
7367428 c.208G>T p.A70S Chr.15: 58771603-58771603 on Build GRCh38
6388861 c.37C>A p.H13N Chr.15: 58771432-58771432 on Build GRCh38
6374991 c.154A>G p.N52D Chr.15: 58771549-58771549 on Build GRCh38
3690458 c.148G>A p.G50S Chr.15: 58771543-58771543 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770102832 Chr.15: 58771766-58771766 on Build GRCh38
rs747903042 Chr.15: 58771628-58771628 on Build GRCh38
rs748176378 Chr.15: 58771813-58771813 on Build GRCh38
rs1047300335 Chr.15: 58771555-58771555 on Build GRCh38
rs756016282 Chr.15: 58771747-58771747 on Build GRCh38
rs539766602 Chr.15: 58771638-58771638 on Build GRCh38
rs201039791 Chr.15: 58771588-58771588 on Build GRCh38
rs553596891 Chr.15: 58771641-58771641 on Build GRCh38
rs374853839 Chr.15: 58771751-58771751 on Build GRCh38
rs555945171 Chr.15: 58771740-58771740 on Build GRCh38
rs755912172 Chr.15: 58771699-58771699 on Build GRCh38
rs777065911 Chr.15: 58771727-58771727 on Build GRCh38
rs201611014 Chr.15: 58771575-58771575 on Build GRCh38
rs771183759 Chr.15: 58771827-58771827 on Build GRCh38
rs780073430 Chr.15: 58771752-58771752 on Build GRCh38
rs777362398 Chr.15: 58771444-58771444 on Build GRCh38
rs1047390 Chr.15: 58771652-58771652 on Build GRCh38
rs752541990 Chr.15: 58771697-58771697 on Build GRCh38
rs373444154 Chr.15: 58771712-58771712 on Build GRCh38
rs754987830 Chr.15: 58771693-58771693 on Build GRCh38
rs199885473 Chr.15: 58771757-58771757 on Build GRCh38
rs753541761 Chr.15: 58771604-58771604 on Build GRCh38
rs533321521 Chr.15: 58771775-58771775 on Build GRCh38
rs754859746 Chr.15: 58771623-58771623 on Build GRCh38
rs773648199 Chr.15: 58771767-58771767 on Build GRCh38
rs751778422 Chr.15: 58771804-58771804 on Build GRCh38
rs182620597 Chr.15: 58771706-58771706 on Build GRCh38
rs753675620 Chr.15: 58771793-58771793 on Build GRCh38
rs745765854 Chr.15: 58771457-58771457 on Build GRCh38
rs750079041 Chr.15: 58771662-58771662 on Build GRCh38
rs768709836 Chr.15: 58771755-58771755 on Build GRCh38
rs755598950 Chr.15: 58771880-58771880 on Build GRCh38
rs746795225 Chr.15: 58771557-58771557 on Build GRCh38
rs372464935 Chr.15: 58771831-58771831 on Build GRCh38
rs764370579 Chr.15: 58771782-58771782 on Build GRCh38
rs968690430 Chr.15: 58771617-58771617 on Build GRCh38
rs548696115 Chr.15: 58771574-58771574 on Build GRCh38
rs756945687 Chr.15: 58771801-58771801 on Build GRCh38
rs372488999 Chr.15: 58771602-58771602 on Build GRCh38
rs533078522 Chr.15: 58771538-58771538 on Build GRCh38
rs760478601 Chr.15: 58771476-58771476 on Build GRCh38
rs763901310 Chr.15: 58771791-58771791 on Build GRCh38
rs374830274 Chr.15: 58771812-58771812 on Build GRCh38
rs544669057 Chr.15: 58771753-58771753 on Build GRCh38
rs774562152 Chr.15: 58771636-58771636 on Build GRCh38
rs61729219 Chr.15: 58771460-58771460 on Build GRCh38
rs764812639 Chr.15: 58771651-58771651 on Build GRCh38
rs775229500 Chr.15: 58771890-58771890 on Build GRCh38
rs774211067 Chr.15: 58771829-58771829 on Build GRCh38
rs761014613 Chr.15: 58771601-58771601 on Build GRCh38
rs758298665 Chr.15: 58771551-58771551 on Build GRCh38
rs770192873 Chr.15: 58771732-58771732 on Build GRCh38
rs771305481 Chr.15: 58771633-58771633 on Build GRCh38
rs747740122 Chr.15: 58771852-58771853 on Build GRCh38
rs761735588 Chr.15: 58771843-58771843 on Build GRCh38
rs757733869 Chr.15: 58771875-58771875 on Build GRCh38
rs781111103 Chr.15: 58771627-58771627 on Build GRCh38
rs755213669 Chr.15: 58771848-58771848 on Build GRCh38
rs768015371 Chr.15: 58771512-58771512 on Build GRCh38
rs143583599 Chr.15: 58771613-58771613 on Build GRCh38
rs368821954 Chr.15: 58771544-58771544 on Build GRCh38
rs764540000 Chr.15: 58771642-58771642 on Build GRCh38
rs779444438 Chr.15: 58771877-58771877 on Build GRCh38
rs755741260 Chr.15: 58771438-58771438 on Build GRCh38
rs745484008 Chr.15: 58771830-58771830 on Build GRCh38
rs28559610 Chr.15: 58771815-58771815 on Build GRCh38
rs34805327 Chr.15: 58771749-58771749 on Build GRCh38
rs547168026 Chr.15: 58771886-58771886 on Build GRCh38
rs776005436 Chr.15: 58771598-58771598 on Build GRCh38
rs773633276 Chr.15: 58771568-58771568 on Build GRCh38
rs748620361 Chr.15: 58771763-58771763 on Build GRCh38
rs374433778 Chr.15: 58771714-58771714 on Build GRCh38
rs773512320 Chr.15: 58771493-58771493 on Build GRCh38
rs765881093 Chr.15: 58771681-58771681 on Build GRCh38
rs148901278 Chr.15: 58771453-58771453 on Build GRCh38
rs779915824 Chr.15: 58771616-58771616 on Build GRCh38
rs746960227 Chr.15: 58771621-58771621 on Build GRCh38
rs763187587 Chr.15: 58771773-58771773 on Build GRCh38
rs759046395 Chr.15: 58771741-58771741 on Build GRCh38
rs770418677 Chr.15: 58771447-58771447 on Build GRCh38
rs766561644 Chr.15: 58771498-58771498 on Build GRCh38
rs781232058 Chr.15: 58771696-58771696 on Build GRCh38
rs377735527 Chr.15: 58771746-58771746 on Build GRCh38
rs772095570 Chr.15: 58771710-58771710 on Build GRCh38
rs751825887 Chr.15: 58771846-58771846 on Build GRCh38
rs769915400 Chr.15: 58771491-58771491 on Build GRCh38
rs775171731 Chr.15: 58771467-58771467 on Build GRCh38
rs879195007 Chr.15: 58771485-58771485 on Build GRCh38
rs540190279 Chr.15: 58771789-58771789 on Build GRCh38
rs765898070 Chr.15: 58771737-58771737 on Build GRCh38
rs1037468456 Chr.15: 58771525-58771525 on Build GRCh38
rs751499297 Chr.15: 58771684-58771684 on Build GRCh38
rs756929862 Chr.15: 58771608-58771608 on Build GRCh38
rs759937759 Chr.15: 58771504-58771504 on Build GRCh38
rs748798122 Chr.15: 58771445-58771445 on Build GRCh38
rs778074754 Chr.15: 58771818-58771818 on Build GRCh38
rs928998355 Chr.15: 58771552-58771552 on Build GRCh38
rs546842447 Chr.15: 58771545-58771545 on Build GRCh38
rs752827017 Chr.15: 58771857-58771857 on Build GRCh38
rs755573819 Chr.15: 58771635-58771640 on Build GRCh38
rs746198882 Chr.15: 58771885-58771885 on Build GRCh38
rs557921964 Chr.15: 58771572-58771572 on Build GRCh38
rs759171383 Chr.15: 58771780-58771780 on Build GRCh38
rs376730103 Chr.15: 58771495-58771495 on Build GRCh38
rs375666296 Chr.15: 58771854-58771854 on Build GRCh38
rs749751848 Chr.15: 58771873-58771873 on Build GRCh38
rs753031354 Chr.15: 58771513-58771513 on Build GRCh38
rs752098631 Chr.15: 58771580-58771593 on Build GRCh38
rs762224127 Chr.15: 58771729-58771729 on Build GRCh38
rs914928868 Chr.15: 58771549-58771549 on Build GRCh38
rs756805124 Chr.15: 58771546-58771546 on Build GRCh38
rs572997970 Chr.15: 58771840-58771840 on Build GRCh38
rs767911572 Chr.15: 58771594-58771594 on Build GRCh38
rs915747588 Chr.15: 58771469-58771469 on Build GRCh38
rs775174435 Chr.15: 58771832-58771832 on Build GRCh38
rs778398259 Chr.15: 58771869-58771869 on Build GRCh38
rs760041782 Chr.15: 58771838-58771838 on Build GRCh38
rs772777415 Chr.15: 58771734-58771734 on Build GRCh38
rs779976761 Chr.15: 58771554-58771554 on Build GRCh38
rs566561884 Chr.15: 58771547-58771547 on Build GRCh38
rs745947406 Chr.15: 58771637-58771637 on Build GRCh38
rs749672851 Chr.15: 58771826-58771826 on Build GRCh38
rs757910331 Chr.15: 58771672-58771672 on Build GRCh38
rs202092838 Chr.15: 58771842-58771842 on Build GRCh38
rs746074293 Chr.15: 58771707-58771707 on Build GRCh38
rs989200457 Chr.15: 58771523-58771523 on Build GRCh38
rs369822716 Chr.15: 58771896-58771896 on Build GRCh38
rs368390369 Chr.15: 58771561-58771561 on Build GRCh38
rs79904206 Chr.15: 58771590-58771590 on Build GRCh38
rs756086188 Chr.15: 58771859-58771859 on Build GRCh38
rs573394169 Chr.15: 58771645-58771645 on Build GRCh38
rs764106464 Chr.15: 58771839-58771839 on Build GRCh38
rs201095543 Chr.15: 58771845-58771845 on Build GRCh38
rs745335327 Chr.15: 58771550-58771550 on Build GRCh38
rs887395032 Chr.15: 58771559-58771559 on Build GRCh38
rs755092897 Chr.15: 58771807-58771807 on Build GRCh38
rs557342297 Chr.15: 58771610-58771610 on Build GRCh38
rs376666617 Chr.15: 58771713-58771713 on Build GRCh38
rs780535245 Chr.15: 58771847-58771848 on Build GRCh38
rs760687084 Chr.15: 58771790-58771790 on Build GRCh38

View Full Product Details