Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 59668976-59669482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BNIP2
Entrez Gene ID
663
Gene Name
BCL2 interacting protein 2
Gene Aliases
BNIP-2, NIP2
UniGene
Hs.592515
Gene Chromosome Location
Chr.15: 59659146-59689534 on Build GRCh38
Transcripts
NM_001320674.1 NM_001320675.1 NM_004330.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5762797 c.773C>A p.A258D Chr.15: 59669297-59669297 on Build GRCh38
6077599 c.1128A>T p.T376T Chr.15: 59669305-59669305 on Build GRCh38
375307 c.773C>T p.A258V Chr.15: 59669297-59669297 on Build GRCh38
6023114 c.1136C>T p.A379V Chr.15: 59669297-59669297 on Build GRCh38
2219865 c.1123A>G p.R375G Chr.15: 59669310-59669310 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749719262 Chr.15: 59669381-59669381 on Build GRCh38
rs771807709 Chr.15: 59669371-59669372 on Build GRCh38
rs781645829 Chr.15: 59669254-59669254 on Build GRCh38
rs373202880 Chr.15: 59668998-59668998 on Build GRCh38
rs756620444 Chr.15: 59669266-59669266 on Build GRCh38
rs921229482 Chr.15: 59669236-59669236 on Build GRCh38
rs185658195 Chr.15: 59669220-59669220 on Build GRCh38
rs189676136 Chr.15: 59669092-59669092 on Build GRCh38
rs6151564 Chr.15: 59669178-59669178 on Build GRCh38
rs778802346 Chr.15: 59669383-59669383 on Build GRCh38
rs764855990 Chr.15: 59669244-59669244 on Build GRCh38
rs762944172 Chr.15: 59669013-59669013 on Build GRCh38
rs767734708 Chr.15: 59669412-59669436 on Build GRCh38
rs778136249 Chr.15: 59669267-59669267 on Build GRCh38
rs775201503 Chr.15: 59669327-59669327 on Build GRCh38
rs775287881 Chr.15: 59669032-59669032 on Build GRCh38
rs6151563 Chr.15: 59669407-59669407 on Build GRCh38
rs778635651 Chr.15: 59669235-59669236 on Build GRCh38
rs1034767529 Chr.15: 59669088-59669088 on Build GRCh38
rs540122408 Chr.15: 59669224-59669227 on Build GRCh38
rs756532689 Chr.15: 59669372-59669372 on Build GRCh38
rs745487001 Chr.15: 59669236-59669239 on Build GRCh38
rs893658413 Chr.15: 59669299-59669299 on Build GRCh38
rs1003669402 Chr.15: 59669087-59669087 on Build GRCh38
rs756812586 Chr.15: 59669236-59669236 on Build GRCh38
rs759810592 Chr.15: 59669192-59669192 on Build GRCh38
rs776496755 Chr.15: 59669381-59669384 on Build GRCh38
rs373080895 Chr.15: 59669329-59669329 on Build GRCh38
rs146320872 Chr.15: 59669320-59669320 on Build GRCh38
rs999362000 Chr.15: 59669016-59669016 on Build GRCh38
rs764026317 Chr.15: 59669235-59669235 on Build GRCh38
rs200965538 Chr.15: 59669358-59669358 on Build GRCh38
rs775679911 Chr.15: 59669395-59669395 on Build GRCh38
rs201366762 Chr.15: 59669338-59669338 on Build GRCh38
rs2307273 Chr.15: 59669447-59669447 on Build GRCh38
rs778275461 Chr.15: 59669038-59669038 on Build GRCh38
rs764608819 Chr.15: 59669347-59669347 on Build GRCh38
rs370872659 Chr.15: 59669301-59669301 on Build GRCh38
rs766132553 Chr.15: 59669366-59669366 on Build GRCh38
rs745954102 Chr.15: 59669387-59669387 on Build GRCh38
rs760461704 Chr.15: 59669332-59669332 on Build GRCh38
rs759842883 Chr.15: 59669021-59669021 on Build GRCh38
rs761846020 Chr.15: 59669345-59669345 on Build GRCh38
rs774521143 Chr.15: 59669302-59669302 on Build GRCh38
rs747284683 Chr.15: 59669401-59669401 on Build GRCh38
rs564193293 Chr.15: 59669046-59669046 on Build GRCh38
rs1052190039 Chr.15: 59669285-59669285 on Build GRCh38
rs753883024 Chr.15: 59669239-59669239 on Build GRCh38
rs760549528 Chr.15: 59669231-59669231 on Build GRCh38
rs767259589 Chr.15: 59669022-59669022 on Build GRCh38
rs543382587 Chr.15: 59669096-59669096 on Build GRCh38
rs778249080 Chr.15: 59669380-59669380 on Build GRCh38
rs373197847 Chr.15: 59669258-59669258 on Build GRCh38
rs750008558 Chr.15: 59669245-59669245 on Build GRCh38
rs749825061 Chr.15: 59669228-59669232 on Build GRCh38
rs758064780 Chr.15: 59669357-59669357 on Build GRCh38
rs924699763 Chr.15: 59669259-59669259 on Build GRCh38
rs757806351 Chr.15: 59669382-59669382 on Build GRCh38
rs72751178 Chr.15: 59669100-59669100 on Build GRCh38
rs556392162 Chr.15: 59669380-59669381 on Build GRCh38
rs143193624 Chr.15: 59669372-59669372 on Build GRCh38
rs774422562 Chr.15: 59669020-59669020 on Build GRCh38
rs183573691 Chr.15: 59669367-59669367 on Build GRCh38
rs768344686 Chr.15: 59669379-59669380 on Build GRCh38
rs776414544 Chr.15: 59669344-59669344 on Build GRCh38
rs767868223 Chr.15: 59669250-59669251 on Build GRCh38
rs756712716 Chr.15: 59669243-59669243 on Build GRCh38
rs758151044 Chr.15: 59669250-59669250 on Build GRCh38
rs772295514 Chr.15: 59669323-59669323 on Build GRCh38
rs368806052 Chr.15: 59669004-59669004 on Build GRCh38
rs1023232885 Chr.15: 59669049-59669049 on Build GRCh38
rs962875290 Chr.15: 59669400-59669400 on Build GRCh38
rs200051385 Chr.15: 59669392-59669392 on Build GRCh38
rs969083295 Chr.15: 59669060-59669060 on Build GRCh38
rs142043306 Chr.15: 59669279-59669279 on Build GRCh38
rs545417919 Chr.15: 59669017-59669017 on Build GRCh38
rs982954458 Chr.15: 59669157-59669157 on Build GRCh38
rs975681582 Chr.15: 59669203-59669203 on Build GRCh38
rs774460118 Chr.15: 59669393-59669398 on Build GRCh38
rs975630720 Chr.15: 59669438-59669438 on Build GRCh38
rs746783995 Chr.15: 59669372-59669373 on Build GRCh38
rs749968673 Chr.15: 59669352-59669352 on Build GRCh38
rs893709377 Chr.15: 59669042-59669042 on Build GRCh38
rs759553515 Chr.15: 59669402-59669403 on Build GRCh38

View Full Product Details