Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 63340629-63341134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA12
Entrez Gene ID
771
Gene Name
carbonic anhydrase 12
Gene Aliases
CA-XII, CAXII, HsT18816, T18816
UniGene
Hs.210995
Gene Chromosome Location
Chr.15: 63321378-63382166 on Build GRCh38
Transcripts
NM_001218.4 NM_001293642.1 NM_206925.2 NR_135511.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6617089 c.534C>T p.S178S Chr.15: 63340775-63340775 on Build GRCh38
470929 c.559T>C p.F187L Chr.15: 63340750-63340750 on Build GRCh38
963680 c.570T>G p.L190L Chr.15: 63340739-63340739 on Build GRCh38
963681 c.543G>A p.P181P Chr.15: 63340766-63340766 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372096514 Chr.15: 63340831-63340831 on Build GRCh38
rs750273598 Chr.15: 63340678-63340680 on Build GRCh38
rs371863633 Chr.15: 63340766-63340766 on Build GRCh38
rs762247061 Chr.15: 63341059-63341059 on Build GRCh38
rs145237465 Chr.15: 63340732-63340732 on Build GRCh38
rs74369730 Chr.15: 63340965-63340965 on Build GRCh38
rs1025163846 Chr.15: 63340892-63340892 on Build GRCh38
rs201128913 Chr.15: 63340775-63340775 on Build GRCh38
rs945513992 Chr.15: 63341058-63341058 on Build GRCh38
rs959107471 Chr.15: 63340684-63340684 on Build GRCh38
rs985348827 Chr.15: 63341022-63341022 on Build GRCh38
rs754043358 Chr.15: 63340867-63340867 on Build GRCh38
rs919825860 Chr.15: 63340711-63340711 on Build GRCh38
rs959035314 Chr.15: 63340860-63340860 on Build GRCh38
rs764842995 Chr.15: 63340980-63340980 on Build GRCh38
rs748070978 Chr.15: 63340703-63340703 on Build GRCh38
rs546736883 Chr.15: 63341091-63341091 on Build GRCh38
rs981738105 Chr.15: 63340979-63340979 on Build GRCh38
rs773989455 Chr.15: 63340671-63340671 on Build GRCh38
rs1033121355 Chr.15: 63340856-63340856 on Build GRCh38
rs757073469 Chr.15: 63341027-63341033 on Build GRCh38
rs1001082785 Chr.15: 63341070-63341070 on Build GRCh38
rs775475611 Chr.15: 63340691-63340691 on Build GRCh38
rs771790538 Chr.15: 63340742-63340742 on Build GRCh38
rs777675276 Chr.15: 63340727-63340727 on Build GRCh38
rs543054470 Chr.15: 63340679-63340679 on Build GRCh38
rs115818954 Chr.15: 63340956-63340956 on Build GRCh38
rs529006930 Chr.15: 63340710-63340710 on Build GRCh38
rs763460109 Chr.15: 63340699-63340699 on Build GRCh38
rs375480906 Chr.15: 63340713-63340713 on Build GRCh38
rs2306719 Chr.15: 63341039-63341039 on Build GRCh38
rs961177939 Chr.15: 63340898-63340898 on Build GRCh38
rs552901624 Chr.15: 63340851-63340851 on Build GRCh38
rs770776028 Chr.15: 63340954-63340954 on Build GRCh38
rs368224069 Chr.15: 63340824-63340824 on Build GRCh38
rs141310681 Chr.15: 63340760-63340760 on Build GRCh38
rs755459847 Chr.15: 63340714-63340714 on Build GRCh38
rs761443210 Chr.15: 63340763-63340763 on Build GRCh38
rs992320244 Chr.15: 63340687-63340687 on Build GRCh38
rs1001156300 Chr.15: 63340848-63340848 on Build GRCh38
rs1046454401 Chr.15: 63340731-63340731 on Build GRCh38
rs748610754 Chr.15: 63340719-63340719 on Build GRCh38
rs991894864 Chr.15: 63340882-63340882 on Build GRCh38
rs780045316 Chr.15: 63340782-63340783 on Build GRCh38
rs989016036 Chr.15: 63340910-63340910 on Build GRCh38
rs370295741 Chr.15: 63340735-63340735 on Build GRCh38
rs375327149 Chr.15: 63340821-63340821 on Build GRCh38
rs145417517 Chr.15: 63340784-63340784 on Build GRCh38
rs1040396678 Chr.15: 63341038-63341038 on Build GRCh38
rs760616893 Chr.15: 63340694-63340694 on Build GRCh38
rs532989116 Chr.15: 63341041-63341041 on Build GRCh38
rs768045632 Chr.15: 63341076-63341076 on Build GRCh38
rs757427796 Chr.15: 63340829-63340829 on Build GRCh38
rs927593964 Chr.15: 63340999-63340999 on Build GRCh38
rs375002808 Chr.15: 63340752-63340752 on Build GRCh38
rs370563418 Chr.15: 63340700-63340700 on Build GRCh38
rs910520541 Chr.15: 63341031-63341031 on Build GRCh38
rs181871612 Chr.15: 63341033-63341033 on Build GRCh38
rs939862723 Chr.15: 63340701-63340701 on Build GRCh38
rs753144843 Chr.15: 63340804-63340804 on Build GRCh38
rs781692852 Chr.15: 63340715-63340715 on Build GRCh38
rs904095530 Chr.15: 63340846-63340846 on Build GRCh38
rs1013651913 Chr.15: 63341102-63341102 on Build GRCh38
rs1026988 Chr.15: 63340835-63340835 on Build GRCh38
rs771040822 Chr.15: 63340734-63340734 on Build GRCh38
rs140120607 Chr.15: 63341086-63341086 on Build GRCh38
rs759378424 Chr.15: 63340676-63340676 on Build GRCh38
rs1021150053 Chr.15: 63340841-63340841 on Build GRCh38
rs186476589 Chr.15: 63341069-63341069 on Build GRCh38
rs985564225 Chr.15: 63340675-63340675 on Build GRCh38
rs368771609 Chr.15: 63340720-63340720 on Build GRCh38
rs747127290 Chr.15: 63340834-63340835 on Build GRCh38
rs767945277 Chr.15: 63340801-63340801 on Build GRCh38

View Full Product Details