Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 63597657-63598163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL22
Entrez Gene ID
283807
Gene Name
F-box and leucine rich repeat protein 22
Gene Aliases
Fbl22
UniGene
Hs.631163
Gene Chromosome Location
Chr.15: 63596225-63608898 on Build GRCh38
Transcripts
NM_203373.2 XM_005254320.3 XM_006720478.1 XM_011521468.2 XM_011521469.1 XM_011521472.2 XM_017022086.1 NR_034080.1

Gene Symbol
USP3-AS1
Entrez Gene ID
100130855
Gene Name
USP3 antisense RNA 1
Gene Aliases
-
UniGene
Hs.631163
Gene Chromosome Location
Chr.15: 63587710-63600827 on Build GRCh38
Transcripts
NM_203373.2 XM_005254320.3 XM_006720478.1 XM_011521468.2 XM_011521469.1 XM_011521472.2 XM_017022086.1 NR_034080.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5431703 c.306C>G p.V102V Chr.15: 63597716-63597716 on Build GRCh38
5024419 c.306C>T p.V102V Chr.15: 63597716-63597716 on Build GRCh38
6220726 c.322C>T p.R108W Chr.15: 63597732-63597732 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1045655292 Chr.15: 63597957-63597957 on Build GRCh38
rs546801032 Chr.15: 63597848-63597848 on Build GRCh38
rs752845315 Chr.15: 63597944-63597944 on Build GRCh38
rs758756870 Chr.15: 63597746-63597746 on Build GRCh38
rs924368103 Chr.15: 63597849-63597849 on Build GRCh38
rs751019157 Chr.15: 63597698-63597698 on Build GRCh38
rs375470672 Chr.15: 63597749-63597749 on Build GRCh38
rs577834305 Chr.15: 63597732-63597732 on Build GRCh38
rs770075152 Chr.15: 63597790-63597790 on Build GRCh38
rs572640425 Chr.15: 63597705-63597705 on Build GRCh38
rs561362799 Chr.15: 63597788-63597788 on Build GRCh38
rs955733204 Chr.15: 63597759-63597759 on Build GRCh38
rs1054283231 Chr.15: 63597878-63597878 on Build GRCh38
rs146309376 Chr.15: 63597700-63597700 on Build GRCh38
rs535860067 Chr.15: 63598086-63598086 on Build GRCh38
rs887195018 Chr.15: 63598066-63598066 on Build GRCh38
rs542349907 Chr.15: 63597961-63597961 on Build GRCh38
rs979999831 Chr.15: 63598137-63598137 on Build GRCh38
rs369184505 Chr.15: 63597978-63597978 on Build GRCh38
rs8035931 Chr.15: 63597735-63597735 on Build GRCh38
rs867547812 Chr.15: 63597902-63597902 on Build GRCh38
rs571226429 Chr.15: 63597709-63597709 on Build GRCh38
rs139584661 Chr.15: 63597707-63597707 on Build GRCh38
rs559557825 Chr.15: 63597731-63597731 on Build GRCh38
rs1040434955 Chr.15: 63597908-63597908 on Build GRCh38
rs372149528 Chr.15: 63597733-63597733 on Build GRCh38
rs932246174 Chr.15: 63597958-63597958 on Build GRCh38
rs768602453 Chr.15: 63597714-63597714 on Build GRCh38
rs1017063227 Chr.15: 63598091-63598091 on Build GRCh38
rs758363206 Chr.15: 63597694-63597694 on Build GRCh38
rs188260211 Chr.15: 63597922-63597922 on Build GRCh38
rs1023948445 Chr.15: 63598100-63598100 on Build GRCh38
rs752172251 Chr.15: 63597814-63597814 on Build GRCh38
rs769736483 Chr.15: 63597730-63597730 on Build GRCh38
rs566216530 Chr.15: 63598116-63598116 on Build GRCh38
rs1000312275 Chr.15: 63597993-63597993 on Build GRCh38
rs550551575 Chr.15: 63597953-63597953 on Build GRCh38
rs755666400 Chr.15: 63597826-63597826 on Build GRCh38
rs780958764 Chr.15: 63597701-63597701 on Build GRCh38
rs753506154 Chr.15: 63597896-63597896 on Build GRCh38
rs141771513 Chr.15: 63598088-63598088 on Build GRCh38
rs567731106 Chr.15: 63597804-63597804 on Build GRCh38
rs955043330 Chr.15: 63597831-63597831 on Build GRCh38
rs148453374 Chr.15: 63597738-63597738 on Build GRCh38
rs964253497 Chr.15: 63597776-63597776 on Build GRCh38
rs935872685 Chr.15: 63597868-63597868 on Build GRCh38
rs752422490 Chr.15: 63597740-63597740 on Build GRCh38
rs768812575 Chr.15: 63597768-63597768 on Build GRCh38
rs907828609 Chr.15: 63597854-63597854 on Build GRCh38
rs1024167970 Chr.15: 63597802-63597802 on Build GRCh38
rs571432318 Chr.15: 63597853-63597853 on Build GRCh38
rs886455711 Chr.15: 63597688-63597688 on Build GRCh38
rs891579384 Chr.15: 63598085-63598085 on Build GRCh38
rs368375969 Chr.15: 63597727-63597727 on Build GRCh38
rs1017177358 Chr.15: 63597769-63597769 on Build GRCh38
rs1047612608 Chr.15: 63598004-63598004 on Build GRCh38
rs948623488 Chr.15: 63597935-63597935 on Build GRCh38
rs200607556 Chr.15: 63597741-63597741 on Build GRCh38
rs573827635 Chr.15: 63597801-63597802 on Build GRCh38
rs556328889 Chr.15: 63598138-63598138 on Build GRCh38
rs139954123 Chr.15: 63597742-63597742 on Build GRCh38
rs149590841 Chr.15: 63597724-63597724 on Build GRCh38
rs781085089 Chr.15: 63597781-63597781 on Build GRCh38
rs762053791 Chr.15: 63597710-63597710 on Build GRCh38
rs1004056901 Chr.15: 63597739-63597739 on Build GRCh38
rs772586197 Chr.15: 63597744-63597747 on Build GRCh38
rs766002635 Chr.15: 63597697-63597697 on Build GRCh38
rs975602141 Chr.15: 63597825-63597825 on Build GRCh38
rs528623394 Chr.15: 63597803-63597803 on Build GRCh38
rs543050690 Chr.15: 63597756-63597756 on Build GRCh38
rs747737098 Chr.15: 63597704-63597704 on Build GRCh38
rs750716680 Chr.15: 63597743-63597743 on Build GRCh38
rs915714048 Chr.15: 63597887-63597887 on Build GRCh38

View Full Product Details