Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 64165947-64166460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK1G1
Entrez Gene ID
53944
Gene Name
casein kinase 1 gamma 1
Gene Aliases
CK1gamma1
UniGene
Hs.646508
Gene Chromosome Location
Chr.15: 64165517-64356243 on Build GRCh38
Transcripts
NM_022048.3

SNP Detail

SNP ID Chromosome Location
rs750287681 Chr.15: 64166035-64166035 on Build GRCh38
rs374863011 Chr.15: 64166209-64166209 on Build GRCh38
rs966208658 Chr.15: 64166147-64166147 on Build GRCh38
rs940567976 Chr.15: 64166350-64166350 on Build GRCh38
rs553614661 Chr.15: 64166338-64166339 on Build GRCh38
rs1036047154 Chr.15: 64166361-64166363 on Build GRCh38
rs560906005 Chr.15: 64166372-64166372 on Build GRCh38
rs62024252 Chr.15: 64166206-64166206 on Build GRCh38
rs948718974 Chr.15: 64166325-64166325 on Build GRCh38
rs979657849 Chr.15: 64166215-64166215 on Build GRCh38
rs772523505 Chr.15: 64166283-64166283 on Build GRCh38
rs901479429 Chr.15: 64166363-64166363 on Build GRCh38
rs866082780 Chr.15: 64166082-64166082 on Build GRCh38
rs1029586841 Chr.15: 64166173-64166173 on Build GRCh38
rs756234657 Chr.15: 64166339-64166344 on Build GRCh38
rs1045731398 Chr.15: 64166376-64166376 on Build GRCh38
rs149507637 Chr.15: 64166320-64166320 on Build GRCh38
rs778827677 Chr.15: 64166020-64166020 on Build GRCh38
rs1186092 Chr.15: 64166081-64166081 on Build GRCh38
rs978257205 Chr.15: 64166250-64166250 on Build GRCh38
rs534877489 Chr.15: 64165999-64165999 on Build GRCh38
rs997249761 Chr.15: 64166156-64166156 on Build GRCh38
rs929385322 Chr.15: 64166433-64166433 on Build GRCh38
rs752333445 Chr.15: 64166044-64166044 on Build GRCh38
rs750154201 Chr.15: 64166083-64166085 on Build GRCh38
rs764974184 Chr.15: 64166082-64166083 on Build GRCh38
rs753373601 Chr.15: 64166083-64166084 on Build GRCh38
rs779970680 Chr.15: 64166322-64166322 on Build GRCh38
rs907335500 Chr.15: 64166034-64166034 on Build GRCh38
rs779371881 Chr.15: 64166103-64166103 on Build GRCh38
rs916197353 Chr.15: 64166324-64166324 on Build GRCh38
rs1012877210 Chr.15: 64166013-64166013 on Build GRCh38
rs917908381 Chr.15: 64166268-64166268 on Build GRCh38
rs1026119733 Chr.15: 64166104-64166104 on Build GRCh38
rs74417649 Chr.15: 64166094-64166095 on Build GRCh38
rs757711310 Chr.15: 64166097-64166097 on Build GRCh38
rs949518665 Chr.15: 64166279-64166279 on Build GRCh38
rs754169012 Chr.15: 64166347-64166350 on Build GRCh38
rs866364561 Chr.15: 64166214-64166214 on Build GRCh38
rs980844320 Chr.15: 64166307-64166307 on Build GRCh38
rs997684729 Chr.15: 64166364-64166366 on Build GRCh38
rs956947409 Chr.15: 64166236-64166236 on Build GRCh38
rs368506081 Chr.15: 64165994-64165994 on Build GRCh38
rs907227340 Chr.15: 64166387-64166387 on Build GRCh38
rs927120203 Chr.15: 64166319-64166320 on Build GRCh38
rs796261731 Chr.15: 64166083-64166084 on Build GRCh38
rs936167817 Chr.15: 64166276-64166276 on Build GRCh38
rs746930093 Chr.15: 64166377-64166377 on Build GRCh38
rs1017949478 Chr.15: 64166177-64166177 on Build GRCh38
rs73462567 Chr.15: 64166130-64166130 on Build GRCh38
rs553310203 Chr.15: 64166129-64166129 on Build GRCh38
rs1001865565 Chr.15: 64166037-64166037 on Build GRCh38
rs924778318 Chr.15: 64166260-64166260 on Build GRCh38
rs1028704849 Chr.15: 64166364-64166364 on Build GRCh38
rs966476586 Chr.15: 64166225-64166225 on Build GRCh38
rs767122339 Chr.15: 64166344-64166345 on Build GRCh38
rs575278502 Chr.15: 64166247-64166247 on Build GRCh38
rs563373217 Chr.15: 64166219-64166219 on Build GRCh38
rs571331741 Chr.15: 64166191-64166191 on Build GRCh38
rs751453829 Chr.15: 64166320-64166320 on Build GRCh38
rs143314613 Chr.15: 64166083-64166083 on Build GRCh38
rs767343722 Chr.15: 64166000-64166000 on Build GRCh38
rs1048967671 Chr.15: 64166111-64166111 on Build GRCh38

View Full Product Details