Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 64921840-64922345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKDD1A
Entrez Gene ID
348094
Gene Name
ankyrin repeat and death domain containing 1A
Gene Aliases
-
UniGene
Hs.207157
Gene Chromosome Location
Chr.15: 64911902-64958700 on Build GRCh38
Transcripts
NM_182703.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5777462 c.358G>A p.E120K Chr.15: 64922011-64922011 on Build GRCh38
3503115 c.3G>A p.M1I Chr.15: 64921929-64921929 on Build GRCh38
5454368 c.8C>T p.A3V Chr.15: 64921934-64921934 on Build GRCh38
3503116 c.289C>T p.L97L Chr.15: 64921942-64921942 on Build GRCh38
3503117 c.16C>T p.L6L Chr.15: 64921942-64921942 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747634110 Chr.15: 64922070-64922070 on Build GRCh38
rs999341910 Chr.15: 64922076-64922076 on Build GRCh38
rs939917988 Chr.15: 64922143-64922143 on Build GRCh38
rs933353784 Chr.15: 64922215-64922215 on Build GRCh38
rs777161469 Chr.15: 64921998-64921998 on Build GRCh38
rs901316643 Chr.15: 64922082-64922082 on Build GRCh38
rs767882746 Chr.15: 64921935-64921935 on Build GRCh38
rs76626463 Chr.15: 64922011-64922011 on Build GRCh38
rs769551689 Chr.15: 64921992-64921992 on Build GRCh38
rs201916280 Chr.15: 64921967-64921967 on Build GRCh38
rs996972844 Chr.15: 64922084-64922084 on Build GRCh38
rs199588777 Chr.15: 64921957-64921957 on Build GRCh38
rs1028508383 Chr.15: 64922085-64922085 on Build GRCh38
rs529443304 Chr.15: 64922301-64922301 on Build GRCh38
rs759721172 Chr.15: 64922182-64922182 on Build GRCh38
rs753865851 Chr.15: 64922034-64922034 on Build GRCh38
rs772696532 Chr.15: 64922000-64922000 on Build GRCh38
rs832887 Chr.15: 64922163-64922163 on Build GRCh38
rs747829826 Chr.15: 64921986-64921986 on Build GRCh38
rs540297470 Chr.15: 64922052-64922052 on Build GRCh38
rs952981381 Chr.15: 64922130-64922130 on Build GRCh38
rs748539051 Chr.15: 64922038-64922044 on Build GRCh38
rs1054394659 Chr.15: 64922100-64922100 on Build GRCh38
rs778701569 Chr.15: 64921958-64921958 on Build GRCh38
rs746468585 Chr.15: 64921877-64921877 on Build GRCh38
rs564877484 Chr.15: 64922072-64922072 on Build GRCh38
rs372452058 Chr.15: 64922055-64922055 on Build GRCh38
rs750196539 Chr.15: 64921960-64921960 on Build GRCh38
rs768419675 Chr.15: 64921893-64921893 on Build GRCh38
rs936754571 Chr.15: 64922275-64922275 on Build GRCh38
rs760659314 Chr.15: 64921952-64921952 on Build GRCh38
rs371881411 Chr.15: 64922039-64922039 on Build GRCh38
rs766137636 Chr.15: 64922057-64922057 on Build GRCh38
rs752892501 Chr.15: 64921944-64921944 on Build GRCh38
rs755449435 Chr.15: 64921997-64921998 on Build GRCh38
rs903207768 Chr.15: 64922073-64922073 on Build GRCh38
rs758112448 Chr.15: 64922053-64922053 on Build GRCh38
rs200840260 Chr.15: 64921923-64921923 on Build GRCh38
rs138202030 Chr.15: 64921966-64921966 on Build GRCh38
rs57353708 Chr.15: 64922117-64922117 on Build GRCh38
rs764555472 Chr.15: 64921946-64921946 on Build GRCh38
rs915846710 Chr.15: 64921978-64921978 on Build GRCh38
rs1041169151 Chr.15: 64922077-64922077 on Build GRCh38
rs759977930 Chr.15: 64921996-64921996 on Build GRCh38
rs764178898 Chr.15: 64921956-64921956 on Build GRCh38
rs78734947 Chr.15: 64922104-64922104 on Build GRCh38
rs776132762 Chr.15: 64921905-64921905 on Build GRCh38
rs563949657 Chr.15: 64922125-64922125 on Build GRCh38
rs1033457620 Chr.15: 64922099-64922099 on Build GRCh38
rs762070383 Chr.15: 64922048-64922048 on Build GRCh38
rs145919740 Chr.15: 64921934-64921934 on Build GRCh38
rs754901202 Chr.15: 64922064-64922064 on Build GRCh38
rs984008233 Chr.15: 64922111-64922111 on Build GRCh38
rs114602209 Chr.15: 64921872-64921872 on Build GRCh38
rs765411845 Chr.15: 64922049-64922049 on Build GRCh38
rs889246179 Chr.15: 64922250-64922250 on Build GRCh38
rs952644042 Chr.15: 64922106-64922106 on Build GRCh38
rs774900398 Chr.15: 64921993-64921993 on Build GRCh38
rs758369266 Chr.15: 64921962-64921962 on Build GRCh38
rs780682731 Chr.15: 64921974-64921974 on Build GRCh38
rs781702512 Chr.15: 64921999-64922002 on Build GRCh38
rs115268452 Chr.15: 64921914-64921914 on Build GRCh38
rs1050462910 Chr.15: 64922216-64922218 on Build GRCh38
rs745836587 Chr.15: 64921870-64921870 on Build GRCh38
rs764112527 Chr.15: 64922025-64922025 on Build GRCh38
rs751452049 Chr.15: 64922060-64922060 on Build GRCh38
rs751965693 Chr.15: 64921914-64921914 on Build GRCh38
rs1053925766 Chr.15: 64922288-64922288 on Build GRCh38
rs779661994 Chr.15: 64921876-64921876 on Build GRCh38

View Full Product Details