Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 64926797-64927287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKDD1A
Entrez Gene ID
348094
Gene Name
ankyrin repeat and death domain containing 1A
Gene Aliases
-
UniGene
Hs.207157
Gene Chromosome Location
Chr.15: 64911902-64958700 on Build GRCh38
Transcripts
NM_182703.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5043046 c.484G>T p.A162S Chr.15: 64926913-64926913 on Build GRCh38
3503118 c.261C>T p.L87L Chr.15: 64926963-64926963 on Build GRCh38
3786549 c.234C>T p.H78H Chr.15: 64926936-64926936 on Build GRCh38
5825914 c.485C>T p.A162V Chr.15: 64926914-64926914 on Build GRCh38
3786548 c.507C>T p.H169H Chr.15: 64926936-64926936 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200317955 Chr.15: 64926949-64926949 on Build GRCh38
rs982034780 Chr.15: 64927171-64927171 on Build GRCh38
rs200097861 Chr.15: 64926963-64926963 on Build GRCh38
rs990448867 Chr.15: 64927225-64927225 on Build GRCh38
rs762640114 Chr.15: 64926931-64926931 on Build GRCh38
rs1034545708 Chr.15: 64927174-64927174 on Build GRCh38
rs144154294 Chr.15: 64926889-64926889 on Build GRCh38
rs538724850 Chr.15: 64927059-64927059 on Build GRCh38
rs950119475 Chr.15: 64927232-64927232 on Build GRCh38
rs199532025 Chr.15: 64927023-64927023 on Build GRCh38
rs745913897 Chr.15: 64926945-64926945 on Build GRCh38
rs750587502 Chr.15: 64927025-64927025 on Build GRCh38
rs550198020 Chr.15: 64926882-64926882 on Build GRCh38
rs548032723 Chr.15: 64927193-64927193 on Build GRCh38
rs944557928 Chr.15: 64927100-64927100 on Build GRCh38
rs566230605 Chr.15: 64927214-64927214 on Build GRCh38
rs531962954 Chr.15: 64927084-64927084 on Build GRCh38
rs749576103 Chr.15: 64926905-64926905 on Build GRCh38
rs182519632 Chr.15: 64927166-64927166 on Build GRCh38
rs374105135 Chr.15: 64927010-64927010 on Build GRCh38
rs762440455 Chr.15: 64927001-64927001 on Build GRCh38
rs925121107 Chr.15: 64927182-64927182 on Build GRCh38
rs1044032920 Chr.15: 64926975-64926975 on Build GRCh38
rs192904448 Chr.15: 64927008-64927008 on Build GRCh38
rs554342619 Chr.15: 64926915-64926915 on Build GRCh38
rs557963917 Chr.15: 64927236-64927236 on Build GRCh38
rs746116516 Chr.15: 64926895-64926895 on Build GRCh38
rs1001673427 Chr.15: 64927074-64927074 on Build GRCh38
rs1042156677 Chr.15: 64927096-64927096 on Build GRCh38
rs758001334 Chr.15: 64926862-64926862 on Build GRCh38
rs374429278 Chr.15: 64927022-64927022 on Build GRCh38
rs780408018 Chr.15: 64926955-64926955 on Build GRCh38
rs775467057 Chr.15: 64926899-64926899 on Build GRCh38
rs914940069 Chr.15: 64927089-64927089 on Build GRCh38
rs565411229 Chr.15: 64927164-64927164 on Build GRCh38
rs887817311 Chr.15: 64927140-64927140 on Build GRCh38
rs905914955 Chr.15: 64927241-64927241 on Build GRCh38
rs527665223 Chr.15: 64927183-64927183 on Build GRCh38
rs878998843 Chr.15: 64927114-64927114 on Build GRCh38
rs906024238 Chr.15: 64927065-64927065 on Build GRCh38
rs563021852 Chr.15: 64927060-64927060 on Build GRCh38
rs750091312 Chr.15: 64926854-64926854 on Build GRCh38
rs758564155 Chr.15: 64927027-64927027 on Build GRCh38
rs1052380367 Chr.15: 64927185-64927185 on Build GRCh38
rs563336858 Chr.15: 64927093-64927093 on Build GRCh38
rs201127218 Chr.15: 64926937-64926937 on Build GRCh38
rs765820682 Chr.15: 64927004-64927004 on Build GRCh38
rs747170042 Chr.15: 64926957-64926957 on Build GRCh38
rs530420524 Chr.15: 64927099-64927099 on Build GRCh38
rs572588342 Chr.15: 64926928-64926928 on Build GRCh38
rs760540390 Chr.15: 64926906-64926906 on Build GRCh38
rs753034410 Chr.15: 64926883-64926883 on Build GRCh38
rs927549692 Chr.15: 64927076-64927076 on Build GRCh38
rs187320003 Chr.15: 64927235-64927235 on Build GRCh38
rs145050267 Chr.15: 64926968-64926968 on Build GRCh38
rs200881619 Chr.15: 64927009-64927009 on Build GRCh38
rs971527024 Chr.15: 64927167-64927167 on Build GRCh38
rs142085847 Chr.15: 64927110-64927110 on Build GRCh38
rs996138610 Chr.15: 64927138-64927138 on Build GRCh38
rs765982673 Chr.15: 64926936-64926936 on Build GRCh38
rs139437993 Chr.15: 64926964-64926964 on Build GRCh38
rs946458915 Chr.15: 64927094-64927094 on Build GRCh38
rs1042187404 Chr.15: 64926960-64926960 on Build GRCh38
rs757656828 Chr.15: 64926942-64926942 on Build GRCh38
rs375134364 Chr.15: 64926852-64926852 on Build GRCh38
rs761588607 Chr.15: 64926914-64926914 on Build GRCh38
rs774000189 Chr.15: 64927005-64927005 on Build GRCh38
rs757802548 Chr.15: 64926946-64926946 on Build GRCh38
rs976227861 Chr.15: 64927109-64927109 on Build GRCh38
rs148599136 Chr.15: 64927083-64927083 on Build GRCh38
rs769594085 Chr.15: 64926981-64926981 on Build GRCh38
rs772745997 Chr.15: 64926986-64926986 on Build GRCh38
rs148678562 Chr.15: 64926911-64926911 on Build GRCh38
rs200670478 Chr.15: 64927011-64927011 on Build GRCh38
rs147441878 Chr.15: 64926943-64926943 on Build GRCh38
rs144515181 Chr.15: 64926912-64926912 on Build GRCh38
rs891795106 Chr.15: 64927224-64927224 on Build GRCh38
rs772403955 Chr.15: 64926897-64926897 on Build GRCh38
rs375221475 Chr.15: 64926894-64926894 on Build GRCh38
rs201766403 Chr.15: 64926952-64926952 on Build GRCh38
rs756683458 Chr.15: 64926887-64926887 on Build GRCh38
rs568645848 Chr.15: 64927111-64927111 on Build GRCh38
rs553529071 Chr.15: 64927258-64927258 on Build GRCh38
rs1028062383 Chr.15: 64927139-64927139 on Build GRCh38
rs143698274 Chr.15: 64926888-64926888 on Build GRCh38
rs559092473 Chr.15: 64927062-64927062 on Build GRCh38

View Full Product Details