Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65663973-65664523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND4A
Entrez Gene ID
10260
Gene Name
DENN domain containing 4A
Gene Aliases
IRLB, MYCPBP
UniGene
Hs.654567
Gene Chromosome Location
Chr.15: 65659123-65792293 on Build GRCh38
Transcripts
NM_001144823.2 NM_001320835.1 NM_005848.4 XM_005254120.4 XM_005254121.3 XM_005254122.4 XM_006720371.3 XM_011521157.2 XM_017021862.1 XM_017021863.1 XM_017021865.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
964107 c.5404G>T p.E1802* Chr.15: 65664381-65664381 on Build GRCh38
4056336 c.5520-2A>G p.? Chr.15: 65664396-65664396 on Build GRCh38
1588043 c.5533G>T p.E1845* Chr.15: 65664381-65664381 on Build GRCh38
2256745 c.5569G>A p.E1857K Chr.15: 65664345-65664345 on Build GRCh38
4056337 c.5391-2A>G p.? Chr.15: 65664396-65664396 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953466057 Chr.15: 65664478-65664478 on Build GRCh38
rs146675846 Chr.15: 65664146-65664146 on Build GRCh38
rs763127759 Chr.15: 65664338-65664338 on Build GRCh38
rs796829774 Chr.15: 65664267-65664269 on Build GRCh38
rs984855035 Chr.15: 65664492-65664492 on Build GRCh38
rs117756005 Chr.15: 65664012-65664012 on Build GRCh38
rs751549830 Chr.15: 65664367-65664367 on Build GRCh38
rs752545903 Chr.15: 65664413-65664413 on Build GRCh38
rs773418411 Chr.15: 65664314-65664314 on Build GRCh38
rs545905566 Chr.15: 65664324-65664324 on Build GRCh38
rs759435120 Chr.15: 65664393-65664393 on Build GRCh38
rs956402231 Chr.15: 65664138-65664138 on Build GRCh38
rs939021344 Chr.15: 65664050-65664050 on Build GRCh38
rs1031940175 Chr.15: 65664126-65664126 on Build GRCh38
rs769923457 Chr.15: 65664302-65664302 on Build GRCh38
rs545361293 Chr.15: 65664254-65664256 on Build GRCh38
rs72625763 Chr.15: 65664267-65664267 on Build GRCh38
rs940714248 Chr.15: 65664461-65664461 on Build GRCh38
rs540940127 Chr.15: 65664088-65664088 on Build GRCh38
rs766180344 Chr.15: 65664359-65664359 on Build GRCh38
rs1031255443 Chr.15: 65664209-65664209 on Build GRCh38
rs1028834387 Chr.15: 65664079-65664079 on Build GRCh38
rs1029432963 Chr.15: 65664398-65664398 on Build GRCh38
rs777408080 Chr.15: 65664444-65664444 on Build GRCh38
rs777800018 Chr.15: 65664403-65664404 on Build GRCh38
rs904028868 Chr.15: 65664157-65664157 on Build GRCh38
rs770195525 Chr.15: 65664293-65664293 on Build GRCh38
rs997795875 Chr.15: 65664074-65664074 on Build GRCh38
rs755760712 Chr.15: 65664441-65664441 on Build GRCh38
rs555563272 Chr.15: 65664059-65664059 on Build GRCh38
rs577254426 Chr.15: 65664090-65664090 on Build GRCh38
rs8033091 Chr.15: 65664060-65664060 on Build GRCh38
rs563444672 Chr.15: 65664405-65664405 on Build GRCh38
rs918215968 Chr.15: 65664008-65664008 on Build GRCh38
rs997514130 Chr.15: 65664379-65664379 on Build GRCh38
rs953380495 Chr.15: 65664092-65664092 on Build GRCh38
rs979335967 Chr.15: 65664118-65664118 on Build GRCh38
rs202146741 Chr.15: 65664265-65664265 on Build GRCh38
rs1056558896 Chr.15: 65664058-65664058 on Build GRCh38
rs981982456 Chr.15: 65664262-65664262 on Build GRCh38
rs999738957 Chr.15: 65664192-65664192 on Build GRCh38
rs533207008 Chr.15: 65664246-65664246 on Build GRCh38
rs372297633 Chr.15: 65664410-65664410 on Build GRCh38
rs57260104 Chr.15: 65664265-65664267 on Build GRCh38

View Full Product Details