Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65667668-65668206 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND4A
Entrez Gene ID
10260
Gene Name
DENN domain containing 4A
Gene Aliases
IRLB, MYCPBP
UniGene
Hs.654567
Gene Chromosome Location
Chr.15: 65659123-65792293 on Build GRCh38
Transcripts
NM_001144823.2 NM_001320835.1 NM_005848.4 XM_005254120.4 XM_005254121.3 XM_005254122.4 XM_006720371.3 XM_011521157.2 XM_017021862.1 XM_017021863.1 XM_017021864.1 XM_017021865.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5893811 c.4801G>A p.E1601K Chr.15: 65668107-65668107 on Build GRCh38
6394536 c.4814C>T p.T1605I Chr.15: 65668094-65668094 on Build GRCh38
5817158 c.4856A>G p.D1619G Chr.15: 65667702-65667702 on Build GRCh38
3816586 c.4988C>A p.S1663Y Chr.15: 65667699-65667699 on Build GRCh38
3816587 c.4859C>A p.S1620Y Chr.15: 65667699-65667699 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191718975 Chr.15: 65667880-65667880 on Build GRCh38
rs767483799 Chr.15: 65667985-65667985 on Build GRCh38
rs893417239 Chr.15: 65668088-65668088 on Build GRCh38
rs751580307 Chr.15: 65667739-65667739 on Build GRCh38
rs773378189 Chr.15: 65667959-65667959 on Build GRCh38
rs920545637 Chr.15: 65667850-65667850 on Build GRCh38
rs762419834 Chr.15: 65667696-65667696 on Build GRCh38
rs748015562 Chr.15: 65667876-65667876 on Build GRCh38
rs746530544 Chr.15: 65668091-65668091 on Build GRCh38
rs149498131 Chr.15: 65667730-65667730 on Build GRCh38
rs767785579 Chr.15: 65668170-65668170 on Build GRCh38
rs749046554 Chr.15: 65668034-65668034 on Build GRCh38
rs762210262 Chr.15: 65667933-65667933 on Build GRCh38
rs754336159 Chr.15: 65667694-65667694 on Build GRCh38
rs765836181 Chr.15: 65667947-65667947 on Build GRCh38
rs773730669 Chr.15: 65668096-65668096 on Build GRCh38
rs1045875852 Chr.15: 65667924-65667924 on Build GRCh38
rs755938432 Chr.15: 65668023-65668023 on Build GRCh38
rs371610511 Chr.15: 65667829-65667829 on Build GRCh38
rs770141369 Chr.15: 65668092-65668092 on Build GRCh38
rs144052781 Chr.15: 65668058-65668058 on Build GRCh38
rs6494531 Chr.15: 65667857-65667857 on Build GRCh38
rs186836490 Chr.15: 65667748-65667748 on Build GRCh38
rs770653639 Chr.15: 65667913-65667913 on Build GRCh38
rs561615981 Chr.15: 65667786-65667786 on Build GRCh38
rs755231924 Chr.15: 65667982-65667982 on Build GRCh38
rs201297991 Chr.15: 65667921-65667921 on Build GRCh38
rs746791411 Chr.15: 65667764-65667764 on Build GRCh38
rs763261526 Chr.15: 65668097-65668097 on Build GRCh38
rs757151542 Chr.15: 65668038-65668038 on Build GRCh38
rs1020717748 Chr.15: 65668098-65668098 on Build GRCh38
rs759160168 Chr.15: 65667768-65667768 on Build GRCh38
rs183508766 Chr.15: 65668029-65668029 on Build GRCh38
rs771679645 Chr.15: 65667925-65667925 on Build GRCh38
rs946925608 Chr.15: 65667781-65667781 on Build GRCh38
rs774756011 Chr.15: 65668163-65668163 on Build GRCh38
rs752890550 Chr.15: 65667997-65667997 on Build GRCh38
rs750854266 Chr.15: 65667725-65667725 on Build GRCh38
rs749100391 Chr.15: 65667884-65667884 on Build GRCh38
rs763320337 Chr.15: 65667965-65667965 on Build GRCh38
rs766538090 Chr.15: 65667968-65667968 on Build GRCh38
rs746694011 Chr.15: 65668158-65668158 on Build GRCh38
rs778272150 Chr.15: 65667692-65667692 on Build GRCh38
rs774974300 Chr.15: 65668165-65668169 on Build GRCh38
rs560689503 Chr.15: 65668063-65668063 on Build GRCh38
rs751809446 Chr.15: 65667970-65667970 on Build GRCh38
rs759752230 Chr.15: 65668164-65668164 on Build GRCh38
rs986750318 Chr.15: 65668043-65668043 on Build GRCh38
rs1047872460 Chr.15: 65667863-65667863 on Build GRCh38
rs778895336 Chr.15: 65668049-65668049 on Build GRCh38
rs987297782 Chr.15: 65667766-65667766 on Build GRCh38
rs774172407 Chr.15: 65667914-65667914 on Build GRCh38
rs752663197 Chr.15: 65668172-65668172 on Build GRCh38
rs756136327 Chr.15: 65667879-65667879 on Build GRCh38
rs1056843759 Chr.15: 65667807-65667807 on Build GRCh38
rs886175689 Chr.15: 65667886-65667886 on Build GRCh38
rs772623561 Chr.15: 65668135-65668135 on Build GRCh38
rs367897260 Chr.15: 65667992-65667992 on Build GRCh38
rs758609921 Chr.15: 65667727-65667727 on Build GRCh38
rs943256164 Chr.15: 65667802-65667802 on Build GRCh38
rs775112024 Chr.15: 65667930-65667930 on Build GRCh38
rs765494675 Chr.15: 65667707-65667707 on Build GRCh38
rs200310738 Chr.15: 65667742-65667742 on Build GRCh38
rs902731139 Chr.15: 65667800-65667800 on Build GRCh38
rs945824994 Chr.15: 65668173-65668173 on Build GRCh38
rs766697327 Chr.15: 65668150-65668150 on Build GRCh38
rs939620683 Chr.15: 65667806-65667806 on Build GRCh38

View Full Product Details