Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 66294909-66295417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIS3L
Entrez Gene ID
115752
Gene Name
DIS3 like exosome 3'-5' exoribonuclease
Gene Aliases
DIS3L1
UniGene
Hs.446251
Gene Chromosome Location
Chr.15: 66293257-66333898 on Build GRCh38
Transcripts
NM_001143688.2 NM_001323936.1 NM_001323937.1 NM_001323938.1 NM_001323939.1 NM_001323940.1 NM_001323941.1 NM_001323943.1 NM_001323944.1 NM_001323945.1 NM_001323946.1 NM_001323948.1 NM_133375.4 XM_005254146.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6015829 c.265C>T p.Q89* Chr.15: 66295113-66295113 on Build GRCh38
5920806 c.220G>A p.E74K Chr.15: 66295068-66295068 on Build GRCh38
1203648 c.268G>T p.A90S Chr.15: 66295116-66295116 on Build GRCh38
3794341 c.265C>G p.Q89E Chr.15: 66295113-66295113 on Build GRCh38
964174 c.35G>A p.R12K Chr.15: 66295132-66295132 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769755641 Chr.15: 66295023-66295023 on Build GRCh38
rs897514202 Chr.15: 66295303-66295303 on Build GRCh38
rs999773548 Chr.15: 66295328-66295328 on Build GRCh38
rs759126986 Chr.15: 66295073-66295073 on Build GRCh38
rs375683090 Chr.15: 66294968-66294968 on Build GRCh38
rs921081070 Chr.15: 66295216-66295216 on Build GRCh38
rs748235246 Chr.15: 66295189-66295189 on Build GRCh38
rs761805530 Chr.15: 66295017-66295017 on Build GRCh38
rs895467366 Chr.15: 66295383-66295383 on Build GRCh38
rs199880913 Chr.15: 66295263-66295264 on Build GRCh38
rs575116022 Chr.15: 66295082-66295082 on Build GRCh38
rs769023700 Chr.15: 66295006-66295006 on Build GRCh38
rs560686161 Chr.15: 66295202-66295202 on Build GRCh38
rs558641168 Chr.15: 66294948-66294948 on Build GRCh38
rs545093772 Chr.15: 66295378-66295378 on Build GRCh38
rs747194548 Chr.15: 66295005-66295005 on Build GRCh38
rs569780371 Chr.15: 66295293-66295293 on Build GRCh38
rs79939593 Chr.15: 66294969-66294969 on Build GRCh38
rs757194957 Chr.15: 66295277-66295277 on Build GRCh38
rs375124003 Chr.15: 66295061-66295061 on Build GRCh38
rs902725442 Chr.15: 66295283-66295283 on Build GRCh38
rs752244737 Chr.15: 66295133-66295133 on Build GRCh38
rs750936205 Chr.15: 66295152-66295152 on Build GRCh38
rs777586613 Chr.15: 66295261-66295261 on Build GRCh38
rs929977095 Chr.15: 66295010-66295010 on Build GRCh38
rs76770477 Chr.15: 66295141-66295141 on Build GRCh38
rs373325779 Chr.15: 66295158-66295158 on Build GRCh38
rs188177866 Chr.15: 66295022-66295022 on Build GRCh38
rs780395916 Chr.15: 66295002-66295002 on Build GRCh38
rs912487829 Chr.15: 66295129-66295130 on Build GRCh38
rs111304305 Chr.15: 66295159-66295159 on Build GRCh38
rs776796518 Chr.15: 66295009-66295009 on Build GRCh38
rs766157127 Chr.15: 66294996-66294996 on Build GRCh38
rs767159031 Chr.15: 66295088-66295088 on Build GRCh38
rs765599287 Chr.15: 66295159-66295163 on Build GRCh38
rs779358711 Chr.15: 66294981-66294981 on Build GRCh38
rs924627006 Chr.15: 66295147-66295147 on Build GRCh38
rs773071004 Chr.15: 66295041-66295041 on Build GRCh38
rs557876063 Chr.15: 66294994-66294994 on Build GRCh38
rs751203338 Chr.15: 66295071-66295071 on Build GRCh38
rs776907056 Chr.15: 66294966-66294967 on Build GRCh38
rs757714248 Chr.15: 66294980-66294980 on Build GRCh38
rs969249741 Chr.15: 66295042-66295042 on Build GRCh38
rs762366336 Chr.15: 66295023-66295023 on Build GRCh38
rs754652586 Chr.15: 66294962-66294962 on Build GRCh38
rs1035955623 Chr.15: 66295336-66295336 on Build GRCh38
rs537875202 Chr.15: 66294970-66294970 on Build GRCh38
rs923145534 Chr.15: 66295185-66295185 on Build GRCh38
rs1056764210 Chr.15: 66295361-66295361 on Build GRCh38
rs768934011 Chr.15: 66295174-66295174 on Build GRCh38
rs947189035 Chr.15: 66295240-66295240 on Build GRCh38
rs779456791 Chr.15: 66295148-66295148 on Build GRCh38
rs77920095 Chr.15: 66295143-66295143 on Build GRCh38
rs370066824 Chr.15: 66295166-66295166 on Build GRCh38
rs762777416 Chr.15: 66295058-66295058 on Build GRCh38
rs752741218 Chr.15: 66295211-66295211 on Build GRCh38
rs80126717 Chr.15: 66295243-66295243 on Build GRCh38
rs781337568 Chr.15: 66295184-66295184 on Build GRCh38
rs780565926 Chr.15: 66295168-66295168 on Build GRCh38
rs751373542 Chr.15: 66295069-66295069 on Build GRCh38
rs993125455 Chr.15: 66295322-66295322 on Build GRCh38
rs762003538 Chr.15: 66294975-66294975 on Build GRCh38

View Full Product Details