Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 67203903-67204327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAGAB
Entrez Gene ID
79719
Gene Name
alpha- and gamma-adaptin binding protein
Gene Aliases
KPPP1, PPKP1, PPKP1A, p34
UniGene
Hs.254642
Gene Chromosome Location
Chr.15: 67200675-67255198 on Build GRCh38
Transcripts
NM_001271885.1 NM_001271886.1 NM_024666.4 XM_011522020.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3361497 c.754C>T p.L252F Chr.15: 67204110-67204110 on Build GRCh38
1374225 c.782A>G p.N261S Chr.15: 67204082-67204082 on Build GRCh38
2257433 c.744A>T p.E248D Chr.15: 67204120-67204120 on Build GRCh38
6432454 c.727G>T p.D243Y Chr.15: 67204137-67204137 on Build GRCh38
1749287 c.780G>A p.E260E Chr.15: 67204084-67204084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1010715115 Chr.15: 67204174-67204174 on Build GRCh38
rs749583839 Chr.15: 67204081-67204081 on Build GRCh38
rs564451900 Chr.15: 67203985-67203985 on Build GRCh38
rs759490495 Chr.15: 67204251-67204251 on Build GRCh38
rs755480469 Chr.15: 67204027-67204027 on Build GRCh38
rs764806559 Chr.15: 67204134-67204134 on Build GRCh38
rs970113502 Chr.15: 67204283-67204283 on Build GRCh38
rs367555610 Chr.15: 67204050-67204050 on Build GRCh38
rs777838692 Chr.15: 67204067-67204067 on Build GRCh38
rs960581588 Chr.15: 67204232-67204232 on Build GRCh38
rs765401006 Chr.15: 67204104-67204105 on Build GRCh38
rs776392886 Chr.15: 67203999-67203999 on Build GRCh38
rs148271197 Chr.15: 67203962-67203962 on Build GRCh38
rs375444996 Chr.15: 67204137-67204137 on Build GRCh38
rs371685408 Chr.15: 67204089-67204089 on Build GRCh38
rs141177071 Chr.15: 67204180-67204180 on Build GRCh38
rs118158950 Chr.15: 67203989-67203989 on Build GRCh38
rs748943571 Chr.15: 67203995-67203995 on Build GRCh38
rs915935476 Chr.15: 67204277-67204277 on Build GRCh38
rs781299385 Chr.15: 67204284-67204285 on Build GRCh38
rs965413939 Chr.15: 67204204-67204204 on Build GRCh38
rs372325047 Chr.15: 67204120-67204120 on Build GRCh38
rs756536786 Chr.15: 67204155-67204155 on Build GRCh38
rs768110809 Chr.15: 67204151-67204151 on Build GRCh38
rs1031900548 Chr.15: 67204230-67204230 on Build GRCh38
rs753310459 Chr.15: 67204154-67204154 on Build GRCh38
rs975548724 Chr.15: 67203950-67203950 on Build GRCh38
rs146169842 Chr.15: 67204190-67204190 on Build GRCh38
rs572298456 Chr.15: 67204202-67204202 on Build GRCh38
rs375593998 Chr.15: 67204049-67204049 on Build GRCh38
rs143201986 Chr.15: 67203948-67203948 on Build GRCh38
rs992775995 Chr.15: 67204271-67204271 on Build GRCh38
rs368693236 Chr.15: 67204142-67204142 on Build GRCh38
rs764587719 Chr.15: 67204173-67204173 on Build GRCh38
rs745960635 Chr.15: 67204103-67204103 on Build GRCh38
rs775445355 Chr.15: 67204110-67204110 on Build GRCh38
rs761459547 Chr.15: 67204123-67204123 on Build GRCh38
rs931214910 Chr.15: 67204098-67204098 on Build GRCh38
rs867395597 Chr.15: 67204125-67204125 on Build GRCh38
rs1046940979 Chr.15: 67204100-67204100 on Build GRCh38
rs1028592136 Chr.15: 67204243-67204243 on Build GRCh38
rs756589595 Chr.15: 67204066-67204066 on Build GRCh38
rs939736095 Chr.15: 67204039-67204039 on Build GRCh38
rs867797999 Chr.15: 67204146-67204146 on Build GRCh38
rs767464285 Chr.15: 67204065-67204066 on Build GRCh38
rs774644135 Chr.15: 67204195-67204195 on Build GRCh38
rs919809911 Chr.15: 67204097-67204097 on Build GRCh38
rs915968056 Chr.15: 67204003-67204003 on Build GRCh38
rs760748779 Chr.15: 67204092-67204094 on Build GRCh38
rs1046128547 Chr.15: 67204045-67204045 on Build GRCh38
rs767852348 Chr.15: 67204269-67204269 on Build GRCh38
rs369410834 Chr.15: 67204116-67204116 on Build GRCh38
rs969376188 Chr.15: 67203955-67203955 on Build GRCh38
rs912549905 Chr.15: 67203944-67203944 on Build GRCh38
rs772953729 Chr.15: 67203974-67203974 on Build GRCh38
rs752656407 Chr.15: 67204065-67204065 on Build GRCh38
rs761214742 Chr.15: 67204009-67204009 on Build GRCh38
rs754158336 Chr.15: 67204176-67204176 on Build GRCh38
rs998889315 Chr.15: 67204200-67204200 on Build GRCh38
rs953058266 Chr.15: 67204284-67204284 on Build GRCh38
rs746782588 Chr.15: 67204112-67204112 on Build GRCh38
rs750458790 Chr.15: 67204200-67204203 on Build GRCh38
rs770852267 Chr.15: 67204025-67204026 on Build GRCh38
rs778984821 Chr.15: 67204094-67204094 on Build GRCh38
rs769461133 Chr.15: 67203957-67203957 on Build GRCh38
rs376820468 Chr.15: 67204107-67204107 on Build GRCh38
rs1044858345 Chr.15: 67204096-67204096 on Build GRCh38
rs768157646 Chr.15: 67204038-67204038 on Build GRCh38
rs764866327 Chr.15: 67204023-67204023 on Build GRCh38
rs975689227 Chr.15: 67203952-67203952 on Build GRCh38
rs975789741 Chr.15: 67204223-67204223 on Build GRCh38
rs749904131 Chr.15: 67204026-67204026 on Build GRCh38
rs938657650 Chr.15: 67203988-67203988 on Build GRCh38

View Full Product Details