Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 67231808-67232323 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAGAB
Entrez Gene ID
79719
Gene Name
alpha- and gamma-adaptin binding protein
Gene Aliases
KPPP1, PPKP1, PPKP1A, p34
UniGene
Hs.254642
Gene Chromosome Location
Chr.15: 67200675-67255198 on Build GRCh38
Transcripts
NM_001271885.1 NM_001271886.1 NM_024666.4 XM_011522020.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5893806 c.460C>T p.P154S Chr.15: 67231889-67231889 on Build GRCh38
278485 c.463G>T p.E155* Chr.15: 67231886-67231886 on Build GRCh38
470991 c.482G>A p.R161Q Chr.15: 67231867-67231867 on Build GRCh38
3503369 c.498G>A p.L166L Chr.15: 67231851-67231851 on Build GRCh38
4495703 c.459C>T p.F153F Chr.15: 67231890-67231890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369008165 Chr.15: 67231874-67231874 on Build GRCh38
rs915741397 Chr.15: 67231951-67231951 on Build GRCh38
rs534437770 Chr.15: 67232181-67232181 on Build GRCh38
rs900835146 Chr.15: 67232231-67232231 on Build GRCh38
rs949090056 Chr.15: 67231954-67231954 on Build GRCh38
rs979965235 Chr.15: 67232279-67232279 on Build GRCh38
rs1006417643 Chr.15: 67232093-67232093 on Build GRCh38
rs112308565 Chr.15: 67231961-67231961 on Build GRCh38
rs779092857 Chr.15: 67231898-67231898 on Build GRCh38
rs779782680 Chr.15: 67231867-67231867 on Build GRCh38
rs536640776 Chr.15: 67232203-67232203 on Build GRCh38
rs1042942346 Chr.15: 67231992-67231992 on Build GRCh38
rs778164504 Chr.15: 67232249-67232250 on Build GRCh38
rs936578962 Chr.15: 67231941-67231941 on Build GRCh38
rs757112292 Chr.15: 67231842-67231842 on Build GRCh38
rs1028531830 Chr.15: 67232070-67232070 on Build GRCh38
rs554620003 Chr.15: 67232128-67232128 on Build GRCh38
rs749862948 Chr.15: 67231885-67231885 on Build GRCh38
rs143805221 Chr.15: 67232043-67232043 on Build GRCh38
rs753883773 Chr.15: 67231841-67231841 on Build GRCh38
rs771436681 Chr.15: 67231889-67231889 on Build GRCh38
rs1034513645 Chr.15: 67232282-67232282 on Build GRCh38
rs374671581 Chr.15: 67231856-67231856 on Build GRCh38
rs781596375 Chr.15: 67231876-67231876 on Build GRCh38
rs776854267 Chr.15: 67231922-67231922 on Build GRCh38
rs958139186 Chr.15: 67232127-67232127 on Build GRCh38
rs568829964 Chr.15: 67232122-67232122 on Build GRCh38
rs188013300 Chr.15: 67232185-67232185 on Build GRCh38
rs988487963 Chr.15: 67231949-67231949 on Build GRCh38
rs546464690 Chr.15: 67231920-67231920 on Build GRCh38
rs934306678 Chr.15: 67232016-67232016 on Build GRCh38
rs902182919 Chr.15: 67231957-67231958 on Build GRCh38
rs991580363 Chr.15: 67232130-67232130 on Build GRCh38
rs750244758 Chr.15: 67231854-67231854 on Build GRCh38
rs373141411 Chr.15: 67231879-67231879 on Build GRCh38
rs567585902 Chr.15: 67232079-67232079 on Build GRCh38
rs957152674 Chr.15: 67232284-67232284 on Build GRCh38
rs746266917 Chr.15: 67231900-67231900 on Build GRCh38
rs377192980 Chr.15: 67231904-67231904 on Build GRCh38
rs34877999 Chr.15: 67232250-67232251 on Build GRCh38
rs932344567 Chr.15: 67232254-67232254 on Build GRCh38
rs146351661 Chr.15: 67232123-67232123 on Build GRCh38
rs962301436 Chr.15: 67232105-67232105 on Build GRCh38
rs764092859 Chr.15: 67231839-67231839 on Build GRCh38
rs1016501487 Chr.15: 67232097-67232097 on Build GRCh38
rs926304194 Chr.15: 67232221-67232221 on Build GRCh38
rs903133839 Chr.15: 67232000-67232000 on Build GRCh38
rs937751527 Chr.15: 67232280-67232280 on Build GRCh38
rs748650736 Chr.15: 67231915-67231918 on Build GRCh38
rs1048526403 Chr.15: 67232022-67232022 on Build GRCh38
rs546049633 Chr.15: 67232290-67232290 on Build GRCh38
rs568574741 Chr.15: 67232146-67232146 on Build GRCh38
rs913860705 Chr.15: 67232133-67232133 on Build GRCh38
rs985950921 Chr.15: 67231893-67231893 on Build GRCh38
rs373768307 Chr.15: 67231929-67231929 on Build GRCh38
rs746488412 Chr.15: 67231868-67231868 on Build GRCh38
rs1040729559 Chr.15: 67232041-67232041 on Build GRCh38
rs376314046 Chr.15: 67232044-67232044 on Build GRCh38
rs1028337685 Chr.15: 67232092-67232101 on Build GRCh38
rs972022765 Chr.15: 67232114-67232114 on Build GRCh38
rs1049471570 Chr.15: 67232272-67232272 on Build GRCh38
rs372734262 Chr.15: 67231882-67231882 on Build GRCh38
rs949306443 Chr.15: 67232184-67232184 on Build GRCh38
rs904911618 Chr.15: 67231983-67231983 on Build GRCh38
rs888628274 Chr.15: 67232027-67232027 on Build GRCh38
rs1041071351 Chr.15: 67232207-67232207 on Build GRCh38
rs947205331 Chr.15: 67232162-67232162 on Build GRCh38
rs577163033 Chr.15: 67232202-67232202 on Build GRCh38
rs761783564 Chr.15: 67231928-67231928 on Build GRCh38
rs370777101 Chr.15: 67231911-67231911 on Build GRCh38
rs552242337 Chr.15: 67232076-67232076 on Build GRCh38
rs764873967 Chr.15: 67231843-67231843 on Build GRCh38
rs760818075 Chr.15: 67231918-67231918 on Build GRCh38
rs536725304 Chr.15: 67232121-67232121 on Build GRCh38
rs398039462 Chr.15: 67232266-67232267 on Build GRCh38
rs1035695352 Chr.15: 67232119-67232119 on Build GRCh38
rs755738808 Chr.15: 67231880-67231880 on Build GRCh38
rs750140875 Chr.15: 67231947-67231947 on Build GRCh38
rs757934884 Chr.15: 67231865-67231865 on Build GRCh38

View Full Product Details