Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 67236217-67236723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAGAB
Entrez Gene ID
79719
Gene Name
alpha- and gamma-adaptin binding protein
Gene Aliases
KPPP1, PPKP1, PPKP1A, p34
UniGene
Hs.254642
Gene Chromosome Location
Chr.15: 67200675-67255198 on Build GRCh38
Transcripts
NM_001271885.1 NM_001271886.1 NM_024666.4 XM_011522020.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
401570 c.355G>C p.E119Q Chr.15: 67236414-67236414 on Build GRCh38
4056429 c.338T>C p.V113A Chr.15: 67236431-67236431 on Build GRCh38
964272 c.342C>T p.C114C Chr.15: 67236427-67236427 on Build GRCh38
964273 c.265-1G>T p.? Chr.15: 67236505-67236505 on Build GRCh38
6464039 c.329T>A p.M110K Chr.15: 67236440-67236440 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368829803 Chr.15: 67236690-67236690 on Build GRCh38
rs767024623 Chr.15: 67236410-67236410 on Build GRCh38
rs768092505 Chr.15: 67236619-67236619 on Build GRCh38
rs186662479 Chr.15: 67236440-67236440 on Build GRCh38
rs779893221 Chr.15: 67236667-67236667 on Build GRCh38
rs16950788 Chr.15: 67236314-67236314 on Build GRCh38
rs895673292 Chr.15: 67236391-67236391 on Build GRCh38
rs775172947 Chr.15: 67236379-67236379 on Build GRCh38
rs375202846 Chr.15: 67236247-67236248 on Build GRCh38
rs182392210 Chr.15: 67236384-67236384 on Build GRCh38
rs763590567 Chr.15: 67236394-67236394 on Build GRCh38
rs375976894 Chr.15: 67236600-67236600 on Build GRCh38
rs772009019 Chr.15: 67236539-67236539 on Build GRCh38
rs760548483 Chr.15: 67236432-67236432 on Build GRCh38
rs767933811 Chr.15: 67236630-67236630 on Build GRCh38
rs946760528 Chr.15: 67236674-67236674 on Build GRCh38
rs905750369 Chr.15: 67236620-67236620 on Build GRCh38
rs74358343 Chr.15: 67236597-67236597 on Build GRCh38
rs893854724 Chr.15: 67236411-67236411 on Build GRCh38
rs201793128 Chr.15: 67236463-67236463 on Build GRCh38
rs922166344 Chr.15: 67236568-67236568 on Build GRCh38
rs746809836 Chr.15: 67236582-67236582 on Build GRCh38
rs756227926 Chr.15: 67236688-67236688 on Build GRCh38
rs746792468 Chr.15: 67236677-67236677 on Build GRCh38
rs745873731 Chr.15: 67236533-67236533 on Build GRCh38
rs946756886 Chr.15: 67236327-67236327 on Build GRCh38
rs767924700 Chr.15: 67236496-67236496 on Build GRCh38
rs908694678 Chr.15: 67236306-67236306 on Build GRCh38
rs747735433 Chr.15: 67236696-67236696 on Build GRCh38
rs565607839 Chr.15: 67236279-67236279 on Build GRCh38
rs923960005 Chr.15: 67236578-67236578 on Build GRCh38
rs537340808 Chr.15: 67236633-67236633 on Build GRCh38
rs767126385 Chr.15: 67236377-67236377 on Build GRCh38
rs766965793 Chr.15: 67236489-67236489 on Build GRCh38
rs770819514 Chr.15: 67236450-67236450 on Build GRCh38
rs756296586 Chr.15: 67236433-67236433 on Build GRCh38
rs769442825 Chr.15: 67236295-67236295 on Build GRCh38
rs745640728 Chr.15: 67236457-67236457 on Build GRCh38
rs1055058820 Chr.15: 67236405-67236405 on Build GRCh38
rs1044353187 Chr.15: 67236427-67236427 on Build GRCh38
rs191746737 Chr.15: 67236335-67236335 on Build GRCh38
rs917060604 Chr.15: 67236603-67236603 on Build GRCh38
rs748466481 Chr.15: 67236430-67236430 on Build GRCh38
rs771975734 Chr.15: 67236422-67236423 on Build GRCh38
rs776166893 Chr.15: 67236479-67236479 on Build GRCh38
rs751937153 Chr.15: 67236412-67236412 on Build GRCh38
rs775155034 Chr.15: 67236332-67236332 on Build GRCh38
rs763675606 Chr.15: 67236478-67236478 on Build GRCh38
rs764359056 Chr.15: 67236635-67236635 on Build GRCh38
rs771913310 Chr.15: 67236459-67236459 on Build GRCh38
rs761390863 Chr.15: 67236613-67236613 on Build GRCh38
rs983590733 Chr.15: 67236246-67236246 on Build GRCh38
rs758889595 Chr.15: 67236402-67236402 on Build GRCh38
rs764063999 Chr.15: 67236541-67236542 on Build GRCh38
rs772914150 Chr.15: 67236350-67236350 on Build GRCh38
rs966273435 Chr.15: 67236528-67236528 on Build GRCh38
rs375434660 Chr.15: 67236614-67236614 on Build GRCh38
rs778810033 Chr.15: 67236525-67236525 on Build GRCh38
rs758289239 Chr.15: 67236664-67236664 on Build GRCh38
rs912082600 Chr.15: 67236302-67236302 on Build GRCh38
rs776580912 Chr.15: 67236267-67236267 on Build GRCh38
rs974367168 Chr.15: 67236492-67236492 on Build GRCh38
rs749184555 Chr.15: 67236357-67236357 on Build GRCh38
rs371926366 Chr.15: 67236426-67236426 on Build GRCh38
rs201761651 Chr.15: 67236612-67236612 on Build GRCh38
rs561525955 Chr.15: 67236246-67236249 on Build GRCh38
rs968277972 Chr.15: 67236570-67236570 on Build GRCh38
rs753927877 Chr.15: 67236522-67236522 on Build GRCh38
rs752166113 Chr.15: 67236491-67236491 on Build GRCh38
rs768513159 Chr.15: 67236258-67236258 on Build GRCh38
rs759361906 Chr.15: 67236375-67236375 on Build GRCh38
rs760899710 Chr.15: 67236550-67236553 on Build GRCh38
rs761269874 Chr.15: 67236484-67236484 on Build GRCh38
rs755439411 Chr.15: 67236416-67236416 on Build GRCh38
rs759156533 Chr.15: 67236377-67236378 on Build GRCh38
rs865986004 Chr.15: 67236414-67236414 on Build GRCh38
rs983593668 Chr.15: 67236532-67236532 on Build GRCh38
rs930391607 Chr.15: 67236277-67236277 on Build GRCh38
rs777892178 Chr.15: 67236515-67236515 on Build GRCh38
rs750521244 Chr.15: 67236659-67236659 on Build GRCh38
rs770872438 Chr.15: 67236360-67236360 on Build GRCh38
rs757377231 Chr.15: 67236524-67236524 on Build GRCh38
rs756314278 Chr.15: 67236500-67236500 on Build GRCh38
rs757328023 Chr.15: 67236642-67236642 on Build GRCh38
rs754070263 Chr.15: 67236641-67236641 on Build GRCh38
rs755386550 Chr.15: 67236495-67236495 on Build GRCh38
rs200664620 Chr.15: 67236373-67236373 on Build GRCh38
rs907641824 Chr.15: 67236563-67236563 on Build GRCh38
rs551139491 Chr.15: 67236581-67236581 on Build GRCh38
rs556689819 Chr.15: 67236682-67236682 on Build GRCh38
rs915236527 Chr.15: 67236625-67236625 on Build GRCh38
rs778899986 Chr.15: 67236656-67236656 on Build GRCh38
rs200116466 Chr.15: 67236456-67236456 on Build GRCh38
rs949952804 Chr.15: 67236606-67236606 on Build GRCh38
rs753019024 Chr.15: 67236499-67236499 on Build GRCh38
rs760315068 Chr.15: 67236473-67236473 on Build GRCh38
rs979370190 Chr.15: 67236571-67236571 on Build GRCh38
rs774087773 Chr.15: 67236363-67236363 on Build GRCh38
rs1055345874 Chr.15: 67236616-67236616 on Build GRCh38
rs920308033 Chr.15: 67236247-67236247 on Build GRCh38
rs975053797 Chr.15: 67236312-67236312 on Build GRCh38
rs753300689 Chr.15: 67236400-67236400 on Build GRCh38
rs760092902 Chr.15: 67236390-67236390 on Build GRCh38
rs753066432 Chr.15: 67236632-67236632 on Build GRCh38
rs775489702 Chr.15: 67236425-67236425 on Build GRCh38

View Full Product Details