Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 68192501-68193008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALML4
Entrez Gene ID
91860
Gene Name
calmodulin like 4
Gene Aliases
NY-BR-20
UniGene
Hs.709550
Gene Chromosome Location
Chr.15: 68190705-68206110 on Build GRCh38
Transcripts
NM_001031733.2 NM_001286694.1 NM_001286695.1 NM_033429.2 NR_104583.1

SNP Detail

SNP ID Chromosome Location
rs999026318 Chr.15: 68192805-68192805 on Build GRCh38
rs75173362 Chr.15: 68192953-68192953 on Build GRCh38
rs984597847 Chr.15: 68192619-68192619 on Build GRCh38
rs541389572 Chr.15: 68192649-68192649 on Build GRCh38
rs1028034470 Chr.15: 68192911-68192911 on Build GRCh38
rs894138712 Chr.15: 68192687-68192687 on Build GRCh38
rs902269483 Chr.15: 68192735-68192735 on Build GRCh38
rs939566329 Chr.15: 68192675-68192675 on Build GRCh38
rs759021493 Chr.15: 68192642-68192642 on Build GRCh38
rs935037549 Chr.15: 68192763-68192763 on Build GRCh38
rs188369358 Chr.15: 68192812-68192812 on Build GRCh38
rs954104416 Chr.15: 68192587-68192587 on Build GRCh38
rs555570390 Chr.15: 68192564-68192564 on Build GRCh38
rs1056352999 Chr.15: 68192815-68192818 on Build GRCh38
rs397854265 Chr.15: 68192687-68192687 on Build GRCh38
rs1019105401 Chr.15: 68192952-68192952 on Build GRCh38
rs1037132753 Chr.15: 68192661-68192661 on Build GRCh38
rs1012615301 Chr.15: 68192697-68192697 on Build GRCh38
rs994859864 Chr.15: 68192859-68192859 on Build GRCh38
rs950831016 Chr.15: 68192586-68192586 on Build GRCh38
rs1023535835 Chr.15: 68192823-68192823 on Build GRCh38
rs965262523 Chr.15: 68192538-68192538 on Build GRCh38
rs560467480 Chr.15: 68192671-68192671 on Build GRCh38
rs998092760 Chr.15: 68192937-68192937 on Build GRCh38
rs760384415 Chr.15: 68192719-68192719 on Build GRCh38
rs752078380 Chr.15: 68192669-68192669 on Build GRCh38
rs984114457 Chr.15: 68192975-68192975 on Build GRCh38
rs767043865 Chr.15: 68192652-68192652 on Build GRCh38
rs562709945 Chr.15: 68192730-68192730 on Build GRCh38
rs185568697 Chr.15: 68192737-68192737 on Build GRCh38
rs546040139 Chr.15: 68192717-68192717 on Build GRCh38
rs534852599 Chr.15: 68192819-68192819 on Build GRCh38
rs921260732 Chr.15: 68192583-68192583 on Build GRCh38
rs763787823 Chr.15: 68192766-68192766 on Build GRCh38
rs374439450 Chr.15: 68192567-68192567 on Build GRCh38
rs572449154 Chr.15: 68192615-68192615 on Build GRCh38
rs532411957 Chr.15: 68192686-68192686 on Build GRCh38
rs922222008 Chr.15: 68192973-68192973 on Build GRCh38
rs572522401 Chr.15: 68192774-68192774 on Build GRCh38
rs896314781 Chr.15: 68192818-68192818 on Build GRCh38
rs180792320 Chr.15: 68192886-68192886 on Build GRCh38
rs904251831 Chr.15: 68192802-68192802 on Build GRCh38
rs909990291 Chr.15: 68192630-68192630 on Build GRCh38
rs542343258 Chr.15: 68192533-68192533 on Build GRCh38
rs898381010 Chr.15: 68192831-68192831 on Build GRCh38
rs539773244 Chr.15: 68192965-68192965 on Build GRCh38
rs752946215 Chr.15: 68192581-68192581 on Build GRCh38
rs770584767 Chr.15: 68192670-68192671 on Build GRCh38
rs928143449 Chr.15: 68192674-68192674 on Build GRCh38
rs561584207 Chr.15: 68192828-68192828 on Build GRCh38
rs983591408 Chr.15: 68192629-68192629 on Build GRCh38
rs76047328 Chr.15: 68192961-68192961 on Build GRCh38
rs143848376 Chr.15: 68192905-68192905 on Build GRCh38
rs757664159 Chr.15: 68192963-68192963 on Build GRCh38
rs976666318 Chr.15: 68192562-68192562 on Build GRCh38
rs974958105 Chr.15: 68192654-68192654 on Build GRCh38

View Full Product Details