Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 68205539-68206045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALML4
Entrez Gene ID
91860
Gene Name
calmodulin like 4
Gene Aliases
NY-BR-20
UniGene
Hs.709550
Gene Chromosome Location
Chr.15: 68190705-68206110 on Build GRCh38
Transcripts
NM_001031733.2 NM_001286694.1 NM_001286695.1 NM_033429.2 NR_104583.1

SNP Detail

SNP ID Chromosome Location
rs759781073 Chr.15: 68205671-68205671 on Build GRCh38
rs1044049651 Chr.15: 68205760-68205760 on Build GRCh38
rs192540126 Chr.15: 68206018-68206018 on Build GRCh38
rs529439954 Chr.15: 68205698-68205698 on Build GRCh38
rs1029497116 Chr.15: 68205670-68205670 on Build GRCh38
rs533374270 Chr.15: 68205602-68205602 on Build GRCh38
rs955353317 Chr.15: 68206010-68206010 on Build GRCh38
rs970953047 Chr.15: 68205684-68205684 on Build GRCh38
rs1003646249 Chr.15: 68205687-68205687 on Build GRCh38
rs563849483 Chr.15: 68205655-68205655 on Build GRCh38
rs896705889 Chr.15: 68205739-68205739 on Build GRCh38
rs535221932 Chr.15: 68205913-68205913 on Build GRCh38
rs1054059252 Chr.15: 68206005-68206005 on Build GRCh38
rs138212402 Chr.15: 68205604-68205604 on Build GRCh38
rs540990411 Chr.15: 68205979-68205979 on Build GRCh38
rs948316961 Chr.15: 68205959-68205959 on Build GRCh38
rs547384528 Chr.15: 68205773-68205773 on Build GRCh38
rs1023061847 Chr.15: 68205601-68205601 on Build GRCh38
rs930104465 Chr.15: 68205732-68205732 on Build GRCh38
rs1003813899 Chr.15: 68205861-68205861 on Build GRCh38
rs142896088 Chr.15: 68205809-68205809 on Build GRCh38
rs780686812 Chr.15: 68206019-68206019 on Build GRCh38
rs577679574 Chr.15: 68205729-68205729 on Build GRCh38
rs560935988 Chr.15: 68205784-68205784 on Build GRCh38
rs922415327 Chr.15: 68205982-68205982 on Build GRCh38
rs918634445 Chr.15: 68205870-68205870 on Build GRCh38
rs974117533 Chr.15: 68205835-68205835 on Build GRCh38
rs554998713 Chr.15: 68205914-68205914 on Build GRCh38
rs985801812 Chr.15: 68205813-68205813 on Build GRCh38
rs746611943 Chr.15: 68206013-68206013 on Build GRCh38
rs776442596 Chr.15: 68205650-68205650 on Build GRCh38
rs11071991 Chr.15: 68205714-68205714 on Build GRCh38
rs999878327 Chr.15: 68205778-68205778 on Build GRCh38
rs988624146 Chr.15: 68205658-68205658 on Build GRCh38
rs1032833287 Chr.15: 68205632-68205632 on Build GRCh38
rs1018260372 Chr.15: 68205831-68205831 on Build GRCh38
rs572005917 Chr.15: 68205580-68205580 on Build GRCh38
rs963003516 Chr.15: 68205834-68205834 on Build GRCh38
rs959072802 Chr.15: 68205894-68205894 on Build GRCh38
rs922552580 Chr.15: 68205740-68205740 on Build GRCh38
rs7183205 Chr.15: 68205930-68205930 on Build GRCh38
rs893993092 Chr.15: 68205620-68205620 on Build GRCh38
rs564763405 Chr.15: 68205586-68205586 on Build GRCh38
rs145059814 Chr.15: 68205885-68205885 on Build GRCh38
rs941433316 Chr.15: 68205709-68205709 on Build GRCh38
rs948337905 Chr.15: 68205754-68205754 on Build GRCh38
rs780027358 Chr.15: 68205977-68205977 on Build GRCh38
rs1007628706 Chr.15: 68205581-68205584 on Build GRCh38
rs756287665 Chr.15: 68205808-68205808 on Build GRCh38
rs906824966 Chr.15: 68205843-68205843 on Build GRCh38
rs187969629 Chr.15: 68205755-68205755 on Build GRCh38
rs943339052 Chr.15: 68205777-68205777 on Build GRCh38
rs566124218 Chr.15: 68205736-68205736 on Build GRCh38
rs541366717 Chr.15: 68205585-68205585 on Build GRCh38
rs139477369 Chr.15: 68205719-68205720 on Build GRCh38
rs778706093 Chr.15: 68205859-68205859 on Build GRCh38
rs1021199285 Chr.15: 68205649-68205649 on Build GRCh38
rs951450042 Chr.15: 68205884-68205884 on Build GRCh38
rs767531443 Chr.15: 68205710-68205710 on Build GRCh38
rs1056648502 Chr.15: 68205734-68205734 on Build GRCh38
rs985472366 Chr.15: 68205682-68205682 on Build GRCh38
rs561389962 Chr.15: 68205577-68205577 on Build GRCh38
rs890980701 Chr.15: 68206022-68206022 on Build GRCh38
rs935627679 Chr.15: 68206000-68206000 on Build GRCh38
rs539331804 Chr.15: 68205793-68205793 on Build GRCh38
rs1032700232 Chr.15: 68205780-68205780 on Build GRCh38
rs1016727262 Chr.15: 68205929-68205929 on Build GRCh38
rs909593970 Chr.15: 68205733-68205733 on Build GRCh38

View Full Product Details