Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 68205934-68206432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALML4
Entrez Gene ID
91860
Gene Name
calmodulin like 4
Gene Aliases
NY-BR-20
UniGene
Hs.709550
Gene Chromosome Location
Chr.15: 68190705-68206110 on Build GRCh38
Transcripts
NM_001031733.2 NM_001286694.1 NM_001286695.1 NM_033429.2 NR_104583.1

SNP Detail

SNP ID Chromosome Location
rs887078159 Chr.15: 68206190-68206190 on Build GRCh38
rs192540126 Chr.15: 68206018-68206018 on Build GRCh38
rs914166258 Chr.15: 68206308-68206308 on Build GRCh38
rs562652253 Chr.15: 68206412-68206412 on Build GRCh38
rs955353317 Chr.15: 68206010-68206010 on Build GRCh38
rs185419326 Chr.15: 68206112-68206112 on Build GRCh38
rs773568261 Chr.15: 68206047-68206047 on Build GRCh38
rs747606771 Chr.15: 68206031-68206031 on Build GRCh38
rs993793197 Chr.15: 68206310-68206310 on Build GRCh38
rs981341285 Chr.15: 68206257-68206257 on Build GRCh38
rs774716898 Chr.15: 68206119-68206119 on Build GRCh38
rs1039545193 Chr.15: 68206163-68206163 on Build GRCh38
rs1054059252 Chr.15: 68206005-68206005 on Build GRCh38
rs957200696 Chr.15: 68206361-68206361 on Build GRCh38
rs992273271 Chr.15: 68206086-68206086 on Build GRCh38
rs759518807 Chr.15: 68206125-68206125 on Build GRCh38
rs771242562 Chr.15: 68206083-68206083 on Build GRCh38
rs543531886 Chr.15: 68206204-68206204 on Build GRCh38
rs1013611207 Chr.15: 68206214-68206214 on Build GRCh38
rs540990411 Chr.15: 68205979-68205979 on Build GRCh38
rs948316961 Chr.15: 68205959-68205959 on Build GRCh38
rs112825138 Chr.15: 68206143-68206143 on Build GRCh38
rs148312821 Chr.15: 68206073-68206073 on Build GRCh38
rs549502532 Chr.15: 68206217-68206217 on Build GRCh38
rs776515609 Chr.15: 68206223-68206223 on Build GRCh38
rs759692928 Chr.15: 68206130-68206130 on Build GRCh38
rs988378564 Chr.15: 68206299-68206299 on Build GRCh38
rs780686812 Chr.15: 68206019-68206019 on Build GRCh38
rs890895086 Chr.15: 68206149-68206149 on Build GRCh38
rs549663331 Chr.15: 68206413-68206413 on Build GRCh38
rs1025684150 Chr.15: 68206071-68206071 on Build GRCh38
rs1025441232 Chr.15: 68206249-68206249 on Build GRCh38
rs922415327 Chr.15: 68205982-68205982 on Build GRCh38
rs576139842 Chr.15: 68206113-68206113 on Build GRCh38
rs746611943 Chr.15: 68206013-68206013 on Build GRCh38
rs578215308 Chr.15: 68206034-68206034 on Build GRCh38
rs939569992 Chr.15: 68206211-68206211 on Build GRCh38
rs945690244 Chr.15: 68206339-68206339 on Build GRCh38
rs551780900 Chr.15: 68206281-68206281 on Build GRCh38
rs986431385 Chr.15: 68206341-68206341 on Build GRCh38
rs559757083 Chr.15: 68206252-68206252 on Build GRCh38
rs1006750673 Chr.15: 68206349-68206349 on Build GRCh38
rs980967028 Chr.15: 68206120-68206120 on Build GRCh38
rs945784040 Chr.15: 68206134-68206134 on Build GRCh38
rs1025631823 Chr.15: 68206184-68206184 on Build GRCh38
rs979938824 Chr.15: 68206133-68206133 on Build GRCh38
rs912264120 Chr.15: 68206344-68206344 on Build GRCh38
rs150557996 Chr.15: 68206345-68206345 on Build GRCh38
rs764277214 Chr.15: 68206181-68206181 on Build GRCh38
rs753025373 Chr.15: 68206156-68206156 on Build GRCh38
rs957500772 Chr.15: 68206172-68206172 on Build GRCh38
rs1029423980 Chr.15: 68206336-68206336 on Build GRCh38
rs539763391 Chr.15: 68206353-68206353 on Build GRCh38
rs35229188 Chr.15: 68206360-68206361 on Build GRCh38
rs925494938 Chr.15: 68206123-68206123 on Build GRCh38
rs35382858 Chr.15: 68206402-68206403 on Build GRCh38
rs978150641 Chr.15: 68206175-68206175 on Build GRCh38
rs780027358 Chr.15: 68205977-68205977 on Build GRCh38
rs542555297 Chr.15: 68206355-68206355 on Build GRCh38
rs990045688 Chr.15: 68206379-68206379 on Build GRCh38
rs995472022 Chr.15: 68206048-68206048 on Build GRCh38
rs958267633 Chr.15: 68206287-68206287 on Build GRCh38
rs573595655 Chr.15: 68206174-68206174 on Build GRCh38
rs760652944 Chr.15: 68206168-68206168 on Build GRCh38
rs141742988 Chr.15: 68206280-68206280 on Build GRCh38
rs769402978 Chr.15: 68206046-68206046 on Build GRCh38
rs890980701 Chr.15: 68206022-68206022 on Build GRCh38
rs779937275 Chr.15: 68206383-68206383 on Build GRCh38
rs935627679 Chr.15: 68206000-68206000 on Build GRCh38

View Full Product Details