Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 68278002-68278506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FEM1B
Entrez Gene ID
10116
Gene Name
fem-1 homolog B
Gene Aliases
F1A-ALPHA, F1AA, FEM1-beta
UniGene
Hs.362733
Gene Chromosome Location
Chr.15: 68277803-68295865 on Build GRCh38
Transcripts
NM_015322.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6288106 c.44G>A p.G15D Chr.15: 68278461-68278461 on Build GRCh38
5772718 c.40G>A p.E14K Chr.15: 68278457-68278457 on Build GRCh38
4804694 c.43G>A p.G15S Chr.15: 68278460-68278460 on Build GRCh38
5629531 c.24A>G p.V8V Chr.15: 68278441-68278441 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs998542365 Chr.15: 68278161-68278161 on Build GRCh38
rs748744787 Chr.15: 68278205-68278205 on Build GRCh38
rs35987978 Chr.15: 68278449-68278450 on Build GRCh38
rs763808050 Chr.15: 68278381-68278381 on Build GRCh38
rs988684907 Chr.15: 68278160-68278160 on Build GRCh38
rs548065777 Chr.15: 68278212-68278212 on Build GRCh38
rs1036734356 Chr.15: 68278119-68278119 on Build GRCh38
rs955934912 Chr.15: 68278065-68278065 on Build GRCh38
rs200735664 Chr.15: 68278395-68278395 on Build GRCh38
rs988129188 Chr.15: 68278069-68278069 on Build GRCh38
rs1048531225 Chr.15: 68278091-68278091 on Build GRCh38
rs746085028 Chr.15: 68278475-68278475 on Build GRCh38
rs778031981 Chr.15: 68278436-68278436 on Build GRCh38
rs1031480534 Chr.15: 68278164-68278164 on Build GRCh38
rs145530056 Chr.15: 68278196-68278196 on Build GRCh38
rs552456623 Chr.15: 68278172-68278172 on Build GRCh38
rs771040816 Chr.15: 68278068-68278068 on Build GRCh38
rs978587629 Chr.15: 68278039-68278039 on Build GRCh38
rs375373890 Chr.15: 68278413-68278413 on Build GRCh38
rs373528730 Chr.15: 68278280-68278280 on Build GRCh38
rs965151835 Chr.15: 68278168-68278168 on Build GRCh38
rs926693684 Chr.15: 68278087-68278087 on Build GRCh38
rs940120354 Chr.15: 68278238-68278238 on Build GRCh38
rs185418232 Chr.15: 68278251-68278251 on Build GRCh38
rs906675266 Chr.15: 68278095-68278095 on Build GRCh38
rs955988532 Chr.15: 68278156-68278156 on Build GRCh38
rs576355982 Chr.15: 68278450-68278450 on Build GRCh38
rs774533588 Chr.15: 68278337-68278337 on Build GRCh38
rs946974756 Chr.15: 68278287-68278287 on Build GRCh38
rs770485475 Chr.15: 68278273-68278273 on Build GRCh38
rs761400696 Chr.15: 68278394-68278394 on Build GRCh38
rs556864855 Chr.15: 68278400-68278400 on Build GRCh38
rs774656717 Chr.15: 68278286-68278286 on Build GRCh38
rs372561452 Chr.15: 68278415-68278415 on Build GRCh38
rs756580056 Chr.15: 68278435-68278435 on Build GRCh38
rs374792610 Chr.15: 68278169-68278169 on Build GRCh38
rs368483635 Chr.15: 68278331-68278331 on Build GRCh38
rs147100682 Chr.15: 68278411-68278411 on Build GRCh38
rs1009989798 Chr.15: 68278363-68278363 on Build GRCh38
rs997823117 Chr.15: 68278131-68278131 on Build GRCh38
rs959755098 Chr.15: 68278423-68278423 on Build GRCh38
rs35278866 Chr.15: 68278302-68278303 on Build GRCh38
rs565648577 Chr.15: 68278284-68278285 on Build GRCh38
rs188776003 Chr.15: 68278271-68278271 on Build GRCh38
rs1043137452 Chr.15: 68278303-68278303 on Build GRCh38
rs1021280946 Chr.15: 68278139-68278139 on Build GRCh38
rs757540393 Chr.15: 68278441-68278441 on Build GRCh38
rs1055350822 Chr.15: 68278105-68278105 on Build GRCh38
rs940106762 Chr.15: 68278064-68278064 on Build GRCh38
rs894144044 Chr.15: 68278112-68278112 on Build GRCh38
rs928124729 Chr.15: 68278253-68278253 on Build GRCh38
rs1013034271 Chr.15: 68278123-68278123 on Build GRCh38
rs754184266 Chr.15: 68278438-68278438 on Build GRCh38
rs1037228397 Chr.15: 68278089-68278089 on Build GRCh38
rs954123095 Chr.15: 68278421-68278421 on Build GRCh38
rs760646274 Chr.15: 68278367-68278367 on Build GRCh38
rs984557230 Chr.15: 68278224-68278224 on Build GRCh38
rs1032829968 Chr.15: 68278155-68278155 on Build GRCh38
rs776417687 Chr.15: 68278391-68278391 on Build GRCh38
rs759561935 Chr.15: 68278380-68278380 on Build GRCh38
rs904298609 Chr.15: 68278142-68278142 on Build GRCh38
rs938191517 Chr.15: 68278090-68278090 on Build GRCh38
rs904163844 Chr.15: 68278307-68278307 on Build GRCh38
rs909866719 Chr.15: 68278237-68278237 on Build GRCh38
rs939596421 Chr.15: 68278099-68278099 on Build GRCh38
rs1031346128 Chr.15: 68278366-68278366 on Build GRCh38
rs1033357024 Chr.15: 68278063-68278063 on Build GRCh38
rs949507859 Chr.15: 68278074-68278074 on Build GRCh38
rs889517330 Chr.15: 68278137-68278137 on Build GRCh38
rs1049382146 Chr.15: 68278280-68278280 on Build GRCh38
rs926718733 Chr.15: 68278197-68278197 on Build GRCh38
rs982605639 Chr.15: 68278086-68278086 on Build GRCh38
rs35606342 Chr.15: 68278456-68278456 on Build GRCh38
rs909992307 Chr.15: 68278059-68278059 on Build GRCh38
rs536461256 Chr.15: 68278021-68278021 on Build GRCh38
rs913895080 Chr.15: 68278073-68278073 on Build GRCh38
rs984135964 Chr.15: 68278027-68278027 on Build GRCh38
rs778625815 Chr.15: 68278408-68278423 on Build GRCh38
rs533141310 Chr.15: 68278132-68278132 on Build GRCh38
rs998571765 Chr.15: 68278312-68278312 on Build GRCh38
rs533893343 Chr.15: 68278270-68278270 on Build GRCh38
rs74521321 Chr.15: 68278200-68278200 on Build GRCh38
rs570469364 Chr.15: 68278319-68278319 on Build GRCh38

View Full Product Details