Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 69270409-69270951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLCE
Entrez Gene ID
26035
Gene Name
glucuronic acid epimerase
Gene Aliases
HSEPI
UniGene
Hs.183006
Gene Chromosome Location
Chr.15: 69160555-69272209 on Build GRCh38
Transcripts
NM_001324092.1 NM_001324093.1 NM_001324094.1 NM_015554.2 XM_005254298.3 XM_017022073.1 XM_017022074.1

SNP Detail

SNP ID Chromosome Location
rs530830862 Chr.15: 69270755-69270755 on Build GRCh38
rs936094004 Chr.15: 69270607-69270607 on Build GRCh38
rs1025330300 Chr.15: 69270602-69270603 on Build GRCh38
rs1056384092 Chr.15: 69270714-69270714 on Build GRCh38
rs191629069 Chr.15: 69270920-69270920 on Build GRCh38
rs778801496 Chr.15: 69270682-69270682 on Build GRCh38
rs1002472456 Chr.15: 69270487-69270487 on Build GRCh38
rs1009779036 Chr.15: 69270622-69270622 on Build GRCh38
rs923946907 Chr.15: 69270685-69270685 on Build GRCh38
rs1042339780 Chr.15: 69270632-69270632 on Build GRCh38
rs995277865 Chr.15: 69270821-69270821 on Build GRCh38
rs57184738 Chr.15: 69270608-69270608 on Build GRCh38
rs890353710 Chr.15: 69270635-69270635 on Build GRCh38
rs3743094 Chr.15: 69270540-69270540 on Build GRCh38
rs1056272276 Chr.15: 69270544-69270544 on Build GRCh38
rs890406007 Chr.15: 69270901-69270901 on Build GRCh38
rs149316926 Chr.15: 69270810-69270810 on Build GRCh38
rs945208215 Chr.15: 69270836-69270836 on Build GRCh38
rs560349479 Chr.15: 69270500-69270500 on Build GRCh38
rs971609583 Chr.15: 69270772-69270772 on Build GRCh38
rs886856555 Chr.15: 69270873-69270873 on Build GRCh38
rs879046817 Chr.15: 69270700-69270700 on Build GRCh38
rs954367613 Chr.15: 69270503-69270503 on Build GRCh38
rs1038758094 Chr.15: 69270924-69270924 on Build GRCh38
rs950073679 Chr.15: 69270585-69270585 on Build GRCh38
rs935257428 Chr.15: 69270508-69270508 on Build GRCh38
rs1053615722 Chr.15: 69270491-69270491 on Build GRCh38
rs917525262 Chr.15: 69270770-69270770 on Build GRCh38
rs574562214 Chr.15: 69270623-69270623 on Build GRCh38
rs575199763 Chr.15: 69270581-69270581 on Build GRCh38
rs745545981 Chr.15: 69270820-69270820 on Build GRCh38
rs554727047 Chr.15: 69270621-69270621 on Build GRCh38
rs752222522 Chr.15: 69270834-69270835 on Build GRCh38
rs905935364 Chr.15: 69270480-69270480 on Build GRCh38
rs143140873 Chr.15: 69270504-69270504 on Build GRCh38
rs369816381 Chr.15: 69270788-69270788 on Build GRCh38
rs1044474015 Chr.15: 69270473-69270473 on Build GRCh38
rs560282043 Chr.15: 69270647-69270647 on Build GRCh38
rs974008539 Chr.15: 69270543-69270543 on Build GRCh38
rs1059923 Chr.15: 69270916-69270916 on Build GRCh38
rs749169397 Chr.15: 69270591-69270591 on Build GRCh38
rs927316553 Chr.15: 69270796-69270796 on Build GRCh38
rs562599529 Chr.15: 69270754-69270754 on Build GRCh38
rs946839200 Chr.15: 69270660-69270660 on Build GRCh38
rs1009482643 Chr.15: 69270499-69270499 on Build GRCh38
rs896433426 Chr.15: 69270743-69270744 on Build GRCh38
rs372028285 Chr.15: 69270803-69270803 on Build GRCh38
rs577106635 Chr.15: 69270674-69270674 on Build GRCh38
rs950991982 Chr.15: 69270605-69270605 on Build GRCh38
rs893750675 Chr.15: 69270494-69270494 on Build GRCh38
rs755074078 Chr.15: 69270571-69270571 on Build GRCh38
rs947948810 Chr.15: 69270760-69270760 on Build GRCh38
rs530121200 Chr.15: 69270850-69270850 on Build GRCh38
rs1020841592 Chr.15: 69270502-69270502 on Build GRCh38
rs1008590089 Chr.15: 69270646-69270646 on Build GRCh38
rs944851742 Chr.15: 69270483-69270483 on Build GRCh38
rs1023878283 Chr.15: 69270590-69270590 on Build GRCh38
rs1017383404 Chr.15: 69270516-69270516 on Build GRCh38
rs766593834 Chr.15: 69270452-69270452 on Build GRCh38
rs57502520 Chr.15: 69270835-69270835 on Build GRCh38
rs60602027 Chr.15: 69270639-69270639 on Build GRCh38
rs1042226410 Chr.15: 69270837-69270837 on Build GRCh38
rs867152343 Chr.15: 69270617-69270617 on Build GRCh38
rs900174246 Chr.15: 69270925-69270925 on Build GRCh38
rs920764717 Chr.15: 69270549-69270549 on Build GRCh38
rs950909786 Chr.15: 69270563-69270563 on Build GRCh38
rs138311237 Chr.15: 69270708-69270708 on Build GRCh38
rs16953674 Chr.15: 69270835-69270835 on Build GRCh38
rs964851593 Chr.15: 69270665-69270665 on Build GRCh38
rs773536146 Chr.15: 69270571-69270571 on Build GRCh38
rs927412500 Chr.15: 69270593-69270593 on Build GRCh38

View Full Product Details