Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 71111768-71112280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CT62
Entrez Gene ID
196993
Gene Name
cancer/testis antigen 62
Gene Aliases
-
UniGene
Hs.646533
Gene Chromosome Location
Chr.15: 71110244-71115551 on Build GRCh38
Transcripts
NM_001102658.1 XM_006720429.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4993873 c.5T>C p.M2T Chr.15: 71112278-71112278 on Build GRCh38
4913155 c.88A>T p.S30C Chr.15: 71112195-71112195 on Build GRCh38
701344 c.76C>A p.H26N Chr.15: 71112207-71112207 on Build GRCh38
1608642 c.61G>A p.A21T Chr.15: 71112222-71112222 on Build GRCh38
4151120 c.54G>A p.Q18Q Chr.15: 71112229-71112229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373577358 Chr.15: 71112140-71112140 on Build GRCh38
rs539549109 Chr.15: 71111897-71111897 on Build GRCh38
rs939619483 Chr.15: 71112120-71112120 on Build GRCh38
rs750342581 Chr.15: 71112134-71112134 on Build GRCh38
rs553474345 Chr.15: 71112005-71112005 on Build GRCh38
rs147445710 Chr.15: 71111857-71111857 on Build GRCh38
rs182883425 Chr.15: 71112159-71112159 on Build GRCh38
rs761920809 Chr.15: 71112216-71112216 on Build GRCh38
rs1038135026 Chr.15: 71112198-71112198 on Build GRCh38
rs558039779 Chr.15: 71112052-71112052 on Build GRCh38
rs755622396 Chr.15: 71112153-71112153 on Build GRCh38
rs746371117 Chr.15: 71112168-71112168 on Build GRCh38
rs570189518 Chr.15: 71111923-71111923 on Build GRCh38
rs886851563 Chr.15: 71111892-71111892 on Build GRCh38
rs758033553 Chr.15: 71112011-71112011 on Build GRCh38
rs186954308 Chr.15: 71111805-71111805 on Build GRCh38
rs536977721 Chr.15: 71112083-71112083 on Build GRCh38
rs780399018 Chr.15: 71112196-71112196 on Build GRCh38
rs946926146 Chr.15: 71111818-71111818 on Build GRCh38
rs923012682 Chr.15: 71111845-71111845 on Build GRCh38
rs898333957 Chr.15: 71111960-71111960 on Build GRCh38
rs190269080 Chr.15: 71112069-71112069 on Build GRCh38
rs772382768 Chr.15: 71112175-71112175 on Build GRCh38
rs139883238 Chr.15: 71111977-71111977 on Build GRCh38
rs747607578 Chr.15: 71112045-71112045 on Build GRCh38
rs566072790 Chr.15: 71112008-71112008 on Build GRCh38
rs200246125 Chr.15: 71112147-71112147 on Build GRCh38
rs369812828 Chr.15: 71112114-71112114 on Build GRCh38
rs1040088449 Chr.15: 71111833-71111833 on Build GRCh38
rs1004003700 Chr.15: 71111931-71111931 on Build GRCh38
rs751334099 Chr.15: 71112141-71112141 on Build GRCh38
rs1050502962 Chr.15: 71111879-71111879 on Build GRCh38
rs573799203 Chr.15: 71112149-71112149 on Build GRCh38
rs552659111 Chr.15: 71112222-71112222 on Build GRCh38
rs772462053 Chr.15: 71112081-71112081 on Build GRCh38
rs759108144 Chr.15: 71112003-71112025 on Build GRCh38
rs747696629 Chr.15: 71112151-71112151 on Build GRCh38
rs774882862 Chr.15: 71112055-71112055 on Build GRCh38
rs768889271 Chr.15: 71112211-71112211 on Build GRCh38
rs1038568906 Chr.15: 71111948-71111948 on Build GRCh38
rs765115529 Chr.15: 71112115-71112115 on Build GRCh38
rs1045334023 Chr.15: 71111819-71111819 on Build GRCh38
rs776793608 Chr.15: 71112212-71112212 on Build GRCh38
rs935088254 Chr.15: 71111832-71111832 on Build GRCh38
rs549547628 Chr.15: 71112007-71112007 on Build GRCh38
rs59864115 Chr.15: 71112158-71112158 on Build GRCh38
rs756807429 Chr.15: 71111820-71111820 on Build GRCh38
rs778362317 Chr.15: 71111991-71111991 on Build GRCh38
rs771587682 Chr.15: 71112054-71112054 on Build GRCh38
rs764233341 Chr.15: 71112098-71112098 on Build GRCh38
rs144480009 Chr.15: 71112112-71112112 on Build GRCh38
rs752265923 Chr.15: 71112225-71112226 on Build GRCh38
rs1023620660 Chr.15: 71112097-71112097 on Build GRCh38
rs762953777 Chr.15: 71112245-71112245 on Build GRCh38
rs760880783 Chr.15: 71112084-71112084 on Build GRCh38
rs535029069 Chr.15: 71112032-71112032 on Build GRCh38
rs141415059 Chr.15: 71111855-71111855 on Build GRCh38
rs747311738 Chr.15: 71112209-71112209 on Build GRCh38
rs767065597 Chr.15: 71112076-71112076 on Build GRCh38
rs979313369 Chr.15: 71112118-71112118 on Build GRCh38
rs368872597 Chr.15: 71112224-71112224 on Build GRCh38

View Full Product Details