Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 88836122-88836636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAN
Entrez Gene ID
176
Gene Name
aggrecan
Gene Aliases
AGC1, AGCAN, CSPG1, CSPGCP, MSK16, SEDK
UniGene
Hs.2159
Gene Chromosome Location
Chr.15: 88803442-88875354 on Build GRCh38
Transcripts
NM_001135.3 NM_013227.3 XM_006720419.1 XM_011521313.1 XM_011521314.1 XM_017021985.1 XM_017021986.1 XM_017021987.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4151182 c.40A>T p.I14F Chr.15: 88836246-88836246 on Build GRCh38
5677699 c.36G>T p.R12S Chr.15: 88836242-88836242 on Build GRCh38
7290998 c.22T>G p.F8V Chr.15: 88836228-88836228 on Build GRCh38
5677698 c.36G>T p.R12S Chr.15: 88836242-88836242 on Build GRCh38
6591777 c.56C>T p.T19I Chr.15: 88836262-88836262 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1033583712 Chr.15: 88836364-88836364 on Build GRCh38
rs941971428 Chr.15: 88836400-88836400 on Build GRCh38
rs956101881 Chr.15: 88836176-88836176 on Build GRCh38
rs1002139229 Chr.15: 88836361-88836361 on Build GRCh38
rs759649540 Chr.15: 88836202-88836202 on Build GRCh38
rs572035952 Chr.15: 88836178-88836178 on Build GRCh38
rs945122179 Chr.15: 88836531-88836531 on Build GRCh38
rs557798737 Chr.15: 88836281-88836281 on Build GRCh38
rs768643230 Chr.15: 88836325-88836325 on Build GRCh38
rs373354169 Chr.15: 88836167-88836167 on Build GRCh38
rs917698599 Chr.15: 88836349-88836349 on Build GRCh38
rs747727343 Chr.15: 88836190-88836190 on Build GRCh38
rs553801256 Chr.15: 88836171-88836171 on Build GRCh38
rs760468016 Chr.15: 88836233-88836233 on Build GRCh38
rs778467717 Chr.15: 88836602-88836602 on Build GRCh38
rs185836629 Chr.15: 88836285-88836285 on Build GRCh38
rs528697242 Chr.15: 88836562-88836562 on Build GRCh38
rs778533738 Chr.15: 88836179-88836179 on Build GRCh38
rs114629087 Chr.15: 88836485-88836485 on Build GRCh38
rs773598650 Chr.15: 88836191-88836191 on Build GRCh38
rs138849113 Chr.15: 88836289-88836289 on Build GRCh38
rs202166561 Chr.15: 88836217-88836217 on Build GRCh38
rs1035045006 Chr.15: 88836506-88836506 on Build GRCh38
rs766063753 Chr.15: 88836255-88836255 on Build GRCh38
rs754672128 Chr.15: 88836267-88836267 on Build GRCh38
rs991770830 Chr.15: 88836323-88836323 on Build GRCh38
rs753706123 Chr.15: 88836260-88836260 on Build GRCh38
rs779714893 Chr.15: 88836175-88836175 on Build GRCh38
rs1055086859 Chr.15: 88836582-88836582 on Build GRCh38
rs1051765491 Chr.15: 88836457-88836457 on Build GRCh38
rs540971342 Chr.15: 88836374-88836374 on Build GRCh38
rs776631256 Chr.15: 88836231-88836231 on Build GRCh38
rs906157675 Chr.15: 88836350-88836350 on Build GRCh38
rs751874711 Chr.15: 88836521-88836521 on Build GRCh38
rs913365596 Chr.15: 88836528-88836528 on Build GRCh38
rs559234468 Chr.15: 88836196-88836196 on Build GRCh38
rs552314184 Chr.15: 88836522-88836522 on Build GRCh38
rs777104551 Chr.15: 88836199-88836199 on Build GRCh38
rs766617563 Chr.15: 88836459-88836459 on Build GRCh38
rs778292021 Chr.15: 88836288-88836288 on Build GRCh38
rs1051773987 Chr.15: 88836609-88836609 on Build GRCh38
rs545563554 Chr.15: 88836197-88836197 on Build GRCh38
rs769950308 Chr.15: 88836311-88836311 on Build GRCh38
rs371249232 Chr.15: 88836219-88836219 on Build GRCh38
rs750516644 Chr.15: 88836413-88836413 on Build GRCh38
rs529108340 Chr.15: 88836593-88836593 on Build GRCh38
rs891889049 Chr.15: 88836458-88836458 on Build GRCh38
rs1030450511 Chr.15: 88836153-88836153 on Build GRCh38
rs995076300 Chr.15: 88836383-88836383 on Build GRCh38
rs900398377 Chr.15: 88836450-88836450 on Build GRCh38
rs781613114 Chr.15: 88836559-88836559 on Build GRCh38
rs373848594 Chr.15: 88836292-88836292 on Build GRCh38
rs933342636 Chr.15: 88836456-88836456 on Build GRCh38
rs144695270 Chr.15: 88836564-88836564 on Build GRCh38
rs1001694490 Chr.15: 88836502-88836502 on Build GRCh38
rs576077940 Chr.15: 88836282-88836282 on Build GRCh38
rs770071622 Chr.15: 88836211-88836211 on Build GRCh38
rs745412789 Chr.15: 88836610-88836610 on Build GRCh38
rs755350654 Chr.15: 88836541-88836541 on Build GRCh38
rs936219579 Chr.15: 88836519-88836519 on Build GRCh38
rs1024555533 Chr.15: 88836530-88836530 on Build GRCh38
rs761719517 Chr.15: 88836256-88836256 on Build GRCh38
rs866240136 Chr.15: 88836373-88836373 on Build GRCh38
rs943627727 Chr.15: 88836160-88836160 on Build GRCh38
rs974595914 Chr.15: 88836588-88836588 on Build GRCh38
rs16942299 Chr.15: 88836438-88836438 on Build GRCh38
rs755150554 Chr.15: 88836426-88836426 on Build GRCh38
rs775539020 Chr.15: 88836212-88836212 on Build GRCh38
rs367732871 Chr.15: 88836304-88836304 on Build GRCh38
rs1046495964 Chr.15: 88836334-88836334 on Build GRCh38
rs909004295 Chr.15: 88836387-88836387 on Build GRCh38
rs371016456 Chr.15: 88836286-88836286 on Build GRCh38
rs748913067 Chr.15: 88836177-88836177 on Build GRCh38
rs1021809746 Chr.15: 88836472-88836472 on Build GRCh38
rs987536287 Chr.15: 88836448-88836448 on Build GRCh38
rs781402812 Chr.15: 88836303-88836303 on Build GRCh38
rs775698414 Chr.15: 88836321-88836321 on Build GRCh38
rs960271155 Chr.15: 88836518-88836518 on Build GRCh38
rs765345363 Chr.15: 88836358-88836358 on Build GRCh38

View Full Product Details