Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 89585696-89586241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TICRR
Entrez Gene ID
90381
Gene Name
TOPBP1 interacting checkpoint and replication regulator
Gene Aliases
C15orf42, SLD3, Treslin
UniGene
Hs.441708
Gene Chromosome Location
Chr.15: 89575587-89628023 on Build GRCh38
Transcripts
NM_001308025.1 NM_152259.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1177372 c.1389G>A p.S463S Chr.15: 89585920-89585920 on Build GRCh38
3361580 c.1212C>T p.P404P Chr.15: 89585743-89585743 on Build GRCh38
383871 c.1351G>A p.D451N Chr.15: 89585882-89585882 on Build GRCh38
1608806 c.1267C>T p.R423C Chr.15: 89585798-89585798 on Build GRCh38
7393834 c.1382A>G p.H461R Chr.15: 89585913-89585913 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs968888218 Chr.15: 89585985-89585985 on Build GRCh38
rs921802761 Chr.15: 89586159-89586159 on Build GRCh38
rs548831287 Chr.15: 89585828-89585828 on Build GRCh38
rs1048496133 Chr.15: 89585767-89585767 on Build GRCh38
rs112898657 Chr.15: 89585981-89585981 on Build GRCh38
rs746206801 Chr.15: 89585968-89585969 on Build GRCh38
rs753995122 Chr.15: 89585969-89585969 on Build GRCh38
rs776393142 Chr.15: 89585916-89585916 on Build GRCh38
rs942770811 Chr.15: 89585822-89585822 on Build GRCh38
rs933144033 Chr.15: 89586167-89586167 on Build GRCh38
rs185009717 Chr.15: 89586175-89586175 on Build GRCh38
rs1055722714 Chr.15: 89585912-89585912 on Build GRCh38
rs753690295 Chr.15: 89586032-89586032 on Build GRCh38
rs544987715 Chr.15: 89585744-89585744 on Build GRCh38
rs190266555 Chr.15: 89586053-89586053 on Build GRCh38
rs757472988 Chr.15: 89585786-89585786 on Build GRCh38
rs960186663 Chr.15: 89586006-89586006 on Build GRCh38
rs376881571 Chr.15: 89585774-89585774 on Build GRCh38
rs756279503 Chr.15: 89585875-89585875 on Build GRCh38
rs893017916 Chr.15: 89586005-89586005 on Build GRCh38
rs201206531 Chr.15: 89585855-89585855 on Build GRCh38
rs577647790 Chr.15: 89586075-89586075 on Build GRCh38
rs750477340 Chr.15: 89585960-89585961 on Build GRCh38
rs11629584 Chr.15: 89585735-89585735 on Build GRCh38
rs778991598 Chr.15: 89585884-89585884 on Build GRCh38
rs370553025 Chr.15: 89585798-89585798 on Build GRCh38
rs746832520 Chr.15: 89585913-89585913 on Build GRCh38
rs758811891 Chr.15: 89585765-89585765 on Build GRCh38
rs1039788705 Chr.15: 89586118-89586118 on Build GRCh38
rs547995043 Chr.15: 89586034-89586034 on Build GRCh38
rs181686852 Chr.15: 89586103-89586103 on Build GRCh38
rs563412740 Chr.15: 89586135-89586135 on Build GRCh38
rs1020245947 Chr.15: 89586020-89586020 on Build GRCh38
rs368492482 Chr.15: 89585976-89585976 on Build GRCh38
rs780227765 Chr.15: 89585962-89585962 on Build GRCh38
rs144998725 Chr.15: 89585761-89585761 on Build GRCh38
rs1540504 Chr.15: 89585957-89585957 on Build GRCh38
rs147564898 Chr.15: 89585858-89585858 on Build GRCh38
rs375688083 Chr.15: 89586114-89586114 on Build GRCh38
rs747297194 Chr.15: 89585780-89585780 on Build GRCh38
rs781072768 Chr.15: 89585792-89585792 on Build GRCh38
rs370946859 Chr.15: 89585826-89585826 on Build GRCh38
rs537462042 Chr.15: 89586007-89586007 on Build GRCh38
rs542219933 Chr.15: 89586180-89586180 on Build GRCh38
rs150074180 Chr.15: 89586207-89586207 on Build GRCh38
rs1540503 Chr.15: 89585996-89585996 on Build GRCh38
rs868062880 Chr.15: 89585799-89585799 on Build GRCh38
rs765805643 Chr.15: 89585752-89585752 on Build GRCh38
rs920766820 Chr.15: 89586136-89586136 on Build GRCh38
rs2165155 Chr.15: 89586216-89586216 on Build GRCh38
rs777664042 Chr.15: 89585904-89585904 on Build GRCh38
rs751878810 Chr.15: 89585939-89585939 on Build GRCh38
rs556047273 Chr.15: 89586027-89586027 on Build GRCh38
rs983661656 Chr.15: 89586208-89586208 on Build GRCh38
rs564080651 Chr.15: 89585733-89585733 on Build GRCh38
rs8024752 Chr.15: 89586028-89586028 on Build GRCh38
rs139386417 Chr.15: 89585873-89585873 on Build GRCh38
rs111907494 Chr.15: 89585931-89585931 on Build GRCh38
rs201748816 Chr.15: 89585739-89585739 on Build GRCh38
rs767260029 Chr.15: 89585726-89585726 on Build GRCh38
rs928068688 Chr.15: 89585854-89585854 on Build GRCh38
rs763889492 Chr.15: 89586014-89586014 on Build GRCh38
rs750727324 Chr.15: 89585956-89585956 on Build GRCh38
rs185371124 Chr.15: 89586003-89586003 on Build GRCh38
rs865850609 Chr.15: 89586090-89586090 on Build GRCh38
rs200018187 Chr.15: 89585859-89585859 on Build GRCh38
rs982546733 Chr.15: 89586040-89586040 on Build GRCh38
rs747948490 Chr.15: 89585800-89585800 on Build GRCh38
rs528414553 Chr.15: 89586214-89586214 on Build GRCh38
rs571857728 Chr.15: 89585801-89585801 on Build GRCh38
rs373277423 Chr.15: 89585811-89585811 on Build GRCh38
rs767622900 Chr.15: 89585951-89585951 on Build GRCh38
rs553340283 Chr.15: 89586067-89586067 on Build GRCh38
rs765638752 Chr.15: 89585844-89585844 on Build GRCh38
rs748034619 Chr.15: 89585895-89585895 on Build GRCh38
rs112376978 Chr.15: 89585775-89585775 on Build GRCh38
rs772149858 Chr.15: 89585897-89585897 on Build GRCh38
rs758956031 Chr.15: 89585919-89585919 on Build GRCh38
rs770486089 Chr.15: 89585825-89585825 on Build GRCh38
rs371781392 Chr.15: 89585943-89585943 on Build GRCh38
rs781504186 Chr.15: 89585864-89585864 on Build GRCh38
rs376340305 Chr.15: 89585851-89585851 on Build GRCh38
rs532927184 Chr.15: 89585741-89585741 on Build GRCh38
rs774984664 Chr.15: 89585920-89585920 on Build GRCh38
rs139431492 Chr.15: 89585736-89585736 on Build GRCh38
rs367605057 Chr.15: 89585914-89585914 on Build GRCh38
rs779146878 Chr.15: 89585737-89585738 on Build GRCh38
rs111488829 Chr.15: 89585990-89585990 on Build GRCh38
rs764372307 Chr.15: 89585857-89585857 on Build GRCh38
rs900266452 Chr.15: 89586002-89586002 on Build GRCh38
rs759125274 Chr.15: 89585837-89585837 on Build GRCh38
rs763389729 Chr.15: 89585925-89585925 on Build GRCh38
rs376813582 Chr.15: 89585823-89585823 on Build GRCh38

View Full Product Details