Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 89790310-89790817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANPEP
Entrez Gene ID
290
Gene Name
alanyl aminopeptidase, membrane
Gene Aliases
APN, CD13, GP150, LAP1, P150, PEPN
UniGene
Hs.1239
Gene Chromosome Location
Chr.15: 89784895-89814852 on Build GRCh38
Transcripts
NM_001150.2 XM_005254892.4 XM_011521473.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2139599 c.2719C>T p.R907* Chr.15: 89790492-89790492 on Build GRCh38
702158 c.2734G>A p.E912K Chr.15: 89790477-89790477 on Build GRCh38
7179436 c.2715G>C p.V905V Chr.15: 89790496-89790496 on Build GRCh38
966401 c.2693_2695delTCT p.F898delF Chr.15: 89790516-89790518 on Build GRCh38
5833000 c.2672_2674delATG p.Y891_G892>C Chr.15: 89790537-89790539 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764473840 Chr.15: 89790502-89790502 on Build GRCh38
rs187084546 Chr.15: 89790433-89790433 on Build GRCh38
rs932905885 Chr.15: 89790699-89790699 on Build GRCh38
rs764769952 Chr.15: 89790554-89790554 on Build GRCh38
rs902636391 Chr.15: 89790716-89790716 on Build GRCh38
rs764741981 Chr.15: 89790472-89790472 on Build GRCh38
rs757565877 Chr.15: 89790512-89790512 on Build GRCh38
rs767821671 Chr.15: 89790589-89790589 on Build GRCh38
rs753161909 Chr.15: 89790489-89790489 on Build GRCh38
rs931891996 Chr.15: 89790666-89790666 on Build GRCh38
rs967284426 Chr.15: 89790360-89790360 on Build GRCh38
rs541460633 Chr.15: 89790782-89790782 on Build GRCh38
rs1032959458 Chr.15: 89790610-89790610 on Build GRCh38
rs1992250 Chr.15: 89790749-89790749 on Build GRCh38
rs756323779 Chr.15: 89790537-89790539 on Build GRCh38
rs536876812 Chr.15: 89790751-89790751 on Build GRCh38
rs762359560 Chr.15: 89790574-89790574 on Build GRCh38
rs754015242 Chr.15: 89790553-89790554 on Build GRCh38
rs182752150 Chr.15: 89790384-89790384 on Build GRCh38
rs768095085 Chr.15: 89790715-89790715 on Build GRCh38
rs770031010 Chr.15: 89790473-89790473 on Build GRCh38
rs756198835 Chr.15: 89790527-89790527 on Build GRCh38
rs1024137400 Chr.15: 89790790-89790790 on Build GRCh38
rs149507309 Chr.15: 89790710-89790710 on Build GRCh38
rs764266719 Chr.15: 89790546-89790569 on Build GRCh38
rs979119001 Chr.15: 89790365-89790365 on Build GRCh38
rs201931192 Chr.15: 89790588-89790588 on Build GRCh38
rs759150965 Chr.15: 89790469-89790469 on Build GRCh38
rs866381238 Chr.15: 89790402-89790402 on Build GRCh38
rs8192299 Chr.15: 89790385-89790385 on Build GRCh38
rs925770864 Chr.15: 89790379-89790379 on Build GRCh38
rs751522046 Chr.15: 89790457-89790458 on Build GRCh38
rs150785426 Chr.15: 89790414-89790414 on Build GRCh38
rs776451769 Chr.15: 89790547-89790547 on Build GRCh38
rs548766181 Chr.15: 89790793-89790793 on Build GRCh38
rs138092014 Chr.15: 89790415-89790415 on Build GRCh38
rs778922354 Chr.15: 89790421-89790421 on Build GRCh38
rs547347224 Chr.15: 89790353-89790353 on Build GRCh38
rs1035136461 Chr.15: 89790499-89790499 on Build GRCh38
rs1012848358 Chr.15: 89790783-89790783 on Build GRCh38
rs921491545 Chr.15: 89790693-89790693 on Build GRCh38
rs773007870 Chr.15: 89790538-89790538 on Build GRCh38
rs768651053 Chr.15: 89790342-89790342 on Build GRCh38
rs1054343955 Chr.15: 89790678-89790678 on Build GRCh38
rs8192298 Chr.15: 89790423-89790423 on Build GRCh38
rs900450297 Chr.15: 89790388-89790388 on Build GRCh38
rs1035170787 Chr.15: 89790733-89790733 on Build GRCh38
rs770780272 Chr.15: 89790543-89790543 on Build GRCh38
rs1033701129 Chr.15: 89790755-89790755 on Build GRCh38
rs770961758 Chr.15: 89790424-89790424 on Build GRCh38
rs1803099 Chr.15: 89790481-89790481 on Build GRCh38
rs564546671 Chr.15: 89790612-89790612 on Build GRCh38
rs776376212 Chr.15: 89790442-89790442 on Build GRCh38
rs533076761 Chr.15: 89790740-89790740 on Build GRCh38
rs17239917 Chr.15: 89790620-89790620 on Build GRCh38
rs746021797 Chr.15: 89790526-89790526 on Build GRCh38
rs528993669 Chr.15: 89790680-89790680 on Build GRCh38
rs368934787 Chr.15: 89790508-89790508 on Build GRCh38
rs766184929 Chr.15: 89790439-89790439 on Build GRCh38
rs555994765 Chr.15: 89790657-89790657 on Build GRCh38
rs7167568 Chr.15: 89790720-89790720 on Build GRCh38
rs544014752 Chr.15: 89790649-89790649 on Build GRCh38
rs1020943954 Chr.15: 89790566-89790566 on Build GRCh38
rs561792946 Chr.15: 89790661-89790661 on Build GRCh38
rs117177414 Chr.15: 89790750-89790750 on Build GRCh38
rs941051866 Chr.15: 89790663-89790663 on Build GRCh38
rs8192301 Chr.15: 89790396-89790396 on Build GRCh38
rs763321827 Chr.15: 89790578-89790578 on Build GRCh38
rs1051223150 Chr.15: 89790477-89790477 on Build GRCh38
rs556275270 Chr.15: 89790417-89790417 on Build GRCh38
rs1021374979 Chr.15: 89790356-89790356 on Build GRCh38
rs8192300 Chr.15: 89790334-89790334 on Build GRCh38
rs746812140 Chr.15: 89790539-89790539 on Build GRCh38
rs991298695 Chr.15: 89790647-89790647 on Build GRCh38
rs946760329 Chr.15: 89790698-89790698 on Build GRCh38
rs1039066059 Chr.15: 89790431-89790431 on Build GRCh38
rs759763680 Chr.15: 89790650-89790650 on Build GRCh38
rs866601955 Chr.15: 89790368-89790368 on Build GRCh38
rs759006687 Chr.15: 89790497-89790497 on Build GRCh38
rs944425434 Chr.15: 89790635-89790635 on Build GRCh38
rs1040213927 Chr.15: 89790386-89790386 on Build GRCh38
rs371867461 Chr.15: 89790571-89790571 on Build GRCh38
rs554129147 Chr.15: 89790549-89790549 on Build GRCh38
rs1042887103 Chr.15: 89790743-89790743 on Build GRCh38
rs200346681 Chr.15: 89790579-89790579 on Build GRCh38
rs749297050 Chr.15: 89790537-89790537 on Build GRCh38

View Full Product Details