Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 89790871-89791360 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANPEP
Entrez Gene ID
290
Gene Name
alanyl aminopeptidase, membrane
Gene Aliases
APN, CD13, GP150, LAP1, P150, PEPN
UniGene
Hs.1239
Gene Chromosome Location
Chr.15: 89784895-89814852 on Build GRCh38
Transcripts
NM_001150.2 XM_005254892.4 XM_011521473.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1180823 c.2664delT p.F888fs*18 Chr.15: 89790958-89790958 on Build GRCh38
4128566 c.2613T>C p.I871I Chr.15: 89791009-89791009 on Build GRCh38
5012934 c.2635T>C p.F879L Chr.15: 89790987-89790987 on Build GRCh38
7120163 c.2668G>T p.D890Y Chr.15: 89790954-89790954 on Build GRCh38
5470415 c.2602A>G p.N868D Chr.15: 89791020-89791020 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35391151 Chr.15: 89790945-89790946 on Build GRCh38
rs7166568 Chr.15: 89790902-89790902 on Build GRCh38
rs758137651 Chr.15: 89790898-89790898 on Build GRCh38
rs74766672 Chr.15: 89791078-89791078 on Build GRCh38
rs544074967 Chr.15: 89790921-89790921 on Build GRCh38
rs776930737 Chr.15: 89791060-89791060 on Build GRCh38
rs773650895 Chr.15: 89791051-89791051 on Build GRCh38
rs762357050 Chr.15: 89790933-89790933 on Build GRCh38
rs973825926 Chr.15: 89791090-89791090 on Build GRCh38
rs772363423 Chr.15: 89791228-89791228 on Build GRCh38
rs144085303 Chr.15: 89790973-89790973 on Build GRCh38
rs1015428579 Chr.15: 89791066-89791066 on Build GRCh38
rs777268404 Chr.15: 89791216-89791217 on Build GRCh38
rs376865285 Chr.15: 89791134-89791134 on Build GRCh38
rs1049246877 Chr.15: 89791163-89791163 on Build GRCh38
rs758736666 Chr.15: 89791095-89791096 on Build GRCh38
rs1021462362 Chr.15: 89791251-89791251 on Build GRCh38
rs79149569 Chr.15: 89791323-89791323 on Build GRCh38
rs779172784 Chr.15: 89790942-89790943 on Build GRCh38
rs773797848 Chr.15: 89791136-89791136 on Build GRCh38
rs772625359 Chr.15: 89791044-89791044 on Build GRCh38
rs879672554 Chr.15: 89791328-89791328 on Build GRCh38
rs773892132 Chr.15: 89791032-89791032 on Build GRCh38
rs945567797 Chr.15: 89790924-89790924 on Build GRCh38
rs774951046 Chr.15: 89790918-89790918 on Build GRCh38
rs779764342 Chr.15: 89791098-89791098 on Build GRCh38
rs1004139689 Chr.15: 89791236-89791236 on Build GRCh38
rs572377622 Chr.15: 89790900-89790900 on Build GRCh38
rs7168329 Chr.15: 89791114-89791114 on Build GRCh38
rs1029065232 Chr.15: 89791223-89791223 on Build GRCh38
rs148098345 Chr.15: 89791042-89791042 on Build GRCh38
rs757448256 Chr.15: 89790932-89790933 on Build GRCh38
rs767570451 Chr.15: 89791080-89791080 on Build GRCh38
rs748800977 Chr.15: 89791131-89791131 on Build GRCh38
rs768013713 Chr.15: 89790922-89790922 on Build GRCh38
rs539849281 Chr.15: 89791305-89791305 on Build GRCh38
rs949393920 Chr.15: 89791204-89791204 on Build GRCh38
rs967150932 Chr.15: 89791296-89791296 on Build GRCh38
rs781216760 Chr.15: 89790923-89790923 on Build GRCh38
rs25654 Chr.15: 89791009-89791009 on Build GRCh38
rs754006589 Chr.15: 89790905-89790905 on Build GRCh38
rs754954275 Chr.15: 89790908-89790908 on Build GRCh38
rs143655390 Chr.15: 89791015-89791015 on Build GRCh38
rs374844432 Chr.15: 89790904-89790904 on Build GRCh38
rs916565974 Chr.15: 89791199-89791199 on Build GRCh38
rs920936294 Chr.15: 89790901-89790901 on Build GRCh38
rs756105308 Chr.15: 89791025-89791025 on Build GRCh38
rs372938510 Chr.15: 89791079-89791079 on Build GRCh38
rs925484078 Chr.15: 89791164-89791164 on Build GRCh38
rs760934157 Chr.15: 89790950-89790950 on Build GRCh38
rs767981424 Chr.15: 89790937-89790937 on Build GRCh38
rs936732315 Chr.15: 89791183-89791183 on Build GRCh38
rs201439023 Chr.15: 89790955-89790955 on Build GRCh38
rs200363198 Chr.15: 89791100-89791100 on Build GRCh38
rs771502186 Chr.15: 89791135-89791135 on Build GRCh38
rs7169161 Chr.15: 89791184-89791184 on Build GRCh38
rs765520167 Chr.15: 89791070-89791070 on Build GRCh38
rs535598126 Chr.15: 89791213-89791213 on Build GRCh38
rs6496604 Chr.15: 89791246-89791246 on Build GRCh38
rs907075545 Chr.15: 89791225-89791225 on Build GRCh38
rs1012837008 Chr.15: 89791018-89791018 on Build GRCh38
rs755962012 Chr.15: 89791097-89791097 on Build GRCh38
rs566024664 Chr.15: 89791250-89791250 on Build GRCh38
rs750290072 Chr.15: 89791089-89791089 on Build GRCh38
rs878933575 Chr.15: 89791177-89791177 on Build GRCh38
rs747422950 Chr.15: 89791138-89791138 on Build GRCh38
rs973670191 Chr.15: 89790891-89790891 on Build GRCh38
rs887904368 Chr.15: 89791180-89791180 on Build GRCh38
rs1005143484 Chr.15: 89791182-89791182 on Build GRCh38
rs572391101 Chr.15: 89790914-89790914 on Build GRCh38
rs761103812 Chr.15: 89791058-89791058 on Build GRCh38
rs986997722 Chr.15: 89791102-89791102 on Build GRCh38
rs759722593 Chr.15: 89791069-89791069 on Build GRCh38
rs746281500 Chr.15: 89790957-89790958 on Build GRCh38
rs200383502 Chr.15: 89790979-89790979 on Build GRCh38
rs77935658 Chr.15: 89790915-89790915 on Build GRCh38
rs201233331 Chr.15: 89791125-89791125 on Build GRCh38
rs967007676 Chr.15: 89791123-89791123 on Build GRCh38
rs750853759 Chr.15: 89790942-89790942 on Build GRCh38
rs1027942019 Chr.15: 89791093-89791093 on Build GRCh38
rs752797827 Chr.15: 89790984-89790984 on Build GRCh38
rs781266711 Chr.15: 89790996-89790996 on Build GRCh38
rs182894281 Chr.15: 89790917-89790917 on Build GRCh38
rs752887360 Chr.15: 89791071-89791071 on Build GRCh38
rs768277577 Chr.15: 89791019-89791019 on Build GRCh38
rs781008050 Chr.15: 89791014-89791014 on Build GRCh38
rs369201239 Chr.15: 89791059-89791059 on Build GRCh38
rs567076190 Chr.15: 89791202-89791202 on Build GRCh38
rs757296390 Chr.15: 89790994-89790994 on Build GRCh38
rs553883672 Chr.15: 89790895-89790895 on Build GRCh38
rs761230301 Chr.15: 89791282-89791282 on Build GRCh38
rs766635145 Chr.15: 89790954-89790954 on Build GRCh38
rs376323078 Chr.15: 89790972-89790972 on Build GRCh38
rs745692624 Chr.15: 89790999-89790999 on Build GRCh38
rs775305541 Chr.15: 89790919-89790919 on Build GRCh38
rs778063366 Chr.15: 89791067-89791067 on Build GRCh38
rs953170959 Chr.15: 89791238-89791238 on Build GRCh38
rs774862244 Chr.15: 89790929-89790929 on Build GRCh38
rs928595922 Chr.15: 89791339-89791339 on Build GRCh38
rs948104736 Chr.15: 89791155-89791155 on Build GRCh38

View Full Product Details