Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 89835574-89836072 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP3S2
Entrez Gene ID
10239
Gene Name
adaptor related protein complex 3 sigma 2 subunit
Gene Aliases
AP3S3, sigma3b
UniGene
Hs.632161
Gene Chromosome Location
Chr.15: 89830599-89894385 on Build GRCh38
Transcripts
NM_005829.4 NR_023361.1 NR_037582.1 NM_001199058.1

Gene Symbol
C15orf38-AP3S2
Entrez Gene ID
100526783
Gene Name
C15orf38-AP3S2 readthrough
Gene Aliases
-
UniGene
Hs.744890
Gene Chromosome Location
Chr.15: 89830599-89912990 on Build GRCh38
Transcripts
NM_005829.4 NR_023361.1 NR_037582.1 NM_001199058.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
702147 c.477G>T p.A159A Chr.15: 89835620-89835620 on Build GRCh38
6242408 c.475G>A p.A159T Chr.15: 89835622-89835622 on Build GRCh38
7341105 c.523C>T p.R175W Chr.15: 89835574-89835574 on Build GRCh38
702148 c.513A>G p.P171P Chr.15: 89835584-89835584 on Build GRCh38
5749675 c.505A>T p.N169Y Chr.15: 89835592-89835592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770651629 Chr.15: 89835613-89835613 on Build GRCh38
rs201507770 Chr.15: 89836042-89836045 on Build GRCh38
rs934626446 Chr.15: 89835912-89835912 on Build GRCh38
rs981651606 Chr.15: 89835917-89835917 on Build GRCh38
rs923385919 Chr.15: 89835911-89835911 on Build GRCh38
rs563795132 Chr.15: 89835765-89835765 on Build GRCh38
rs929628485 Chr.15: 89836037-89836037 on Build GRCh38
rs1026500097 Chr.15: 89835794-89835794 on Build GRCh38
rs780857665 Chr.15: 89835614-89835614 on Build GRCh38
rs976532495 Chr.15: 89835897-89835897 on Build GRCh38
rs896250272 Chr.15: 89835954-89835954 on Build GRCh38
rs1034690600 Chr.15: 89835774-89835774 on Build GRCh38
rs946646179 Chr.15: 89835930-89835930 on Build GRCh38
rs777422761 Chr.15: 89835602-89835602 on Build GRCh38
rs778860299 Chr.15: 89835597-89835597 on Build GRCh38
rs752550716 Chr.15: 89835600-89835600 on Build GRCh38
rs762729044 Chr.15: 89835872-89835872 on Build GRCh38
rs192949364 Chr.15: 89835969-89835969 on Build GRCh38
rs752743482 Chr.15: 89835646-89835646 on Build GRCh38
rs913804675 Chr.15: 89835925-89835925 on Build GRCh38
rs778674508 Chr.15: 89835642-89835642 on Build GRCh38
rs200425463 Chr.15: 89835730-89835730 on Build GRCh38
rs1043573010 Chr.15: 89835932-89835932 on Build GRCh38
rs755691802 Chr.15: 89835675-89835675 on Build GRCh38
rs976287917 Chr.15: 89836031-89836031 on Build GRCh38
rs369148847 Chr.15: 89835797-89835797 on Build GRCh38
rs764056325 Chr.15: 89835703-89835704 on Build GRCh38
rs758251712 Chr.15: 89835661-89835661 on Build GRCh38
rs927581588 Chr.15: 89835638-89835638 on Build GRCh38
rs965204632 Chr.15: 89836023-89836023 on Build GRCh38
rs745703206 Chr.15: 89835642-89835643 on Build GRCh38
rs993186548 Chr.15: 89835793-89835793 on Build GRCh38
rs749866510 Chr.15: 89835690-89835690 on Build GRCh38
rs988831266 Chr.15: 89835916-89835916 on Build GRCh38
rs865809167 Chr.15: 89835856-89835856 on Build GRCh38
rs745335818 Chr.15: 89835616-89835616 on Build GRCh38
rs143234818 Chr.15: 89835617-89835617 on Build GRCh38
rs957403103 Chr.15: 89835854-89835854 on Build GRCh38
rs112350158 Chr.15: 89836027-89836030 on Build GRCh38
rs768976238 Chr.15: 89835695-89835695 on Build GRCh38
rs1017813667 Chr.15: 89835991-89835991 on Build GRCh38
rs760853637 Chr.15: 89835630-89835630 on Build GRCh38
rs759846821 Chr.15: 89836027-89836027 on Build GRCh38
rs112739850 Chr.15: 89835620-89835620 on Build GRCh38
rs1962662 Chr.15: 89835776-89835776 on Build GRCh38
rs538209457 Chr.15: 89835612-89835612 on Build GRCh38
rs762025364 Chr.15: 89835622-89835622 on Build GRCh38
rs974354222 Chr.15: 89836011-89836011 on Build GRCh38
rs1032209590 Chr.15: 89835846-89835846 on Build GRCh38
rs542611048 Chr.15: 89835960-89835960 on Build GRCh38
rs147800094 Chr.15: 89835871-89835871 on Build GRCh38
rs776607280 Chr.15: 89835619-89835619 on Build GRCh38
rs529216885 Chr.15: 89835942-89835942 on Build GRCh38
rs1025965062 Chr.15: 89835968-89835968 on Build GRCh38
rs766542726 Chr.15: 89835896-89835896 on Build GRCh38
rs776911457 Chr.15: 89835702-89835704 on Build GRCh38
rs547668456 Chr.15: 89835947-89835947 on Build GRCh38
rs758249917 Chr.15: 89835601-89835601 on Build GRCh38
rs769659993 Chr.15: 89835826-89835826 on Build GRCh38
rs919619774 Chr.15: 89835733-89835733 on Build GRCh38
rs779426951 Chr.15: 89835683-89835683 on Build GRCh38
rs745494677 Chr.15: 89835669-89835669 on Build GRCh38
rs369373472 Chr.15: 89835723-89835724 on Build GRCh38
rs776114232 Chr.15: 89835618-89835618 on Build GRCh38
rs1000763259 Chr.15: 89835832-89835832 on Build GRCh38
rs188914042 Chr.15: 89835663-89835663 on Build GRCh38
rs552374461 Chr.15: 89836043-89836046 on Build GRCh38
rs5814404 Chr.15: 89835722-89835722 on Build GRCh38
rs773302840 Chr.15: 89835625-89835625 on Build GRCh38
rs780947613 Chr.15: 89835666-89835666 on Build GRCh38
rs984910357 Chr.15: 89835859-89835859 on Build GRCh38
rs376854476 Chr.15: 89835643-89835643 on Build GRCh38
rs1020371145 Chr.15: 89835891-89835891 on Build GRCh38
rs545581831 Chr.15: 89835747-89835747 on Build GRCh38
rs552721414 Chr.15: 89836020-89836020 on Build GRCh38
rs371762248 Chr.15: 89835724-89835724 on Build GRCh38
rs1002397368 Chr.15: 89835766-89835766 on Build GRCh38
rs1006467546 Chr.15: 89835976-89835976 on Build GRCh38
rs929618615 Chr.15: 89835761-89835761 on Build GRCh38
rs140842385 Chr.15: 89835662-89835662 on Build GRCh38
rs371542418 Chr.15: 89835937-89835937 on Build GRCh38
rs767686464 Chr.15: 89835623-89835623 on Build GRCh38
rs774639100 Chr.15: 89835621-89835621 on Build GRCh38
rs1052315414 Chr.15: 89835626-89835626 on Build GRCh38

View Full Product Details