Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 89876919-89877418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP3S2
Entrez Gene ID
10239
Gene Name
adaptor related protein complex 3 sigma 2 subunit
Gene Aliases
AP3S3, sigma3b
UniGene
Hs.632161
Gene Chromosome Location
Chr.15: 89830599-89894385 on Build GRCh38
Transcripts
NM_005829.4 NR_023361.1 NR_037582.1 NM_001199058.1

Gene Symbol
C15orf38-AP3S2
Entrez Gene ID
100526783
Gene Name
C15orf38-AP3S2 readthrough
Gene Aliases
-
UniGene
Hs.744890
Gene Chromosome Location
Chr.15: 89830599-89912990 on Build GRCh38
Transcripts
NM_005829.4 NR_023361.1 NR_037582.1 NM_001199058.1

SNP Detail

SNP ID Chromosome Location
rs750752993 Chr.15: 89877341-89877341 on Build GRCh38
rs771295642 Chr.15: 89877334-89877334 on Build GRCh38
rs915258264 Chr.15: 89877171-89877171 on Build GRCh38
rs749486817 Chr.15: 89877347-89877347 on Build GRCh38
rs770832996 Chr.15: 89877380-89877380 on Build GRCh38
rs1042677940 Chr.15: 89877019-89877019 on Build GRCh38
rs945746400 Chr.15: 89876996-89876996 on Build GRCh38
rs1042709354 Chr.15: 89877272-89877272 on Build GRCh38
rs547343586 Chr.15: 89877215-89877215 on Build GRCh38
rs974653021 Chr.15: 89877222-89877222 on Build GRCh38
rs779037751 Chr.15: 89877362-89877362 on Build GRCh38
rs1011904282 Chr.15: 89876988-89876988 on Build GRCh38
rs567630809 Chr.15: 89877058-89877058 on Build GRCh38
rs770689229 Chr.15: 89877284-89877284 on Build GRCh38
rs763629378 Chr.15: 89877319-89877319 on Build GRCh38
rs187112700 Chr.15: 89877254-89877254 on Build GRCh38
rs527343339 Chr.15: 89877151-89877154 on Build GRCh38
rs1014710813 Chr.15: 89877387-89877387 on Build GRCh38
rs984133828 Chr.15: 89877118-89877118 on Build GRCh38
rs896039181 Chr.15: 89877335-89877335 on Build GRCh38
rs921884691 Chr.15: 89877104-89877104 on Build GRCh38
rs776427974 Chr.15: 89877287-89877287 on Build GRCh38
rs987254493 Chr.15: 89877252-89877252 on Build GRCh38
rs752072645 Chr.15: 89877326-89877326 on Build GRCh38
rs951429943 Chr.15: 89877094-89877094 on Build GRCh38
rs762500034 Chr.15: 89877312-89877312 on Build GRCh38
rs1015050780 Chr.15: 89877061-89877061 on Build GRCh38
rs114847603 Chr.15: 89877342-89877342 on Build GRCh38
rs548333206 Chr.15: 89876954-89876954 on Build GRCh38
rs953258599 Chr.15: 89877107-89877107 on Build GRCh38
rs778711210 Chr.15: 89877163-89877163 on Build GRCh38
rs1027256391 Chr.15: 89877037-89877037 on Build GRCh38
rs1001756664 Chr.15: 89877044-89877044 on Build GRCh38
rs867742226 Chr.15: 89877082-89877082 on Build GRCh38
rs536308029 Chr.15: 89877329-89877329 on Build GRCh38
rs1034940041 Chr.15: 89877048-89877048 on Build GRCh38
rs1014453236 Chr.15: 89877340-89877340 on Build GRCh38
rs528814307 Chr.15: 89877102-89877102 on Build GRCh38
rs891541944 Chr.15: 89876974-89876974 on Build GRCh38
rs1004923240 Chr.15: 89877355-89877355 on Build GRCh38
rs1016920901 Chr.15: 89877161-89877161 on Build GRCh38
rs144254794 Chr.15: 89877248-89877248 on Build GRCh38
rs182010295 Chr.15: 89877042-89877042 on Build GRCh38
rs755075313 Chr.15: 89877351-89877351 on Build GRCh38
rs746932427 Chr.15: 89877372-89877372 on Build GRCh38
rs1025757580 Chr.15: 89877089-89877089 on Build GRCh38
rs72756562 Chr.15: 89877247-89877247 on Build GRCh38
rs759191690 Chr.15: 89877290-89877290 on Build GRCh38
rs1014834894 Chr.15: 89877087-89877087 on Build GRCh38
rs746956589 Chr.15: 89877280-89877280 on Build GRCh38
rs965915163 Chr.15: 89877081-89877081 on Build GRCh38
rs764828000 Chr.15: 89877297-89877297 on Build GRCh38
rs191653751 Chr.15: 89877271-89877271 on Build GRCh38
rs36115166 Chr.15: 89877056-89877057 on Build GRCh38
rs775005209 Chr.15: 89877303-89877303 on Build GRCh38
rs904803705 Chr.15: 89877041-89877041 on Build GRCh38
rs912999626 Chr.15: 89877258-89877258 on Build GRCh38
rs887207492 Chr.15: 89877371-89877371 on Build GRCh38

View Full Product Details