Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90530100-90530590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTC3
Entrez Gene ID
64784
Gene Name
CREB regulated transcription coactivator 3
Gene Aliases
TORC-3, TORC3
UniGene
Hs.567572
Gene Chromosome Location
Chr.15: 90529886-90645345 on Build GRCh38
Transcripts
NM_001042574.2 NM_022769.4 XM_005254968.2 XM_011521906.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3667771 c.60G>T p.A20A Chr.15: 90530131-90530131 on Build GRCh38
4632105 c.105G>A p.Q35Q Chr.15: 90530176-90530176 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs867583186 Chr.15: 90530173-90530173 on Build GRCh38
rs754618349 Chr.15: 90530221-90530242 on Build GRCh38
rs185055811 Chr.15: 90530278-90530278 on Build GRCh38
rs561774830 Chr.15: 90530214-90530214 on Build GRCh38
rs374653632 Chr.15: 90530238-90530238 on Build GRCh38
rs947107512 Chr.15: 90530533-90530533 on Build GRCh38
rs970129273 Chr.15: 90530218-90530219 on Build GRCh38
rs777148263 Chr.15: 90530213-90530213 on Build GRCh38
rs918259982 Chr.15: 90530277-90530277 on Build GRCh38
rs781735252 Chr.15: 90530237-90530237 on Build GRCh38
rs190838915 Chr.15: 90530324-90530324 on Build GRCh38
rs988583077 Chr.15: 90530481-90530481 on Build GRCh38
rs139118057 Chr.15: 90530233-90530238 on Build GRCh38
rs896999415 Chr.15: 90530549-90530549 on Build GRCh38
rs766247155 Chr.15: 90530145-90530145 on Build GRCh38
rs938620495 Chr.15: 90530544-90530544 on Build GRCh38
rs1016735059 Chr.15: 90530246-90530246 on Build GRCh38
rs770232230 Chr.15: 90530239-90530239 on Build GRCh38
rs1025539638 Chr.15: 90530498-90530498 on Build GRCh38
rs986462611 Chr.15: 90530520-90530520 on Build GRCh38
rs976270606 Chr.15: 90530463-90530463 on Build GRCh38
rs550916637 Chr.15: 90530433-90530433 on Build GRCh38
rs1056863740 Chr.15: 90530546-90530546 on Build GRCh38
rs757850149 Chr.15: 90530227-90530227 on Build GRCh38
rs757969797 Chr.15: 90530204-90530204 on Build GRCh38
rs926101566 Chr.15: 90530359-90530359 on Build GRCh38
rs760524428 Chr.15: 90530140-90530140 on Build GRCh38
rs549061946 Chr.15: 90530524-90530524 on Build GRCh38
rs1053404105 Chr.15: 90530425-90530425 on Build GRCh38
rs914222206 Chr.15: 90530499-90530499 on Build GRCh38
rs116192256 Chr.15: 90530438-90530438 on Build GRCh38
rs1044691960 Chr.15: 90530541-90530541 on Build GRCh38
rs1045756064 Chr.15: 90530461-90530461 on Build GRCh38
rs866420341 Chr.15: 90530139-90530139 on Build GRCh38
rs1047846981 Chr.15: 90530566-90530566 on Build GRCh38
rs754730082 Chr.15: 90530174-90530174 on Build GRCh38
rs1017859712 Chr.15: 90530534-90530534 on Build GRCh38
rs867540303 Chr.15: 90530130-90530130 on Build GRCh38
rs753634298 Chr.15: 90530152-90530152 on Build GRCh38
rs993949576 Chr.15: 90530558-90530558 on Build GRCh38
rs188476026 Chr.15: 90530197-90530197 on Build GRCh38
rs535016867 Chr.15: 90530552-90530552 on Build GRCh38
rs962950534 Chr.15: 90530255-90530255 on Build GRCh38
rs927237581 Chr.15: 90530543-90530543 on Build GRCh38
rs764056442 Chr.15: 90530119-90530119 on Build GRCh38
rs764893511 Chr.15: 90530179-90530179 on Build GRCh38
rs1039616289 Chr.15: 90530326-90530326 on Build GRCh38
rs180995634 Chr.15: 90530256-90530256 on Build GRCh38
rs532788446 Chr.15: 90530410-90530410 on Build GRCh38
rs955742315 Chr.15: 90530473-90530473 on Build GRCh38
rs866917303 Chr.15: 90530129-90530129 on Build GRCh38
rs779183674 Chr.15: 90530532-90530532 on Build GRCh38
rs986410392 Chr.15: 90530300-90530300 on Build GRCh38
rs911009719 Chr.15: 90530317-90530317 on Build GRCh38
rs780945415 Chr.15: 90530232-90530237 on Build GRCh38
rs867396572 Chr.15: 90530242-90530242 on Build GRCh38
rs543610628 Chr.15: 90530201-90530201 on Build GRCh38
rs1017297208 Chr.15: 90530473-90530486 on Build GRCh38
rs1014254002 Chr.15: 90530195-90530195 on Build GRCh38
rs773241141 Chr.15: 90530122-90530122 on Build GRCh38
rs887764898 Chr.15: 90530567-90530567 on Build GRCh38
rs111997730 Chr.15: 90530234-90530234 on Build GRCh38
rs936266265 Chr.15: 90530391-90530391 on Build GRCh38
rs530563336 Chr.15: 90530472-90530472 on Build GRCh38
rs867267649 Chr.15: 90530153-90530153 on Build GRCh38

View Full Product Details