Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90887097-90887591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FES
Entrez Gene ID
2242
Gene Name
FES proto-oncogene, tyrosine kinase
Gene Aliases
FPS
UniGene
Hs.7636
Gene Chromosome Location
Chr.15: 90884421-90895776 on Build GRCh38
Transcripts
NM_001143783.1 NM_001143784.1 NM_001143785.1 NM_002005.3 XM_005254880.1 XM_005254882.1 XM_017022005.1 XM_017022006.1 XM_017022007.1 XM_017022008.1 XM_017022009.1 XM_017022010.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3944328 c.618C>T p.P206P Chr.15: 90887320-90887320 on Build GRCh38
6663223 c.554G>A p.R185H Chr.15: 90887256-90887256 on Build GRCh38
7243831 c.564G>T p.L188L Chr.15: 90887266-90887266 on Build GRCh38
3505440 c.568G>A p.V190M Chr.15: 90887270-90887270 on Build GRCh38
3505438 c.490G>A p.D164N Chr.15: 90887192-90887192 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763029771 Chr.15: 90887321-90887321 on Build GRCh38
rs777383884 Chr.15: 90887282-90887282 on Build GRCh38
rs36113227 Chr.15: 90887445-90887445 on Build GRCh38
rs528288914 Chr.15: 90887469-90887469 on Build GRCh38
rs758936539 Chr.15: 90887363-90887363 on Build GRCh38
rs759560947 Chr.15: 90887174-90887174 on Build GRCh38
rs769517026 Chr.15: 90887492-90887492 on Build GRCh38
rs150679526 Chr.15: 90887369-90887369 on Build GRCh38
rs754861288 Chr.15: 90887386-90887386 on Build GRCh38
rs746544835 Chr.15: 90887378-90887378 on Build GRCh38
rs140247798 Chr.15: 90887269-90887269 on Build GRCh38
rs571259657 Chr.15: 90887480-90887480 on Build GRCh38
rs992559271 Chr.15: 90887546-90887546 on Build GRCh38
rs1018101780 Chr.15: 90887150-90887150 on Build GRCh38
rs770601368 Chr.15: 90887227-90887227 on Build GRCh38
rs749725382 Chr.15: 90887196-90887196 on Build GRCh38
rs546632838 Chr.15: 90887474-90887474 on Build GRCh38
rs1003297054 Chr.15: 90887130-90887131 on Build GRCh38
rs138511924 Chr.15: 90887255-90887255 on Build GRCh38
rs893812660 Chr.15: 90887323-90887323 on Build GRCh38
rs375285407 Chr.15: 90887393-90887393 on Build GRCh38
rs1039041282 Chr.15: 90887416-90887416 on Build GRCh38
rs922794480 Chr.15: 90887497-90887497 on Build GRCh38
rs35617264 Chr.15: 90887444-90887444 on Build GRCh38
rs201073806 Chr.15: 90887143-90887143 on Build GRCh38
rs772841979 Chr.15: 90887225-90887225 on Build GRCh38
rs750585753 Chr.15: 90887267-90887267 on Build GRCh38
rs779391057 Chr.15: 90887274-90887274 on Build GRCh38
rs757886415 Chr.15: 90887281-90887281 on Build GRCh38
rs766254990 Chr.15: 90887165-90887165 on Build GRCh38
rs765278179 Chr.15: 90887242-90887242 on Build GRCh38
rs775461048 Chr.15: 90887391-90887391 on Build GRCh38
rs747663762 Chr.15: 90887208-90887208 on Build GRCh38
rs946782172 Chr.15: 90887559-90887559 on Build GRCh38
rs755326118 Chr.15: 90887348-90887348 on Build GRCh38
rs757859464 Chr.15: 90887389-90887389 on Build GRCh38
rs761313675 Chr.15: 90887151-90887151 on Build GRCh38
rs926993421 Chr.15: 90887514-90887514 on Build GRCh38
rs373485327 Chr.15: 90887244-90887244 on Build GRCh38
rs779401815 Chr.15: 90887320-90887320 on Build GRCh38
rs960035488 Chr.15: 90887520-90887520 on Build GRCh38
rs769777721 Chr.15: 90887306-90887306 on Build GRCh38
rs748716030 Chr.15: 90887280-90887280 on Build GRCh38
rs750723414 Chr.15: 90887177-90887177 on Build GRCh38
rs926997032 Chr.15: 90887351-90887351 on Build GRCh38
rs114657862 Chr.15: 90887496-90887496 on Build GRCh38
rs568342645 Chr.15: 90887293-90887293 on Build GRCh38
rs377603197 Chr.15: 90887322-90887322 on Build GRCh38
rs377277703 Chr.15: 90887175-90887175 on Build GRCh38
rs1050290178 Chr.15: 90887513-90887513 on Build GRCh38
rs145301509 Chr.15: 90887295-90887295 on Build GRCh38
rs368888354 Chr.15: 90887353-90887353 on Build GRCh38
rs759344901 Chr.15: 90887237-90887237 on Build GRCh38
rs769233559 Chr.15: 90887203-90887203 on Build GRCh38
rs955857231 Chr.15: 90887448-90887448 on Build GRCh38
rs556532119 Chr.15: 90887504-90887504 on Build GRCh38
rs72755383 Chr.15: 90887176-90887176 on Build GRCh38
rs775554967 Chr.15: 90887311-90887311 on Build GRCh38
rs772926958 Chr.15: 90887158-90887158 on Build GRCh38
rs773888266 Chr.15: 90887149-90887149 on Build GRCh38
rs2071383 Chr.15: 90887206-90887206 on Build GRCh38
rs148010744 Chr.15: 90887305-90887305 on Build GRCh38
rs771729065 Chr.15: 90887157-90887157 on Build GRCh38
rs888070417 Chr.15: 90887402-90887402 on Build GRCh38
rs746581749 Chr.15: 90887379-90887379 on Build GRCh38
rs760756622 Chr.15: 90887330-90887330 on Build GRCh38
rs776229552 Chr.15: 90887299-90887299 on Build GRCh38
rs761979788 Chr.15: 90887249-90887249 on Build GRCh38
rs201771040 Chr.15: 90887222-90887222 on Build GRCh38
rs28438927 Chr.15: 90887417-90887417 on Build GRCh38
rs749916436 Chr.15: 90887171-90887172 on Build GRCh38
rs766630921 Chr.15: 90887341-90887341 on Build GRCh38
rs568549430 Chr.15: 90887552-90887552 on Build GRCh38
rs576327466 Chr.15: 90887195-90887195 on Build GRCh38
rs765546858 Chr.15: 90887422-90887422 on Build GRCh38
rs545069439 Chr.15: 90887327-90887327 on Build GRCh38
rs552812878 Chr.15: 90887122-90887122 on Build GRCh38
rs2071384 Chr.15: 90887387-90887387 on Build GRCh38
rs200508852 Chr.15: 90887153-90887153 on Build GRCh38
rs776522820 Chr.15: 90887235-90887235 on Build GRCh38
rs768657168 Chr.15: 90887395-90887395 on Build GRCh38
rs781261769 Chr.15: 90887414-90887414 on Build GRCh38
rs141159455 Chr.15: 90887191-90887191 on Build GRCh38
rs922763509 Chr.15: 90887256-90887256 on Build GRCh38
rs373751084 Chr.15: 90887551-90887551 on Build GRCh38
rs768025740 Chr.15: 90887146-90887146 on Build GRCh38
rs779639535 Chr.15: 90887395-90887396 on Build GRCh38
rs750047865 Chr.15: 90887270-90887270 on Build GRCh38
rs141941493 Chr.15: 90887350-90887350 on Build GRCh38
rs753696978 Chr.15: 90887166-90887166 on Build GRCh38
rs773105691 Chr.15: 90887400-90887400 on Build GRCh38
rs116140607 Chr.15: 90887118-90887118 on Build GRCh38
rs754078593 Chr.15: 90887344-90887344 on Build GRCh38
rs1034837962 Chr.15: 90887390-90887390 on Build GRCh38
rs759799074 Chr.15: 90887420-90887420 on Build GRCh38
rs745572655 Chr.15: 90887383-90887383 on Build GRCh38
rs375072112 Chr.15: 90887403-90887403 on Build GRCh38
rs777099415 Chr.15: 90887374-90887374 on Build GRCh38
rs373484714 Chr.15: 90887162-90887162 on Build GRCh38

View Full Product Details