Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90890252-90890783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FES
Entrez Gene ID
2242
Gene Name
FES proto-oncogene, tyrosine kinase
Gene Aliases
FPS
UniGene
Hs.7636
Gene Chromosome Location
Chr.15: 90884421-90895776 on Build GRCh38
Transcripts
NM_001143783.1 NM_001143784.1 NM_001143785.1 NM_002005.3 XM_005254880.1 XM_005254882.1 XM_017022005.1 XM_017022006.1 XM_017022007.1 XM_017022008.1 XM_017022009.1 XM_017022010.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2140362 c.1305C>T p.F435F Chr.15: 90890469-90890469 on Build GRCh38
4848641 c.1256G>A p.G419E Chr.15: 90890420-90890420 on Build GRCh38
4057941 c.1216C>T p.R406C Chr.15: 90890258-90890258 on Build GRCh38
6663221 c.1251delG p.G419fs*113 Chr.15: 90890415-90890415 on Build GRCh38
6408877 c.1235C>T p.S412L Chr.15: 90890277-90890277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897548307 Chr.15: 90890744-90890744 on Build GRCh38
rs398028344 Chr.15: 90890724-90890724 on Build GRCh38
rs750070131 Chr.15: 90890358-90890358 on Build GRCh38
rs757010190 Chr.15: 90890658-90890658 on Build GRCh38
rs985663069 Chr.15: 90890544-90890544 on Build GRCh38
rs753584672 Chr.15: 90890439-90890439 on Build GRCh38
rs191115273 Chr.15: 90890494-90890494 on Build GRCh38
rs553916547 Chr.15: 90890335-90890335 on Build GRCh38
rs753623918 Chr.15: 90890605-90890605 on Build GRCh38
rs765124279 Chr.15: 90890454-90890454 on Build GRCh38
rs398078393 Chr.15: 90890723-90890723 on Build GRCh38
rs993799850 Chr.15: 90890367-90890367 on Build GRCh38
rs913217110 Chr.15: 90890491-90890491 on Build GRCh38
rs756281605 Chr.15: 90890477-90890477 on Build GRCh38
rs372142935 Chr.15: 90890307-90890307 on Build GRCh38
rs973312856 Chr.15: 90890301-90890301 on Build GRCh38
rs761390860 Chr.15: 90890353-90890353 on Build GRCh38
rs749208680 Chr.15: 90890295-90890295 on Build GRCh38
rs911508053 Chr.15: 90890558-90890558 on Build GRCh38
rs141297480 Chr.15: 90890289-90890289 on Build GRCh38
rs905679236 Chr.15: 90890325-90890325 on Build GRCh38
rs756020799 Chr.15: 90890376-90890376 on Build GRCh38
rs768870048 Chr.15: 90890487-90890487 on Build GRCh38
rs143824045 Chr.15: 90890433-90890433 on Build GRCh38
rs767008963 Chr.15: 90890271-90890271 on Build GRCh38
rs376056732 Chr.15: 90890309-90890309 on Build GRCh38
rs376636537 Chr.15: 90890526-90890526 on Build GRCh38
rs769047058 Chr.15: 90890505-90890506 on Build GRCh38
rs146783075 Chr.15: 90890756-90890756 on Build GRCh38
rs757300050 Chr.15: 90890391-90890391 on Build GRCh38
rs977161623 Chr.15: 90890592-90890592 on Build GRCh38
rs11331647 Chr.15: 90890722-90890722 on Build GRCh38
rs776814325 Chr.15: 90890304-90890304 on Build GRCh38
rs993917568 Chr.15: 90890601-90890601 on Build GRCh38
rs764510721 Chr.15: 90890314-90890314 on Build GRCh38
rs898167551 Chr.15: 90890352-90890352 on Build GRCh38
rs965347917 Chr.15: 90890559-90890559 on Build GRCh38
rs985403005 Chr.15: 90890320-90890320 on Build GRCh38
rs200611368 Chr.15: 90890383-90890383 on Build GRCh38
rs944069746 Chr.15: 90890347-90890347 on Build GRCh38
rs199952972 Chr.15: 90890528-90890528 on Build GRCh38
rs192326321 Chr.15: 90890725-90890725 on Build GRCh38
rs779752208 Chr.15: 90890294-90890294 on Build GRCh38
rs568341108 Chr.15: 90890330-90890330 on Build GRCh38
rs964913288 Chr.15: 90890748-90890748 on Build GRCh38
rs759230524 Chr.15: 90890448-90890448 on Build GRCh38
rs765642999 Chr.15: 90890272-90890272 on Build GRCh38
rs375392546 Chr.15: 90890416-90890416 on Build GRCh38
rs546717971 Chr.15: 90890689-90890689 on Build GRCh38
rs949999341 Chr.15: 90890492-90890492 on Build GRCh38
rs752594830 Chr.15: 90890462-90890462 on Build GRCh38
rs150626827 Chr.15: 90890470-90890470 on Build GRCh38
rs754870836 Chr.15: 90890382-90890382 on Build GRCh38
rs777008953 Chr.15: 90890506-90890506 on Build GRCh38
rs754886070 Chr.15: 90890296-90890296 on Build GRCh38
rs138547963 Chr.15: 90890417-90890417 on Build GRCh38
rs779450173 Chr.15: 90890493-90890493 on Build GRCh38
rs369338125 Chr.15: 90890321-90890321 on Build GRCh38
rs143621141 Chr.15: 90890690-90890690 on Build GRCh38
rs772718520 Chr.15: 90890496-90890496 on Build GRCh38
rs755860926 Chr.15: 90890373-90890373 on Build GRCh38
rs1053605267 Chr.15: 90890659-90890659 on Build GRCh38
rs572176840 Chr.15: 90890354-90890354 on Build GRCh38
rs765944755 Chr.15: 90890434-90890434 on Build GRCh38
rs568394657 Chr.15: 90890575-90890575 on Build GRCh38
rs770158169 Chr.15: 90890308-90890308 on Build GRCh38
rs539653804 Chr.15: 90890705-90890705 on Build GRCh38
rs193084120 Chr.15: 90890726-90890726 on Build GRCh38
rs575070040 Chr.15: 90890420-90890420 on Build GRCh38
rs996780569 Chr.15: 90890749-90890749 on Build GRCh38
rs781285046 Chr.15: 90890401-90890401 on Build GRCh38
rs201841209 Chr.15: 90890277-90890277 on Build GRCh38
rs767270159 Chr.15: 90890288-90890288 on Build GRCh38
rs747088456 Chr.15: 90890297-90890297 on Build GRCh38
rs188040097 Chr.15: 90890634-90890634 on Build GRCh38
rs751041379 Chr.15: 90890284-90890284 on Build GRCh38
rs769925657 Chr.15: 90890411-90890411 on Build GRCh38
rs201659079 Chr.15: 90890483-90890483 on Build GRCh38
rs370234292 Chr.15: 90890388-90890388 on Build GRCh38
rs912046375 Chr.15: 90890293-90890293 on Build GRCh38
rs139750858 Chr.15: 90890471-90890471 on Build GRCh38
rs760090969 Chr.15: 90890510-90890510 on Build GRCh38
rs539492430 Chr.15: 90890375-90890375 on Build GRCh38
rs1019574798 Chr.15: 90890671-90890671 on Build GRCh38
rs763455736 Chr.15: 90890313-90890313 on Build GRCh38
rs1044997207 Chr.15: 90890694-90890694 on Build GRCh38
rs1015118061 Chr.15: 90890298-90890298 on Build GRCh38
rs906449559 Chr.15: 90890723-90890723 on Build GRCh38
rs147101703 Chr.15: 90890415-90890415 on Build GRCh38
rs773923402 Chr.15: 90890506-90890506 on Build GRCh38
rs143234970 Chr.15: 90890405-90890405 on Build GRCh38
rs535423583 Chr.15: 90890334-90890334 on Build GRCh38
rs763614714 Chr.15: 90890278-90890278 on Build GRCh38
rs770526007 Chr.15: 90890513-90890513 on Build GRCh38
rs915043208 Chr.15: 90890685-90890685 on Build GRCh38
rs199942096 Chr.15: 90890724-90890724 on Build GRCh38
rs200988464 Chr.15: 90890525-90890525 on Build GRCh38
rs755950934 Chr.15: 90890290-90890290 on Build GRCh38
rs564704861 Chr.15: 90890622-90890622 on Build GRCh38
rs962106027 Chr.15: 90890519-90890519 on Build GRCh38
rs760864102 Chr.15: 90890324-90890324 on Build GRCh38
rs780121381 Chr.15: 90890482-90890482 on Build GRCh38
rs182579231 Chr.15: 90890537-90890537 on Build GRCh38
rs952906062 Chr.15: 90890542-90890542 on Build GRCh38
rs368439059 Chr.15: 90890300-90890300 on Build GRCh38
rs778585609 Chr.15: 90890667-90890667 on Build GRCh38
rs756900483 Chr.15: 90890285-90890285 on Build GRCh38
rs1036448970 Chr.15: 90890743-90890743 on Build GRCh38

View Full Product Details