Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 97960622-97961124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARRDC4
Entrez Gene ID
91947
Gene Name
arrestin domain containing 4
Gene Aliases
-
UniGene
Hs.6093
Gene Chromosome Location
Chr.15: 97960703-97973838 on Build GRCh38
Transcripts
NM_183376.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6404761 c.145C>T p.P49S Chr.15: 97961006-97961006 on Build GRCh38
5668772 c.51G>A p.V17V Chr.15: 97960912-97960912 on Build GRCh38
6397535 c.92G>A p.C31Y Chr.15: 97960953-97960953 on Build GRCh38
3749319 c.9G>C p.G3G Chr.15: 97960870-97960870 on Build GRCh38
6374841 c.47G>A p.R16H Chr.15: 97960908-97960908 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756552442 Chr.15: 97961084-97961084 on Build GRCh38
rs771241062 Chr.15: 97960861-97960861 on Build GRCh38
rs200108685 Chr.15: 97960999-97960999 on Build GRCh38
rs777881690 Chr.15: 97960851-97960851 on Build GRCh38
rs768990179 Chr.15: 97961088-97961099 on Build GRCh38
rs1002680994 Chr.15: 97960733-97960733 on Build GRCh38
rs907059184 Chr.15: 97960732-97960732 on Build GRCh38
rs1019867708 Chr.15: 97961104-97961104 on Build GRCh38
rs12101554 Chr.15: 97961096-97961096 on Build GRCh38
rs780558699 Chr.15: 97961086-97961086 on Build GRCh38
rs886361009 Chr.15: 97960994-97960994 on Build GRCh38
rs562762356 Chr.15: 97960944-97960944 on Build GRCh38
rs116718710 Chr.15: 97960711-97960711 on Build GRCh38
rs575490201 Chr.15: 97960750-97960750 on Build GRCh38
rs557228522 Chr.15: 97960744-97960744 on Build GRCh38
rs983617245 Chr.15: 97960812-97960812 on Build GRCh38
rs746234806 Chr.15: 97961075-97961075 on Build GRCh38
rs937401552 Chr.15: 97960717-97960717 on Build GRCh38
rs975425426 Chr.15: 97960752-97960752 on Build GRCh38
rs111509401 Chr.15: 97960921-97960921 on Build GRCh38
rs4965046 Chr.15: 97960870-97960870 on Build GRCh38
rs570367767 Chr.15: 97960690-97960690 on Build GRCh38
rs535535676 Chr.15: 97960725-97960725 on Build GRCh38
rs774043798 Chr.15: 97961091-97961091 on Build GRCh38
rs746431220 Chr.15: 97960866-97960866 on Build GRCh38
rs760626150 Chr.15: 97960985-97960985 on Build GRCh38
rs921593329 Chr.15: 97960674-97960674 on Build GRCh38
rs773901599 Chr.15: 97960922-97960922 on Build GRCh38
rs200944490 Chr.15: 97960934-97960934 on Build GRCh38
rs898424423 Chr.15: 97960714-97960714 on Build GRCh38
rs551375554 Chr.15: 97960707-97960707 on Build GRCh38
rs768087449 Chr.15: 97961089-97961089 on Build GRCh38
rs964678771 Chr.15: 97960751-97960751 on Build GRCh38
rs557999708 Chr.15: 97960765-97960765 on Build GRCh38
rs943588460 Chr.15: 97960652-97960652 on Build GRCh38
rs748752898 Chr.15: 97961088-97961088 on Build GRCh38
rs781707610 Chr.15: 97960865-97960865 on Build GRCh38
rs751777581 Chr.15: 97961037-97961037 on Build GRCh38
rs933582943 Chr.15: 97960675-97960675 on Build GRCh38
rs192769497 Chr.15: 97960688-97960688 on Build GRCh38
rs886835139 Chr.15: 97960702-97960702 on Build GRCh38
rs758638584 Chr.15: 97960829-97960829 on Build GRCh38
rs956081873 Chr.15: 97960737-97960737 on Build GRCh38
rs781474450 Chr.15: 97961054-97961054 on Build GRCh38
rs974956521 Chr.15: 97960653-97960653 on Build GRCh38
rs527974039 Chr.15: 97961043-97961043 on Build GRCh38
rs1003120270 Chr.15: 97960937-97960937 on Build GRCh38
rs939718188 Chr.15: 97960855-97960855 on Build GRCh38
rs774912989 Chr.15: 97960900-97960900 on Build GRCh38
rs1004750826 Chr.15: 97960748-97960748 on Build GRCh38
rs773872179 Chr.15: 97960770-97960770 on Build GRCh38
rs917300111 Chr.15: 97960936-97960936 on Build GRCh38
rs114245019 Chr.15: 97960858-97960858 on Build GRCh38
rs867349406 Chr.15: 97961095-97961095 on Build GRCh38
rs190697600 Chr.15: 97960978-97960978 on Build GRCh38
rs577774581 Chr.15: 97960763-97960763 on Build GRCh38
rs1021612822 Chr.15: 97960745-97960745 on Build GRCh38
rs568194292 Chr.15: 97960722-97960722 on Build GRCh38
rs1031467133 Chr.15: 97960953-97960953 on Build GRCh38
rs954605401 Chr.15: 97961093-97961093 on Build GRCh38
rs1029147173 Chr.15: 97960838-97960838 on Build GRCh38
rs923475413 Chr.15: 97960666-97960666 on Build GRCh38
rs1043198885 Chr.15: 97960703-97960703 on Build GRCh38
rs996394435 Chr.15: 97961040-97961040 on Build GRCh38
rs914826247 Chr.15: 97960641-97960641 on Build GRCh38
rs201533515 Chr.15: 97960979-97960979 on Build GRCh38
rs1032191927 Chr.15: 97960735-97960735 on Build GRCh38
rs865946359 Chr.15: 97960734-97960734 on Build GRCh38
rs948355128 Chr.15: 97960913-97960913 on Build GRCh38
rs974492133 Chr.15: 97960803-97960803 on Build GRCh38
rs908293787 Chr.15: 97960828-97960828 on Build GRCh38
rs112100339 Chr.15: 97961092-97961092 on Build GRCh38
rs974744850 Chr.15: 97961017-97961017 on Build GRCh38
rs748806356 Chr.15: 97960915-97960915 on Build GRCh38
rs764346863 Chr.15: 97961007-97961007 on Build GRCh38
rs539768160 Chr.15: 97960757-97960757 on Build GRCh38
rs1012874504 Chr.15: 97960723-97960723 on Build GRCh38
rs202148289 Chr.15: 97961006-97961006 on Build GRCh38
rs913520544 Chr.15: 97960882-97960882 on Build GRCh38
rs752777292 Chr.15: 97961002-97961002 on Build GRCh38
rs895786992 Chr.15: 97960729-97960729 on Build GRCh38
rs115137535 Chr.15: 97960681-97960681 on Build GRCh38

View Full Product Details