Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 97965808-97966318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARRDC4
Entrez Gene ID
91947
Gene Name
arrestin domain containing 4
Gene Aliases
-
UniGene
Hs.6093
Gene Chromosome Location
Chr.15: 97960703-97973838 on Build GRCh38
Transcripts
NM_183376.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5023162 c.457G>A p.D153N Chr.15: 97965977-97965977 on Build GRCh38
6037798 c.420C>T p.Y140Y Chr.15: 97965940-97965940 on Build GRCh38
5012942 c.483G>T p.Q161H Chr.15: 97966003-97966003 on Build GRCh38
4887325 c.415C>T p.Q139* Chr.15: 97965935-97965935 on Build GRCh38
2141278 c.473G>A p.R158Q Chr.15: 97965993-97965993 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372153487 Chr.15: 97965862-97965862 on Build GRCh38
rs962793634 Chr.15: 97966003-97966003 on Build GRCh38
rs112924712 Chr.15: 97966044-97966044 on Build GRCh38
rs62023728 Chr.15: 97966089-97966089 on Build GRCh38
rs1029209912 Chr.15: 97966168-97966168 on Build GRCh38
rs761766424 Chr.15: 97965904-97965904 on Build GRCh38
rs920959779 Chr.15: 97966255-97966255 on Build GRCh38
rs933614989 Chr.15: 97966273-97966273 on Build GRCh38
rs62023729 Chr.15: 97966091-97966091 on Build GRCh38
rs112656939 Chr.15: 97966043-97966043 on Build GRCh38
rs913319980 Chr.15: 97965931-97965931 on Build GRCh38
rs201763370 Chr.15: 97965984-97965984 on Build GRCh38
rs981690092 Chr.15: 97966077-97966077 on Build GRCh38
rs769567722 Chr.15: 97965874-97965874 on Build GRCh38
rs1039402918 Chr.15: 97965905-97965905 on Build GRCh38
rs753297131 Chr.15: 97965947-97965947 on Build GRCh38
rs770173605 Chr.15: 97966071-97966072 on Build GRCh38
rs184224769 Chr.15: 97965948-97965948 on Build GRCh38
rs990298689 Chr.15: 97966119-97966119 on Build GRCh38
rs750134440 Chr.15: 97966083-97966083 on Build GRCh38
rs1020845332 Chr.15: 97966212-97966212 on Build GRCh38
rs146146673 Chr.15: 97965937-97965937 on Build GRCh38
rs397942243 Chr.15: 97965840-97965841 on Build GRCh38
rs967561098 Chr.15: 97966227-97966227 on Build GRCh38
rs987619914 Chr.15: 97966202-97966202 on Build GRCh38
rs535589673 Chr.15: 97966145-97966145 on Build GRCh38
rs573789663 Chr.15: 97965968-97965968 on Build GRCh38
rs949207185 Chr.15: 97966064-97966066 on Build GRCh38
rs775525538 Chr.15: 97965903-97965903 on Build GRCh38
rs564131950 Chr.15: 97966117-97966117 on Build GRCh38
rs76209521 Chr.15: 97966283-97966283 on Build GRCh38
rs954909986 Chr.15: 97965844-97965847 on Build GRCh38
rs143500462 Chr.15: 97965901-97965901 on Build GRCh38
rs748597087 Chr.15: 97965962-97965962 on Build GRCh38
rs754105306 Chr.15: 97965930-97965930 on Build GRCh38
rs557183663 Chr.15: 97965840-97965840 on Build GRCh38
rs539331591 Chr.15: 97965914-97965914 on Build GRCh38
rs747117138 Chr.15: 97966004-97966004 on Build GRCh38
rs777545528 Chr.15: 97965856-97965856 on Build GRCh38
rs189894715 Chr.15: 97966012-97966012 on Build GRCh38
rs200384116 Chr.15: 97965963-97965963 on Build GRCh38
rs1005010182 Chr.15: 97966078-97966078 on Build GRCh38
rs759798131 Chr.15: 97966018-97966018 on Build GRCh38
rs924840342 Chr.15: 97966114-97966114 on Build GRCh38
rs77978530 Chr.15: 97966106-97966106 on Build GRCh38
rs374602795 Chr.15: 97965942-97965942 on Build GRCh38
rs138382305 Chr.15: 97965910-97965910 on Build GRCh38
rs756910523 Chr.15: 97965969-97965969 on Build GRCh38
rs954921090 Chr.15: 97966199-97966199 on Build GRCh38
rs1051027655 Chr.15: 97966277-97966277 on Build GRCh38
rs150017247 Chr.15: 97966007-97966007 on Build GRCh38
rs745606400 Chr.15: 97965979-97965979 on Build GRCh38
rs544451290 Chr.15: 97965971-97965971 on Build GRCh38
rs773223195 Chr.15: 97965998-97965998 on Build GRCh38
rs773515883 Chr.15: 97966073-97966073 on Build GRCh38
rs763454177 Chr.15: 97966271-97966271 on Build GRCh38
rs767042279 Chr.15: 97966058-97966058 on Build GRCh38
rs781067926 Chr.15: 97965868-97965868 on Build GRCh38
rs998125293 Chr.15: 97965927-97965927 on Build GRCh38
rs375553745 Chr.15: 97965980-97965980 on Build GRCh38
rs1046615464 Chr.15: 97966067-97966075 on Build GRCh38
rs768589029 Chr.15: 97965993-97965993 on Build GRCh38
rs367878363 Chr.15: 97965992-97965992 on Build GRCh38
rs760293047 Chr.15: 97966240-97966240 on Build GRCh38
rs746718183 Chr.15: 97965860-97965860 on Build GRCh38
rs528029422 Chr.15: 97966210-97966210 on Build GRCh38
rs779829889 Chr.15: 97965991-97965991 on Build GRCh38
rs376482434 Chr.15: 97966021-97966021 on Build GRCh38
rs142999338 Chr.15: 97965925-97965925 on Build GRCh38
rs763485660 Chr.15: 97966023-97966023 on Build GRCh38
rs765693177 Chr.15: 97966019-97966019 on Build GRCh38
rs546726215 Chr.15: 97966250-97966250 on Build GRCh38
rs946526122 Chr.15: 97966217-97966217 on Build GRCh38
rs549333182 Chr.15: 97966029-97966029 on Build GRCh38
rs139298600 Chr.15: 97965871-97965871 on Build GRCh38

View Full Product Details