Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 97970506-97971016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARRDC4
Entrez Gene ID
91947
Gene Name
arrestin domain containing 4
Gene Aliases
-
UniGene
Hs.6093
Gene Chromosome Location
Chr.15: 97960703-97973838 on Build GRCh38
Transcripts
NM_183376.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1375590 c.1072T>C p.S358P Chr.15: 97970615-97970615 on Build GRCh38
376028 c.1157G>A p.C386Y Chr.15: 97970700-97970700 on Build GRCh38
3887839 c.1108C>T p.P370S Chr.15: 97970651-97970651 on Build GRCh38
5705997 c.1196C>T p.S399L Chr.15: 97970739-97970739 on Build GRCh38
5705996 c.1078G>A p.E360K Chr.15: 97970621-97970621 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140190892 Chr.15: 97970538-97970538 on Build GRCh38
rs1042467085 Chr.15: 97970926-97970926 on Build GRCh38
rs386787036 Chr.15: 97970943-97970945 on Build GRCh38
rs534921430 Chr.15: 97970643-97970643 on Build GRCh38
rs779942864 Chr.15: 97970777-97970777 on Build GRCh38
rs779994238 Chr.15: 97970677-97970677 on Build GRCh38
rs752215159 Chr.15: 97970640-97970640 on Build GRCh38
rs111892639 Chr.15: 97970943-97970944 on Build GRCh38
rs562082015 Chr.15: 97970934-97970934 on Build GRCh38
rs747189613 Chr.15: 97970694-97970694 on Build GRCh38
rs751281954 Chr.15: 97970647-97970647 on Build GRCh38
rs75444538 Chr.15: 97970873-97970873 on Build GRCh38
rs575728067 Chr.15: 97970715-97970715 on Build GRCh38
rs553267091 Chr.15: 97970687-97970687 on Build GRCh38
rs914176829 Chr.15: 97970680-97970680 on Build GRCh38
rs1047690666 Chr.15: 97970819-97970819 on Build GRCh38
rs111462734 Chr.15: 97970948-97970949 on Build GRCh38
rs750191609 Chr.15: 97970760-97970760 on Build GRCh38
rs990644882 Chr.15: 97970883-97970883 on Build GRCh38
rs755992521 Chr.15: 97970675-97970675 on Build GRCh38
rs759853831 Chr.15: 97970710-97970710 on Build GRCh38
rs760936532 Chr.15: 97970590-97970590 on Build GRCh38
rs528614364 Chr.15: 97970888-97970888 on Build GRCh38
rs2130882 Chr.15: 97970615-97970615 on Build GRCh38
rs949541197 Chr.15: 97970897-97970897 on Build GRCh38
rs771515166 Chr.15: 97970789-97970791 on Build GRCh38
rs963947570 Chr.15: 97970892-97970892 on Build GRCh38
rs746726598 Chr.15: 97970796-97970797 on Build GRCh38
rs556598670 Chr.15: 97970629-97970629 on Build GRCh38
rs950403494 Chr.15: 97970822-97970822 on Build GRCh38
rs569533756 Chr.15: 97970973-97970973 on Build GRCh38
rs200850009 Chr.15: 97970907-97970908 on Build GRCh38
rs759873347 Chr.15: 97970877-97970877 on Build GRCh38
rs890011564 Chr.15: 97970959-97970959 on Build GRCh38
rs984677704 Chr.15: 97970847-97970847 on Build GRCh38
rs763814536 Chr.15: 97970645-97970645 on Build GRCh38
rs142239479 Chr.15: 97970652-97970652 on Build GRCh38
rs751032895 Chr.15: 97970725-97970725 on Build GRCh38
rs765867939 Chr.15: 97970633-97970633 on Build GRCh38
rs1043988505 Chr.15: 97970707-97970707 on Build GRCh38
rs771485729 Chr.15: 97970567-97970568 on Build GRCh38
rs761467541 Chr.15: 97970747-97970747 on Build GRCh38
rs767097581 Chr.15: 97970759-97970759 on Build GRCh38
rs762535900 Chr.15: 97970716-97970716 on Build GRCh38
rs764476585 Chr.15: 97970580-97970580 on Build GRCh38
rs909112517 Chr.15: 97970852-97970852 on Build GRCh38
rs902511158 Chr.15: 97970654-97970654 on Build GRCh38
rs745659349 Chr.15: 97970662-97970662 on Build GRCh38
rs763761032 Chr.15: 97970724-97970724 on Build GRCh38
rs540534169 Chr.15: 97970908-97970908 on Build GRCh38
rs771071661 Chr.15: 97970705-97970705 on Build GRCh38
rs937809490 Chr.15: 97970537-97970537 on Build GRCh38
rs766837247 Chr.15: 97970603-97970603 on Build GRCh38
rs979714325 Chr.15: 97970948-97970948 on Build GRCh38
rs748246023 Chr.15: 97970555-97970555 on Build GRCh38
rs988174266 Chr.15: 97970676-97970676 on Build GRCh38
rs745421445 Chr.15: 97970689-97970690 on Build GRCh38
rs767608247 Chr.15: 97970909-97970909 on Build GRCh38
rs1056559820 Chr.15: 97970730-97970730 on Build GRCh38
rs772238321 Chr.15: 97970575-97970575 on Build GRCh38
rs774353419 Chr.15: 97970550-97970550 on Build GRCh38
rs917973410 Chr.15: 97970624-97970624 on Build GRCh38
rs143197566 Chr.15: 97970690-97970690 on Build GRCh38
rs201615841 Chr.15: 97970708-97970708 on Build GRCh38
rs1026437484 Chr.15: 97970817-97970817 on Build GRCh38
rs763127839 Chr.15: 97970810-97970810 on Build GRCh38
rs80061417 Chr.15: 97970945-97970945 on Build GRCh38
rs370057027 Chr.15: 97970656-97970656 on Build GRCh38
rs76962941 Chr.15: 97970714-97970714 on Build GRCh38
rs376594605 Chr.15: 97970578-97970578 on Build GRCh38
rs554214084 Chr.15: 97970800-97970817 on Build GRCh38
rs373539425 Chr.15: 97970701-97970701 on Build GRCh38
rs77625572 Chr.15: 97970943-97970943 on Build GRCh38
rs967702690 Chr.15: 97970947-97970947 on Build GRCh38
rs1001558092 Chr.15: 97970658-97970658 on Build GRCh38
rs1054008010 Chr.15: 97970684-97970684 on Build GRCh38
rs926833117 Chr.15: 97970986-97970986 on Build GRCh38
rs760034920 Chr.15: 97970616-97970616 on Build GRCh38
rs946923414 Chr.15: 97970702-97970702 on Build GRCh38
rs757762086 Chr.15: 97970621-97970621 on Build GRCh38
rs773812725 Chr.15: 97970770-97970770 on Build GRCh38
rs1029417735 Chr.15: 97970922-97970922 on Build GRCh38
rs917741473 Chr.15: 97970891-97970891 on Build GRCh38
rs193084708 Chr.15: 97970765-97970765 on Build GRCh38
rs555699515 Chr.15: 97970661-97970664 on Build GRCh38
rs766420293 Chr.15: 97970870-97970870 on Build GRCh38
rs887763916 Chr.15: 97970832-97970832 on Build GRCh38

View Full Product Details