Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 98439320-98439814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM169B
Entrez Gene ID
283777
Gene Name
family with sequence similarity 169 member B
Gene Aliases
-
UniGene
Hs.668070
Gene Chromosome Location
Chr.15: 98437162-98514382 on Build GRCh38
Transcripts
NM_182562.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
383158 c.533G>T p.G178V Chr.15: 98439677-98439677 on Build GRCh38
3370313 c.538G>A p.A180T Chr.15: 98439672-98439672 on Build GRCh38
966765 c.493A>G p.K165E Chr.15: 98439717-98439717 on Build GRCh38
4499708 c.548C>T p.P183L Chr.15: 98439662-98439662 on Build GRCh38
198938 c.495G>T p.K165N Chr.15: 98439715-98439715 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756301717 Chr.15: 98439675-98439675 on Build GRCh38
rs550335558 Chr.15: 98439585-98439585 on Build GRCh38
rs918991731 Chr.15: 98439588-98439588 on Build GRCh38
rs185843455 Chr.15: 98439666-98439666 on Build GRCh38
rs968240351 Chr.15: 98439544-98439544 on Build GRCh38
rs1022507156 Chr.15: 98439541-98439541 on Build GRCh38
rs373917169 Chr.15: 98439729-98439729 on Build GRCh38
rs969612830 Chr.15: 98439389-98439389 on Build GRCh38
rs1040301675 Chr.15: 98439640-98439640 on Build GRCh38
rs748538880 Chr.15: 98439636-98439636 on Build GRCh38
rs1031054448 Chr.15: 98439498-98439498 on Build GRCh38
rs761691471 Chr.15: 98439620-98439621 on Build GRCh38
rs1036271899 Chr.15: 98439434-98439434 on Build GRCh38
rs1035706921 Chr.15: 98439559-98439559 on Build GRCh38
rs563885365 Chr.15: 98439486-98439486 on Build GRCh38
rs754169231 Chr.15: 98439681-98439681 on Build GRCh38
rs996818795 Chr.15: 98439546-98439546 on Build GRCh38
rs770490323 Chr.15: 98439753-98439753 on Build GRCh38
rs930382744 Chr.15: 98439610-98439610 on Build GRCh38
rs915469248 Chr.15: 98439381-98439381 on Build GRCh38
rs898869248 Chr.15: 98439439-98439439 on Build GRCh38
rs377093710 Chr.15: 98439639-98439639 on Build GRCh38
rs776639736 Chr.15: 98439604-98439609 on Build GRCh38
rs189412134 Chr.15: 98439638-98439638 on Build GRCh38
rs892171522 Chr.15: 98439765-98439765 on Build GRCh38
rs1028254660 Chr.15: 98439551-98439551 on Build GRCh38
rs145179819 Chr.15: 98439507-98439507 on Build GRCh38
rs762738868 Chr.15: 98439665-98439665 on Build GRCh38
rs374821353 Chr.15: 98439416-98439416 on Build GRCh38
rs200050409 Chr.15: 98439679-98439679 on Build GRCh38
rs1044718064 Chr.15: 98439448-98439448 on Build GRCh38
rs4129470 Chr.15: 98439398-98439398 on Build GRCh38
rs539511116 Chr.15: 98439629-98439629 on Build GRCh38
rs566162686 Chr.15: 98439630-98439630 on Build GRCh38
rs367979023 Chr.15: 98439760-98439760 on Build GRCh38
rs199951416 Chr.15: 98439702-98439702 on Build GRCh38
rs779204951 Chr.15: 98439688-98439688 on Build GRCh38
rs756352383 Chr.15: 98439621-98439621 on Build GRCh38
rs888996751 Chr.15: 98439450-98439450 on Build GRCh38
rs765271652 Chr.15: 98439735-98439737 on Build GRCh38
rs750487100 Chr.15: 98439672-98439672 on Build GRCh38
rs780454801 Chr.15: 98439623-98439623 on Build GRCh38
rs531203131 Chr.15: 98439560-98439560 on Build GRCh38
rs934060298 Chr.15: 98439380-98439380 on Build GRCh38
rs181551150 Chr.15: 98439364-98439364 on Build GRCh38
rs757356258 Chr.15: 98439614-98439614 on Build GRCh38
rs562470161 Chr.15: 98439457-98439457 on Build GRCh38
rs770543637 Chr.15: 98439642-98439642 on Build GRCh38
rs763273905 Chr.15: 98439410-98439410 on Build GRCh38
rs1056215923 Chr.15: 98439732-98439732 on Build GRCh38
rs545898105 Chr.15: 98439485-98439485 on Build GRCh38
rs769985847 Chr.15: 98439386-98439386 on Build GRCh38
rs779090781 Chr.15: 98439720-98439720 on Build GRCh38
rs759218435 Chr.15: 98439659-98439659 on Build GRCh38
rs746504588 Chr.15: 98439752-98439752 on Build GRCh38
rs34036460 Chr.15: 98439715-98439716 on Build GRCh38
rs778292963 Chr.15: 98439744-98439744 on Build GRCh38
rs184136890 Chr.15: 98439493-98439493 on Build GRCh38
rs766651975 Chr.15: 98439506-98439506 on Build GRCh38
rs752638480 Chr.15: 98439609-98439609 on Build GRCh38
rs8043073 Chr.15: 98439446-98439446 on Build GRCh38
rs751394462 Chr.15: 98439519-98439519 on Build GRCh38
rs199994723 Chr.15: 98439620-98439620 on Build GRCh38
rs149145816 Chr.15: 98439573-98439573 on Build GRCh38
rs776318697 Chr.15: 98439658-98439658 on Build GRCh38
rs975348735 Chr.15: 98439353-98439353 on Build GRCh38
rs1002190827 Chr.15: 98439730-98439732 on Build GRCh38
rs779460817 Chr.15: 98439634-98439634 on Build GRCh38
rs180911515 Chr.15: 98439660-98439660 on Build GRCh38
rs575383659 Chr.15: 98439433-98439433 on Build GRCh38
rs538480419 Chr.15: 98439465-98439465 on Build GRCh38
rs867075219 Chr.15: 98439780-98439780 on Build GRCh38
rs1048779282 Chr.15: 98439449-98439449 on Build GRCh38
rs986889678 Chr.15: 98439616-98439616 on Build GRCh38
rs911708962 Chr.15: 98439432-98439432 on Build GRCh38

View Full Product Details