Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 99971459-99971961 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS17
Entrez Gene ID
170691
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 17
Gene Aliases
-
UniGene
Hs.513200
Gene Chromosome Location
Chr.15: 99971438-100342924 on Build GRCh38
Transcripts
NM_139057.3 XM_005254872.3 XM_011521312.2 XM_017021973.1 XM_017021975.1 XM_017021976.1 XM_017021978.1 XM_017021979.1 XM_017021980.1

SNP Detail

SNP ID Chromosome Location
rs13329612 Chr.15: 99971701-99971701 on Build GRCh38
rs768332296 Chr.15: 99971513-99971513 on Build GRCh38
rs10468182 Chr.15: 99971632-99971632 on Build GRCh38
rs564165754 Chr.15: 99971756-99971756 on Build GRCh38
rs994363579 Chr.15: 99971750-99971750 on Build GRCh38
rs774237671 Chr.15: 99971585-99971585 on Build GRCh38
rs979037516 Chr.15: 99971682-99971682 on Build GRCh38
rs1042815473 Chr.15: 99971492-99971492 on Build GRCh38
rs994958939 Chr.15: 99971645-99971645 on Build GRCh38
rs566216613 Chr.15: 99971712-99971712 on Build GRCh38
rs778520282 Chr.15: 99971801-99971801 on Build GRCh38
rs886050951 Chr.15: 99971870-99971870 on Build GRCh38
rs183671964 Chr.15: 99971824-99971824 on Build GRCh38
rs778254319 Chr.15: 99971736-99971736 on Build GRCh38
rs754478048 Chr.15: 99971804-99971804 on Build GRCh38
rs574839498 Chr.15: 99971502-99971502 on Build GRCh38
rs573353500 Chr.15: 99971922-99971922 on Build GRCh38
rs972171040 Chr.15: 99971838-99971838 on Build GRCh38
rs543559051 Chr.15: 99971774-99971776 on Build GRCh38
rs767798008 Chr.15: 99971671-99971671 on Build GRCh38
rs142112505 Chr.15: 99971762-99971762 on Build GRCh38
rs1056062786 Chr.15: 99971881-99971881 on Build GRCh38
rs529849561 Chr.15: 99971765-99971765 on Build GRCh38
rs973091308 Chr.15: 99971542-99971543 on Build GRCh38
rs189002965 Chr.15: 99971782-99971782 on Build GRCh38
rs960178419 Chr.15: 99971503-99971503 on Build GRCh38
rs879898681 Chr.15: 99971761-99971761 on Build GRCh38
rs1042485479 Chr.15: 99971597-99971597 on Build GRCh38
rs990665106 Chr.15: 99971663-99971663 on Build GRCh38
rs930480209 Chr.15: 99971849-99971849 on Build GRCh38
rs892131014 Chr.15: 99971887-99971887 on Build GRCh38
rs946753220 Chr.15: 99971590-99971590 on Build GRCh38
rs940789616 Chr.15: 99971833-99971833 on Build GRCh38
rs8041971 Chr.15: 99971861-99971861 on Build GRCh38
rs568094335 Chr.15: 99971911-99971912 on Build GRCh38
rs552244538 Chr.15: 99971541-99971541 on Build GRCh38
rs375704403 Chr.15: 99971786-99971786 on Build GRCh38
rs1043451739 Chr.15: 99971524-99971524 on Build GRCh38
rs189702378 Chr.15: 99971650-99971650 on Build GRCh38
rs566389317 Chr.15: 99971551-99971551 on Build GRCh38
rs879249123 Chr.15: 99971723-99971723 on Build GRCh38
rs138099553 Chr.15: 99971930-99971930 on Build GRCh38
rs561880684 Chr.15: 99971662-99971663 on Build GRCh38
rs540696412 Chr.15: 99971923-99971923 on Build GRCh38
rs140048103 Chr.15: 99971787-99971787 on Build GRCh38
rs923909690 Chr.15: 99971603-99971603 on Build GRCh38
rs879359334 Chr.15: 99971649-99971649 on Build GRCh38
rs920164077 Chr.15: 99971933-99971934 on Build GRCh38
rs183457005 Chr.15: 99971655-99971655 on Build GRCh38
rs370402646 Chr.15: 99971823-99971836 on Build GRCh38
rs575688305 Chr.15: 99971687-99971687 on Build GRCh38
rs537007053 Chr.15: 99971523-99971523 on Build GRCh38
rs773361337 Chr.15: 99971640-99971640 on Build GRCh38
rs987473305 Chr.15: 99971814-99971814 on Build GRCh38
rs911999983 Chr.15: 99971718-99971718 on Build GRCh38
rs1029933898 Chr.15: 99971504-99971504 on Build GRCh38
rs1038983671 Chr.15: 99971637-99971637 on Build GRCh38
rs1047253649 Chr.15: 99971829-99971829 on Build GRCh38
rs1007053202 Chr.15: 99971635-99971635 on Build GRCh38
rs956426836 Chr.15: 99971657-99971657 on Build GRCh38
rs932095692 Chr.15: 99971542-99971542 on Build GRCh38
rs151153777 Chr.15: 99971783-99971783 on Build GRCh38
rs111235216 Chr.15: 99971820-99971820 on Build GRCh38
rs894781158 Chr.15: 99971851-99971851 on Build GRCh38
rs981819129 Chr.15: 99971499-99971499 on Build GRCh38
rs1036176176 Chr.15: 99971617-99971617 on Build GRCh38
rs535694449 Chr.15: 99971664-99971664 on Build GRCh38
rs188195772 Chr.15: 99971867-99971867 on Build GRCh38
rs959315931 Chr.15: 99971659-99971659 on Build GRCh38
rs1020410350 Chr.15: 99971669-99971669 on Build GRCh38
rs994729962 Chr.15: 99971622-99971622 on Build GRCh38
rs374262051 Chr.15: 99971618-99971618 on Build GRCh38
rs772067210 Chr.15: 99971621-99971621 on Build GRCh38
rs890324345 Chr.15: 99971589-99971589 on Build GRCh38
rs569280011 Chr.15: 99971872-99971872 on Build GRCh38
rs145561763 Chr.15: 99971891-99971891 on Build GRCh38
rs1051832947 Chr.15: 99971565-99971565 on Build GRCh38
rs913486375 Chr.15: 99971518-99971518 on Build GRCh38
rs116467016 Chr.15: 99971680-99971680 on Build GRCh38
rs933968440 Chr.15: 99971688-99971688 on Build GRCh38
rs1007090109 Chr.15: 99971528-99971528 on Build GRCh38
rs927901243 Chr.15: 99971562-99971562 on Build GRCh38
rs902519864 Chr.15: 99971494-99971494 on Build GRCh38
rs571471522 Chr.15: 99971787-99971801 on Build GRCh38
rs527248314 Chr.15: 99971895-99971896 on Build GRCh38
rs911475873 Chr.15: 99971704-99971704 on Build GRCh38
rs1047449726 Chr.15: 99971753-99971753 on Build GRCh38
rs903805002 Chr.15: 99971868-99971868 on Build GRCh38
rs1038623544 Chr.15: 99971713-99971713 on Build GRCh38
rs981963339 Chr.15: 99971550-99971550 on Build GRCh38
rs8037666 Chr.15: 99971874-99971874 on Build GRCh38
rs1036614146 Chr.15: 99971740-99971740 on Build GRCh38
rs114391483 Chr.15: 99971757-99971757 on Build GRCh38
rs141079260 Chr.15: 99971816-99971829 on Build GRCh38
rs998405702 Chr.15: 99971501-99971501 on Build GRCh38
rs546576510 Chr.15: 99971491-99971493 on Build GRCh38
rs778323773 Chr.15: 99971816-99971816 on Build GRCh38
rs946724687 Chr.15: 99971487-99971490 on Build GRCh38
rs759842799 Chr.15: 99971665-99971665 on Build GRCh38
rs370664837 Chr.15: 99971768-99971768 on Build GRCh38
rs886050950 Chr.15: 99971524-99971524 on Build GRCh38
rs955202180 Chr.15: 99971880-99971880 on Build GRCh38
rs953586992 Chr.15: 99971806-99971806 on Build GRCh38
rs1026359448 Chr.15: 99971548-99971548 on Build GRCh38
rs926558850 Chr.15: 99971873-99971873 on Build GRCh38
rs773407803 Chr.15: 99971746-99971747 on Build GRCh38
rs529656441 Chr.15: 99971612-99971612 on Build GRCh38
rs1009757490 Chr.15: 99971890-99971890 on Build GRCh38
rs191759654 Chr.15: 99971797-99971797 on Build GRCh38
rs898665324 Chr.15: 99971641-99971642 on Build GRCh38
rs761921447 Chr.15: 99971576-99971577 on Build GRCh38
rs766691184 Chr.15: 99971779-99971793 on Build GRCh38
rs10468183 Chr.15: 99971842-99971842 on Build GRCh38
rs903573257 Chr.15: 99971919-99971919 on Build GRCh38
rs921951679 Chr.15: 99971691-99971691 on Build GRCh38
rs1044969322 Chr.15: 99971764-99971764 on Build GRCh38

View Full Product Details