Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 99973128-99973632 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS17
Entrez Gene ID
170691
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 17
Gene Aliases
-
UniGene
Hs.513200
Gene Chromosome Location
Chr.15: 99971438-100342924 on Build GRCh38
Transcripts
NM_139057.3 XM_005254872.3 XM_011521312.2 XM_017021973.1 XM_017021975.1 XM_017021976.1 XM_017021978.1 XM_017021979.1 XM_017021980.1 XM_017021982.1 XM_017021983.1

SNP Detail

SNP ID Chromosome Location
rs2727198 Chr.15: 99973517-99973517 on Build GRCh38
rs555793439 Chr.15: 99973338-99973338 on Build GRCh38
rs1004326206 Chr.15: 99973494-99973494 on Build GRCh38
rs549608045 Chr.15: 99973546-99973546 on Build GRCh38
rs2581359 Chr.15: 99973502-99973502 on Build GRCh38
rs561561821 Chr.15: 99973158-99973158 on Build GRCh38
rs949930618 Chr.15: 99973402-99973402 on Build GRCh38
rs571008658 Chr.15: 99973600-99973600 on Build GRCh38
rs146418744 Chr.15: 99973320-99973320 on Build GRCh38
rs893963425 Chr.15: 99973274-99973274 on Build GRCh38
rs951529025 Chr.15: 99973337-99973337 on Build GRCh38
rs886050958 Chr.15: 99973184-99973184 on Build GRCh38
rs953386260 Chr.15: 99973444-99973444 on Build GRCh38
rs143967511 Chr.15: 99973222-99973222 on Build GRCh38
rs543905312 Chr.15: 99973168-99973168 on Build GRCh38
rs981748454 Chr.15: 99973588-99973588 on Build GRCh38
rs1054363056 Chr.15: 99973406-99973406 on Build GRCh38
rs753577015 Chr.15: 99973299-99973299 on Build GRCh38
rs892958505 Chr.15: 99973302-99973302 on Build GRCh38
rs527611343 Chr.15: 99973303-99973303 on Build GRCh38
rs985453208 Chr.15: 99973201-99973203 on Build GRCh38
rs1054603073 Chr.15: 99973438-99973438 on Build GRCh38
rs982642681 Chr.15: 99973189-99973189 on Build GRCh38
rs971379245 Chr.15: 99973581-99973581 on Build GRCh38
rs934452330 Chr.15: 99973436-99973436 on Build GRCh38
rs957205433 Chr.15: 99973155-99973155 on Build GRCh38
rs924330482 Chr.15: 99973593-99973593 on Build GRCh38
rs1054393704 Chr.15: 99973296-99973296 on Build GRCh38
rs949743426 Chr.15: 99973267-99973267 on Build GRCh38
rs565379470 Chr.15: 99973199-99973199 on Build GRCh38
rs964027320 Chr.15: 99973151-99973151 on Build GRCh38
rs1055618962 Chr.15: 99973219-99973219 on Build GRCh38
rs538240846 Chr.15: 99973412-99973412 on Build GRCh38
rs902942627 Chr.15: 99973268-99973268 on Build GRCh38
rs57279107 Chr.15: 99973413-99973413 on Build GRCh38
rs180868548 Chr.15: 99973442-99973442 on Build GRCh38
rs886050962 Chr.15: 99973386-99973386 on Build GRCh38
rs2581363 Chr.15: 99973159-99973159 on Build GRCh38
rs941320893 Chr.15: 99973215-99973215 on Build GRCh38
rs559730861 Chr.15: 99973521-99973521 on Build GRCh38
rs886050959 Chr.15: 99973337-99973337 on Build GRCh38
rs900062630 Chr.15: 99973315-99973315 on Build GRCh38
rs915604850 Chr.15: 99973227-99973227 on Build GRCh38
rs563070394 Chr.15: 99973439-99973439 on Build GRCh38
rs192315551 Chr.15: 99973601-99973601 on Build GRCh38
rs889232446 Chr.15: 99973597-99973597 on Build GRCh38
rs886050960 Chr.15: 99973348-99973348 on Build GRCh38
rs933172667 Chr.15: 99973548-99973548 on Build GRCh38
rs570538855 Chr.15: 99973228-99973230 on Build GRCh38
rs867101754 Chr.15: 99973414-99973414 on Build GRCh38
rs1050519680 Chr.15: 99973595-99973595 on Build GRCh38
rs188429259 Chr.15: 99973416-99973416 on Build GRCh38
rs552737711 Chr.15: 99973545-99973545 on Build GRCh38
rs2581362 Chr.15: 99973319-99973319 on Build GRCh38
rs11277519 Chr.15: 99973383-99973388 on Build GRCh38
rs116585016 Chr.15: 99973260-99973260 on Build GRCh38
rs909490264 Chr.15: 99973326-99973326 on Build GRCh38
rs971322468 Chr.15: 99973517-99973517 on Build GRCh38
rs962309056 Chr.15: 99973501-99973501 on Build GRCh38
rs10549565 Chr.15: 99973168-99973171 on Build GRCh38
rs1018551132 Chr.15: 99973309-99973309 on Build GRCh38
rs999004930 Chr.15: 99973271-99973271 on Build GRCh38
rs201905982 Chr.15: 99973388-99973388 on Build GRCh38
rs576659910 Chr.15: 99973452-99973452 on Build GRCh38
rs549233393 Chr.15: 99973528-99973528 on Build GRCh38
rs887086825 Chr.15: 99973475-99973475 on Build GRCh38
rs946888861 Chr.15: 99973440-99973440 on Build GRCh38
rs886050963 Chr.15: 99973436-99973436 on Build GRCh38
rs2727196 Chr.15: 99973324-99973324 on Build GRCh38
rs867738416 Chr.15: 99973417-99973417 on Build GRCh38
rs2727197 Chr.15: 99973476-99973476 on Build GRCh38
rs962502639 Chr.15: 99973345-99973345 on Build GRCh38
rs886050964 Chr.15: 99973552-99973552 on Build GRCh38
rs1022717822 Chr.15: 99973552-99973552 on Build GRCh38
rs773298499 Chr.15: 99973154-99973154 on Build GRCh38
rs762728863 Chr.15: 99973382-99973383 on Build GRCh38
rs998834909 Chr.15: 99973450-99973450 on Build GRCh38
rs1028958390 Chr.15: 99973323-99973323 on Build GRCh38
rs886050961 Chr.15: 99973378-99973378 on Build GRCh38
rs867937151 Chr.15: 99973277-99973277 on Build GRCh38
rs766519775 Chr.15: 99973275-99973275 on Build GRCh38
rs952567865 Chr.15: 99973603-99973611 on Build GRCh38
rs879713910 Chr.15: 99973386-99973391 on Build GRCh38
rs949733036 Chr.15: 99973248-99973248 on Build GRCh38
rs2581358 Chr.15: 99973520-99973520 on Build GRCh38
rs909355756 Chr.15: 99973224-99973224 on Build GRCh38
rs187416809 Chr.15: 99973522-99973522 on Build GRCh38
rs915845535 Chr.15: 99973370-99973370 on Build GRCh38
rs909697622 Chr.15: 99973191-99973191 on Build GRCh38
rs571021181 Chr.15: 99973341-99973341 on Build GRCh38
rs377343119 Chr.15: 99973272-99973272 on Build GRCh38
rs199980958 Chr.15: 99973384-99973384 on Build GRCh38
rs945910346 Chr.15: 99973544-99973544 on Build GRCh38
rs768799371 Chr.15: 99973237-99973237 on Build GRCh38
rs761195810 Chr.15: 99973478-99973478 on Build GRCh38
rs991251949 Chr.15: 99973361-99973361 on Build GRCh38
rs141144986 Chr.15: 99973443-99973443 on Build GRCh38
rs560851767 Chr.15: 99973531-99973531 on Build GRCh38
rs1016462751 Chr.15: 99973451-99973451 on Build GRCh38
rs940879390 Chr.15: 99973226-99973226 on Build GRCh38

View Full Product Details